Product References
Eosinophils protect against pulmonary hypertension through 14-HDHA and 17-HDHA.
The European respiratory journal
Shu T,Zhang J,Zhou Y,Chen Z,Li J,Tang Q,Lei W,Xing Y,Wang J,Wang C
250-01 was used in In vitro experiments to show that in PH development without external inflammatory stimulus, peripheral blood exhibits a low EOS level, in which EOS play a protective role through suppressing perivascular inflammation and maintaining PASMC homeostasis via 14/17-HDHA.
Wed Mar 01 00:00:00 EST 2023
Aiolos regulates eosinophil migration into tissues.
Mucosal immunology
Felton JM,Bouffi C,Schwartz JT,Schollaert KL,Malik A,Vallabh S,Wronowski B,Magier AZ,Merlin L,Barski A,Weirauch MT,Fulkerson PC,Rothenberg ME
Mon Nov 01 00:00:00 EDT 2021
Aiolos regulates eosinophil migration into tissues.
Mucosal immunology
Felton JM,Bouffi C,Schwartz JT,Schollaert KL,Malik A,Vallabh S,Wronowski B,Magier AZ,Merlin L,Barski A,Weirauch MT,Fulkerson PC,Rothenberg ME
Mon Nov 01 00:00:00 EDT 2021
Myeloid-IL4Rα is an indispensable link in IL-33-ILCs-IL-13-IL4Rα axis of eosinophil recruitment in murine lungs.
Scientific reports
Patial S,Lewis BW,Vo T,Choudhary I,Paudel K,Mao Y,Singamsetty D,Brombacher F,Saini Y
Thu Jul 29 00:00:00 EDT 2021
Regulation of Eosinophil Recruitment and Allergic Airway Inflammation by Tropomyosin Receptor Kinase A.
Journal of immunology (Baltimore, Md. : 1950)
Dileepan M,Ge XN,Bastan I,Greenberg YG,Liang Y,Sriramarao P,Rao SP
Sat Feb 01 00:00:00 EST 2020
Inflammaging impairs peripheral nerve maintenance and regeneration.
Aging cell
Büttner R,Schulz A,Reuter M,Akula AK,Mindos T,Carlstedt A,Riecken LB,Baader SL,Bauer R,Morrison H
Sat Dec 01 00:00:00 EST 2018
Physiological function of phospholipase D2 in anti-tumor immunity: regulation of CD8+ T lymphocyte proliferation.
Scientific reports
Ngo Thai Bich V,Hongu T,Miura Y,Katagiri N,Ohbayashi N,Yamashita-Kanemaru Y,Shibuya A,Funakoshi Y,Kanaho Y
Thu Apr 19 00:00:00 EDT 2018
Yeast surface display identifies a family of evasins from ticks with novel polyvalent CC chemokine-binding activities.
Scientific reports
Singh K,Davies G,Alenazi Y,Eaton JRO,Kawamura A,Bhattacharya S
Tue Jun 27 00:00:00 EDT 2017
Systematic Screening of Chemokines to Identify Candidates to Model and Create Ectopic Lymph Node Structures for Cancer Immunotherapy.
Scientific reports
Yagawa Y,Robertson-Tessi M,Zhou SL,Anderson ARA,Mulé JJ,Mailloux AW
Wed Nov 22 00:00:00 EST 2017
Regulation of eosinophilia and allergic airway inflammation by the glycan-binding protein galectin-1.
Proceedings of the National Academy of Sciences of the United States of America
Ge XN,Ha SG,Greenberg YG,Rao A,Bastan I,Blidner AG,Rao SP,Rabinovich GA,Sriramarao P
Tue Aug 16 00:00:00 EDT 2016
5-lipoxygenase-dependent recruitment of neutrophils and macrophages by eotaxin-stimulated murine eosinophils.
Mediators of inflammation
Luz RA,Xavier-Elsas P,de Luca B,Masid-de-Brito D,Cauduro PS,Arcanjo LC,dos Santos AC,de Oliveira IC,Gaspar-Elsas MI
250-01 was used in In vivo experiments to investigate eosinophil roles in microbial defence in mouse models.
Mon Dec 15 00:00:00 EST 2014
Eosinophil adoptive transfer system to directly evaluate pulmonary eosinophil trafficking in vivo.
Proceedings of the National Academy of Sciences of the United States of America
Wen T,Besse JA,Mingler MK,Fulkerson PC,Rothenberg ME
Tue Apr 09 00:00:00 EDT 2013
Adam8 limits the development of allergic airway inflammation in mice.
Journal of immunology (Baltimore, Md. : 1950)
Knolle MD,Nakajima T,Hergrueter A,Gupta K,Polverino F,Craig VJ,Fyfe SE,Zahid M,Permaul P,Cernadas M,Montano G,Tesfaigzi Y,Sholl L,Kobzik L,Israel E,Owen CA
Sat Jun 15 00:00:00 EDT 2013
Regulation of eosinophil trafficking by SWAP-70 and its role in allergic airway inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Bahaie NS,Hosseinkhani MR,Ge XN,Kang BN,Ha SG,Blumenthal MS,Jessberger R,Rao SP,Sriramarao P
Wed Feb 01 00:00:00 EST 2012
PTP1B deficiency exacerbates inflammation and accelerates leukocyte trafficking in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Berdnikovs S,Pavlov VI,Abdala-Valencia H,McCary CA,Klumpp DJ,Tremblay ML,Cook-Mills JM
Sun Jan 15 00:00:00 EST 2012
PTP1B deficiency exacerbates inflammation and accelerates leukocyte trafficking in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Berdnikovs S,Pavlov VI,Abdala-Valencia H,McCary CA,Klumpp DJ,Tremblay ML,Cook-Mills JM
Sun Jan 15 00:00:00 EST 2012
IL-4 engagement of the type I IL-4 receptor complex enhances mouse eosinophil migration to eotaxin-1 in vitro.
PloS one
Heller NM,Gwinn WM,Donnelly RP,Constant SL,Keegan AD
Fri Jan 04 00:00:00 EST 2013
N-Glycans differentially regulate eosinophil and neutrophil recruitment during allergic airway inflammation.
The Journal of biological chemistry
Bahaie NS,Kang BN,Frenzel EM,Hosseinkhani MR,Ge XN,Greenberg Y,Ha SG,Demetriou M,Rao SP,Sriramarao P
Fri Nov 04 00:00:00 EDT 2011
A novel recombinant fusion protein encoding a 20-amino acid residue of the third extracellular (E3) domain of CCR2 neutralizes the biological activity of CCL2.
Journal of immunology (Baltimore, Md. : 1950)
Izhak L,Wildbaum G,Zohar Y,Anunu R,Klapper L,Elkeles A,Seagal J,Yefenof E,Ayalon-Soffer M,Karin N
Wed Jul 01 00:00:00 EDT 2009
Structure and function of A41, a vaccinia virus chemokine binding protein.
PLoS pathogens
Bahar MW,Kenyon JC,Putz MM,Abrescia NG,Pease JE,Wise EL,Stuart DI,Smith GL,Grimes JM
Tue Jan 01 00:00:00 EST 2008
CXCL10 blockade protects mice from cyclophosphamide-induced cystitis.
Journal of immune based therapies and vaccines
Sakthivel SK,Singh UP,Singh S,Taub DD,Novakovic KR,Lillard JW
Tue Oct 28 00:00:00 EDT 2008
Eotaxin selectively binds heparin. An interaction that protects eotaxin from proteolysis and potentiates chemotactic activity in vivo.
The Journal of biological chemistry
Ellyard JI,Simson L,Bezos A,Johnston K,Freeman C,Parish CR
Fri May 18 00:00:00 EDT 2007
Leukotriene B4, an activation product of mast cells, is a chemoattractant for their progenitors.
The Journal of experimental medicine
Weller CL,Collington SJ,Brown JK,Miller HR,Al-Kashi A,Clark P,Jose PJ,Hartnell A,Williams TJ
Mon Jun 20 00:00:00 EDT 2005
Negative regulation of eosinophil recruitment to the lung by the chemokine monokine induced by IFN-gamma (Mig, CXCL9).
Proceedings of the National Academy of Sciences of the United States of America
Fulkerson PC,Zimmermann N,Brandt EB,Muntel EE,Doepker MP,Kavanaugh JL,Mishra A,Witte DP,Zhang H,Farber JM,Yang M,Foster PS,Rothenberg ME
Tue Feb 17 00:00:00 EST 2004
Hematopoietic stem cells are uniquely selective in their migratory response to chemokines.
The Journal of experimental medicine
Wright DE,Bowman EP,Wagers AJ,Butcher EC,Weissman IL
Mon May 06 00:00:00 EDT 2002
Stem cell factor can stimulate the formation of eosinophils by two types of murine eosinophil progenitor cells.
Stem cells (Dayton, Ohio)
Metcalf D,Mifsud S,Di Rago L
Wed Mar 12 00:00:00 EST 2003
CCL7 and CXCL10 orchestrate oxidative stress-induced neutrophilic lung inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Michalec L,Choudhury BK,Postlethwait E,Wild JS,Alam R,Lett-Brown M,Sur S
Tue Jan 15 00:00:00 EST 2002
A pathological function for eotaxin and eosinophils in eosinophilic gastrointestinal inflammation.
Nature immunology
Hogan SP,Mishra A,Brandt EB,Royalty MP,Pope SM,Zimmermann N,Foster PS,Rothenberg ME
Sun Apr 01 00:00:00 EST 2001
BLTR mediates leukotriene B(4)-induced chemotaxis and adhesion and plays a dominant role in eosinophil accumulation in a murine model of peritonitis.
The Journal of experimental medicine
Tager AM,Dufour JH,Goodarzi K,Bercury SD,von Andrian UH,Luster AD
Mon Aug 07 00:00:00 EDT 2000
Developmental switches in chemokine response profiles during B cell differentiation and maturation.
The Journal of experimental medicine
Bowman EP,Campbell JJ,Soler D,Dong Z,Manlongat N,Picarella D,Hardy RR,Butcher EC
Mon Apr 17 00:00:00 EDT 2000
The chemotactic cytokine eotaxin acts as a granulocyte-macrophage colony-stimulating factor during lung inflammation.
Blood
Peled A,Gonzalo JA,Lloyd C,Gutierrez-Ramos JC
Sun Mar 15 00:00:00 EST 1998
Murine monocyte chemoattractant protein (MCP)-5: a novel CC chemokine that is a structural and functional homologue of human MCP-1.
The Journal of experimental medicine
Sarafi MN,Garcia-Zepeda EA,MacLean JA,Charo IF,Luster AD
Mon Jan 06 00:00:00 EST 1997