Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recombinant Murine TARC (CCL17) is an 7.9 kDa protein containing 70 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABCD-2; C-C motif chemokine; C-C motif chemokine 17; CC chemokine TARC; small inducible cytokine subfamily A17; Small-inducible cytokine A17; Thymus and activation-regulated chemokine
Gene Aliases: Abcd-2; Ccl17; Scya17; Scya17l; Tarc
UniProt ID: (Mouse) Q9WUZ6
Entrez Gene ID: (Mouse) 20295
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support