Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recombinant Murine Eotaxin-2 is a 10.3 kDa protein containing 93 amino acid residues.
This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 24; CC chemokine CCL24; Eosinophil chemotactic protein 2; Eotaxin-2; Eotaxin2; small inducible cytokine A24; Small-inducible cytokine A24
Gene Aliases: Ccl24; CKb-6; MPIF-2; Scya24
UniProt ID: (Mouse) Q9JKC0
Entrez Gene ID: (Mouse) 56221
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support