Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
It is recommended that the protein be aliquoted for optimal storage.
Reconstituted with sterile deionized water to 0.25mg/mL.
Concentration is measured by UV.
This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD85k; glycoprotein 49 B; leukocyte immunoglobulin-like receptor subfamily B member 4; Leukocyte immunoglobulin-like receptor subfamily B member 4A; Mast cell surface glycoprotein Gp49B
Gene Aliases: CD85K; gp49; Gp49b; HM18; ILT3; Lilrb4a; LIR-5
UniProt ID: (Mouse) Q64281
Entrez Gene ID: (Mouse) 14728
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support