Product References
Glomerulonephritis and autoimmune vasculitis are independent of P2RX7 but may depend on alternative inflammasome pathways.
The Journal of pathology
Prendecki M,McAdoo SP,Turner-Stokes T,Garcia-Diaz A,Orriss I,Woollard KJ,Behmoaras J,Cook HT,Unwin R,Pusey CD,Aitman TJ,Tam FW
Fri Jul 01 00:00:00 EDT 2022
Distinct Effects of Heparin and Interleukin-4 Functionalization on Macrophage Polarization and In Situ Arterial Tissue Regeneration Using Resorbable Supramolecular Vascular Grafts in Rats.
Advanced healthcare materials
Bonito V,Koch SE,Krebber MM,Carvajal-Berrio DA,Marzi J,Duijvelshoff R,Lurier EB,Buscone S,Dekker S,de Jong SMJ,Mes T,Vaessen KRD,Brauchle EM,Bosman AW,Schenke-Layland K,Verhaar MC,Dankers PYW,Smits AIPM,Bouten CVC
Mon Nov 01 00:00:00 EDT 2021
Exosomes Derived from M2 Macrophages Exert a Therapeutic Effect via Inhibition of the PI3K/AKT/mTOR Pathway in Rats with Knee Osteoarthritic.
BioMed research international
Da-Wa ZX,Jun M,Chao-Zheng L,Sen-Lin Y,Chuan L,De-Chun L,Zu-Nan D,Hong-Tao Z,Shu-Qing W,Xian-Wei P,Wenbo L,Ke-Wen L
Thu Jan 20 00:00:00 EST 2022
Carbamylated erythropoietin regulates immune responses and promotes long-term kidney allograft survival through activation of PI3K/AKT signaling.
Signal transduction and targeted therapy
Na N,Zhao D,Zhang J,Wu J,Miao B,Li H,Luo Y,Tang Z,Zhang W,Bellanti JA,Zheng SG
Wed Sep 16 00:00:00 EDT 2020
C-type lectin receptors Mcl and Mincle control development of multiple sclerosis-like neuroinflammation.
The Journal of clinical investigation
N'diaye M,Brauner S,Flytzani S,Kular L,Warnecke A,Adzemovic MZ,Piket E,Min JH,Edwards W,Mela F,Choi HY,Magg V,James T,Linden M,Reichardt HM,Daws MR,van Horssen J,Kockum I,Harris RA,Olsson T,Guerreiro-Cacais AO,Jagodic M
Mon Feb 03 00:00:00 EST 2020
MHC/class-II-positive cells inhibit corticosterone of adrenal gland cells in experimental arthritis: a role for IL-1β, IL-18, and the inflammasome.
Scientific reports
Stangl H,Krammetsvogl A,Lesiak M,Wolff C,Straub RH
Tue Oct 13 00:00:00 EDT 2020
Beneficial effects of IL-4 and IL-6 on rat neonatal target cardiac cells.
Scientific reports
Zogbi C,Oliveira NC,Levy D,Bydlowski SP,Bassaneze V,Neri EA,Krieger JE
Thu Jul 23 00:00:00 EDT 2020
C1q/TNF-related protein-9 promotes macrophage polarization and improves cardiac dysfunction after myocardial infarction.
Journal of cellular physiology
Liu M,Yin L,Li W,Hu J,Wang H,Ye B,Tang Y,Huang C
Thu Aug 01 00:00:00 EDT 2019
Effects of IL-10- and FasL-overexpressing dendritic cells on liver transplantation tolerance in a heterotopic liver transplantation rat model.
Immunology and cell biology
Chen L,Zhang L,Zhu Z,He W,Gao L,Zhang W,Liu J,Huang A
400-04 was used in Cell Culture to demonstrate that the transfusion of IL-10-FasL/imDCs enhanced immune tolerance and prolonged the survival of liver allografts after LT.
Sun Sep 01 00:00:00 EDT 2019
Deletion of F4L (ribonucleotide reductase) in vaccinia virus produces a selective oncolytic virus and promotes anti-tumor immunity with superior safety in bladder cancer models.
EMBO molecular medicine
Potts KG,Irwin CR,Favis NA,Pink DB,Vincent KM,Lewis JD,Moore RB,Hitt MM,Evans DH
Mon May 01 00:00:00 EDT 2017
The novel monoclonal antibody 9F5 reveals expression of a fragment of GPNMB/osteoactivin processed by furin-like protease(s) in a subpopulation of microglia in neonatal rat brain.
Glia
Kawahara K,Hirata H,Ohbuchi K,Nishi K,Maeda A,Kuniyasu A,Yamada D,Maeda T,Tsuji A,Sawada M,Nakayama H
Tue Nov 01 00:00:00 EDT 2016
Interleukin-4 Ameliorates the Functional Recovery of Intracerebral Hemorrhage Through the Alternative Activation of Microglia/Macrophage.
Frontiers in neuroscience
Yang J,Ding S,Huang W,Hu J,Huang S,Zhang Y,Zhuge Q
400-04 was used in Sample Preparation to show that early intracerebral injection of IL-4 potentially promotes neuro-functional recovery, probably through enhancing the alternative activation of microglia/macrophage.
Fri Mar 25 00:00:00 EDT 2016
β2-adrenoceptor signaling reduction in dendritic cells is involved in the inflammatory response in adjuvant-induced arthritic rats.
Scientific reports
Wu H,Chen J,Song S,Yuan P,Liu L,Zhang Y,Zhou A,Chang Y,Zhang L,Wei W
Fri Apr 15 00:00:00 EDT 2016
Combining Exosomes Derived from Immature DCs with Donor Antigen-Specific Treg Cells Induces Tolerance in a Rat Liver Allograft Model.
Scientific reports
Ma B,Yang JY,Song WJ,Ding R,Zhang ZC,Ji HC,Zhang X,Wang JL,Yang XS,Tao KS,Dou KF,Li X
Mon Sep 19 00:00:00 EDT 2016
Comparative miRNA-Based Fingerprinting Reveals Biological Differences in Human Olfactory Mucosa- and Bone-Marrow-Derived Mesenchymal Stromal Cells.
Stem cell reports
Lindsay SL,Johnstone SA,McGrath MA,Mallinson D,Barnett SC
Tue May 10 00:00:00 EDT 2016
Immunologically augmented skin flap as a novel dendritic cell vaccine against head and neck cancer in a rat model.
Cancer science
Inoue K,Saegusa N,Omiya M,Ashizawa T,Miyata H,Komiyama M,Iizuka A,Kume A,Sugino T,Yamaguchi K,Kiyohara Y,Nakagawa M,Akiyama Y
Sun Feb 01 00:00:00 EST 2015
Estradiol promotes M1-like macrophage activation through cadherin-11 to aggravate temporomandibular joint inflammation in rats.
Journal of immunology (Baltimore, Md. : 1950)
Kou XX,Li CS,He DQ,Wang XD,Hao T,Meng Z,Zhou YH,Gan YH
Sun Mar 15 00:00:00 EDT 2015
Novel mechanism of inhibition of dendritic cells maturation by mesenchymal stem cells via interleukin-10 and the JAK1/STAT3 signaling pathway.
PloS one
Liu WH,Liu JJ,Wu J,Zhang LL,Liu F,Yin L,Zhang MM,Yu B
Wed Jul 17 00:00:00 EDT 2013
Rat macrophage C-type lectin is an activating receptor expressed by phagocytic cells.
PloS one
Lobato-Pascual A,Saether PC,Dahle MK,Gaustad P,Dissen E,Fossum S,Daws MR
Tue Sep 10 00:00:00 EDT 2013
Caspase-2-dependent dendritic cell death, maturation, and priming of T cells in response to Brucella abortus infection.
PloS one
Li X,He Y
Tue Jan 29 00:00:00 EST 2013
Host-based Th2 cell therapy for prolongation of cardiac allograft viability.
PloS one
Amarnath S,Chen H,Foley JE,Costanzo CM,Sennesh JD,Solomon MA,Fowler DH
Fri Apr 29 00:00:00 EDT 2011
Uropathogenic E. coli induce different immune response in testicular and peritoneal macrophages: implications for testicular immune privilege.
PloS one
Bhushan S,Hossain H,Lu Y,Geisler A,Tchatalbachev S,Mikulski Z,Schuler G,Klug J,Pilatz A,Wagenlehner F,Chakraborty T,Meinhardt A
Fri Jul 20 00:00:00 EDT 2012
Antigen-specific T-cell response from dendritic cell vaccination using cancer stem-like cell-associated antigens.
Stem cells (Dayton, Ohio)
Xu Q,Liu G,Yuan X,Xu M,Wang H,Ji J,Konda B,Black KL,Yu JS
Sat Aug 01 00:00:00 EDT 2009
Immature dendritic cell-derived exosomes rescue septic animals via milk fat globule epidermal growth factor-factor VIII [corrected].
Journal of immunology (Baltimore, Md. : 1950)
Miksa M,Wu R,Dong W,Komura H,Amin D,Ji Y,Wang Z,Wang H,Ravikumar TS,Tracey KJ,Wang P
Sun Nov 01 00:00:00 EDT 2009
Unique expression of suppressor of cytokine signaling 3 is essential for classical macrophage activation in rodents in vitro and in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Liu Y,Stewart KN,Bishop E,Marek CJ,Kluth DC,Rees AJ,Wilson HM
Thu May 01 00:00:00 EDT 2008
IL-4-induced selective clearance of oligomeric beta-amyloid peptide(1-42) by rat primary type 2 microglia.
Journal of immunology (Baltimore, Md. : 1950)
Shimizu E,Kawahara K,Kajizono M,Sawada M,Nakayama H
Sat Nov 01 00:00:00 EDT 2008
Rapamycin-conditioned, alloantigen-pulsed dendritic cells promote indefinite survival of vascularized skin allografts in association with T regulatory cell expansion.
Transplant immunology
Horibe EK,Sacks J,Unadkat J,Raimondi G,Wang Z,Ikeguchi R,Marsteller D,Ferreira LM,Thomson AW,Lee WP,Feili-Hariri M
Fri Feb 01 00:00:00 EST 2008
Heme oxygenase-1 ameliorates ischemia/reperfusion injury by targeting dendritic cell maturation and migration.
Antioxidants & redox signaling
Kotsch K,Martins PN,Klemz R,Janssen U,Gerstmayer B,Dernier A,Reutzel-Selke A,Kuckelkorn U,Tullius SG,Volk HD
Sat Dec 01 00:00:00 EST 2007
A non-receptor-mediated mechanism for internalization of molecular chaperones.
Methods (San Diego, Calif.)
Pockley AG,Fairburn B,Mirza S,Slack LK,Hopkinson K,Muthana M
Thu Nov 01 00:00:00 EDT 2007
A small molecule, orally active, alpha4beta1/alpha4beta7 dual antagonist reduces leukocyte infiltration and airway hyper-responsiveness in an experimental model of allergic asthma in Brown Norway rats.
British journal of pharmacology
Cortijo J,Sanz MJ,Iranzo A,Montesinos JL,Nabah YN,Alfón J,Gómez LA,Merlos M,Morcillo EJ
Wed Mar 01 00:00:00 EST 2006
Interleukin-13 enhances cyclooxygenase-2 expression in activated rat brain microglia: implications for death of activated microglia.
Journal of immunology (Baltimore, Md. : 1950)
Yang MS,Ji KA,Jeon SB,Jin BK,Kim SU,Jou I,Joe E
Sat Jul 15 00:00:00 EDT 2006
Beneficial autoimmunity to proinflammatory mediators restrains the consequences of self-destructive immunity.
Immunity
Wildbaum G,Nahir MA,Karin N
Sat Nov 01 00:00:00 EST 2003
Transfer of myelin-specific cells deviated in vitro towards IL-4 production ameliorates ongoing experimental allergic neuritis.
Clinical and experimental immunology
Ekerfelt C,Dahle C,Weissert R,Kvarnström M,Olsson T,Ernerudh J
Mon Jan 01 00:00:00 EST 2001
Reciprocal effects of interleukin-4 and interferon-gamma on immunoglobulin E-mediated mast cell degranulation: a role for nitric oxide but not peroxynitrite or cyclic guanosine monophosphate.
Immunology
Deschoolmeester ML,Eastmond NC,Dearman RJ,Kimber I,Basketter DA,Coleman JW
Fri Jan 01 00:00:00 EST 1999