Sequence Codes
Input your sequence (5' to 3').
Use only the IUPAC 1-letter code abbreviations for all standard, non-standard, mixed and S-oligo bases.
Bases | IUPAC Code |
---|---|
Deoxyadenine | A |
Deoxycytosine | C |
Deoxyguanidine | G |
DeoxyThymidine | T |
Deoxyuracil | U |
Deoxyinosine | I |
Phosphorothioate-A | F |
Phosphorothioate-C | O |
Phosphorothioate-G | E |
Phosphorothioate-T | Z |
A+C+G | V |
A+T+G | D |
T+C+G | B |
A+T+C | H |
A+T | W |
C+G | S |
T+G | K |
A+C | M |
C+T | Y |
A+G | R |
A+G+C+T | N |