Three gastrointestinal bacteria. One TaqPath panel.
Transition to molecular testing with the new TaqPath Enteric Bacterial Select Panel.
The TaqPath Enteric Bacterial Select Panel is a highly sensitive testing solution for detecting and differentiating common gastrointestinal (GI) bacteria: Salmonella spp.; Shigella spp./enteroinvasive E. coli (EIEC)*; and Campylobacter jejuni, Campylobacter coli, and Campylobacter upsaliensis**.
* Shigella spp./EIEC are undifferentiated.
** Campylobacter jejuni, Campylobacter coli, and Campylobacter upsaliensis
Benefits of molecular testing
Offering high sensitivity and fast time-to-results, this powerful PCR panel makes it easy and affordable for you to adopt a molecular diagnostic CE-IVD kit for gastrointestinal testing. As the technological migration from culture to molecular testing continues, the need for expanded molecular testing remains beneficial for at-risk populations, including children, older adults, people with weakened immune systems, and travelers.¹ These populations are often at higher risk of severe disease and hospitalization, and need clear answers. PCR testing is designed to deliver accurate results and aid in differential diagnosis, including coinfections, to assist with treatment or other patient management decisions.
Data sheet: Classic Stool Culture vs Molecular GI Testing
Download this free data sheet to learn more about GI testing methods and get introduced to the TaqPath Enteric Bacterial Select Panel.
Download now
Article: Is it Time to Substitute Culture With Molecular Diagnostics for Infectious Diseases?
Read the latest about the benefits of molecular diagnostics testing and the role it looks to play in the feature. Read now ›
Or if you’d like to learn more about how the TaqPath Enteric Bacterial Select Panel can help your lab transition to molecular testing, talk with one of our specialists.
Detect gastrointestinal pathogens using bacterial PCR GI panel
Product details
Targets
  • Salmonella spp. 
  • Shigella spp./enteroinvasive E. coli (EIEC)* 
  • Campylobacter jejuni, C. coli, C. upsaliensis** 
Internal control
Process control (Bacillus atropheus)
Positive control (included)
TaqPath Enteric Bacterial Select Positive Control
Sample type
Stool
Authorized kit
TaqPath Enteric Bacterial Select Panel
Authorized real-time PCR systems
  • QuantStudio 5 (96-well, 0.2 mL)
  • QuantStudio 5 Dx (96-well, 0.2 mL)***
Turnaround time
Extracted sample (or purified nucleic acid) to results in about 2 hours
Software
Pathogen Interpretive Software CE-IVD Edition
Approval status
CE-IVD
***The TaqPath Enteric Bacterial Select Panel is validated on the CE-IVDD version of the QuantStudio 5 Dx Real-Time PCR System. Please contact your sales representative for more information.
TaqPath Enteric Bacterial Select Panel
Add to your diagnostic testing capabilities using the PCR experience you already have. The TaqPath Enteric Bacterial Select Panel detects and differentiates common gastrointestinal (GI) bacteria.
Gatrointestinal (GI) pathogen panel by PCR
Workflow
Simple workflow
Save time with a simple workflow and automatic results interpretation
Tube
Single tube
Detecting three bacteria and a process control, all in a single well, makes it easy to perform routine patient testing
QuantStudio 5
Affordable price
Easily adopt this panel and never overpay for the targets you need
Hand-in-hand
Confidence in results
Results from clinical evaluation show ≥98% sensitivity and ≥98% specificity so you can be confident in your results
Arrows together
Controls included
Bacillus atrophaeus is used for sample internal control, and positive control for PCR reaction validity
Circle Arrow Right
Flexible throughput
The workflow is scalable to fit your needs, allowing you to test 1 to 93 samples per run
Choose Thermo Fisher Scientific
In a time when getting answers quickly is critical, Thermo Fisher Scientific—a global leader in COVID-19 testing—is leveraging its scale and resources to help the world move forward. With innovative solutions and a reliable supply chain already helping to meet the unprecedented demand for trusted COVID-19 testing, we are continuing to expand our high-quality solutions and urgently accelerate innovation to address current and future challenges.
QuantStudio 5
Trusted technology
Applied Biosystems QuantStudio 5 or 5 Dx real-time PCR systems are part of an innovative family of platforms that combine modern hardware and software in an automatable, compact footprint, enabling molecular diagnostics workflows that operate with maximum efficiency and smarter productivity.
Customer Service
Service and support
Rely on one company to service and support you through all steps of the recommended workflow. Our experts will help you get up and running and maintain your growing molecular testing menu.
Arrows
Growing menu
Our expanding portfolio of Applied Biosystems CE-IVD panels allows you to get more from your existing equipment, like the new Applied Biosystems TaqPath Respiratory Viral Select Panel.
Ordering information
Analysis and reporting
The Applied Biosystems Pathogen Interpretive Software (CE-IVD) is part of the required workflow and must be downloaded prior to instrumentation use. You may download the software upon kit purchase. Labs following the authorized workflow are required to complete an eLearning course and pass an exam to download.
Follow the instructions below to obtain the software from your local support team:
  • Go to thermofisher.com/contactus
  • Select “Instrument Service” as category
  • Enter product name or Cat. No. and select your country
  • Call the local number displayed on the screen
PIS Ce-IVD edition
References
1. Amjad M. An Overview of the Molecular Methods in the Diagnosis of Gastrointestinal Infectious Diseases. Int J Microbiol. 2020 Mar 24;2020:8135724. doi: 10.1155/2020/8135724. PMID: 32280346; PMCID: PMC7128059.
2. Salmonella (Non-Typhoidal) Key Facts. World Health Organization.
3. Pouletty M, De Pontual L, Lopez M, Morin L, et al. Multiplex PCR reveals a high prevalence of multiple pathogens in traveller’s diarrhoea in children. Arch Dis Child. 2019 Feb;104(2):141-146. doi: 10.1136/archdischild-2017-314327. Epub 2018 Jul 7. PMID: 29982173.
Style Sheet for Global Design System
For In Vitro Diagnostic Use.
Regulatory requirements vary by country. Product may not be available in your geographic region.