Search Thermo Fisher Scientific
The Invitrogen Collibri portfolio of library prep kits and accessories generate libraries more quickly, with less bias, in order to match the speed and capacity of Illumina instruments. Advantages available only from Invitrogen Collibri Library Prep Kits:
Collibri Library Prep kits for Illumina Systems enhance NGS-based viral surveillance with higher quality sequences and rapid protocols.
Figure 1. An example of the Collibri PS DNA Library Prep Kit for Illumina systems workflow in which each reagent contains a unique tracking dye that provides visual feedback of proper reagent addition and mixing.
“…it became evident that this can be a very useful tool, not only for training, but in production.”
Nik Matthews, Tumor Profiling Unit, Institute for Cancer Research, UK
The Collibri library prep workflows match the pace needed to efficiently utilize Illumina systems. Reduce time spent on placing orders, managing inventory and contacting technical support from multiple companies. Thermo Fisher Scientific offers technical and ordering support for most steps in the total pre-sequencing workflow for improved efficiency of your lab operations.
Figure 2. The Collibri PCR-Free PS DNA Library Prep Kit protocol is completed in 1.5 hours, reducing total workflow time to ~6.5 hours.
Figure 3. The Collibri Stranded RNA Library Prep Kit with rRNA Depletion protocol is completed in 6.5 hours, reducing total workflow time to ~10.5 hours.
Reduce bias in genomic interpretation
Clean data reveals transcriptome complexity
Determine the concentration of your Illumina NGS libraries
Amplify libraries for sequencing on Illumina systems
NGS libraries must be carefully quantified and normalized so that each sample achieves the required number of reads.
For Research Use Only. Not for use in diagnostic procedures.