Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Thermo Scientific HyPerforma Rocker Bioreactor is controlled by the HyPerforma G3Lab Controller and TruBio software, providing a complete, ready-to-use solution. The Rocker Bioreactor uses Thermo Scientific Rocker BioProcess Containers (BPCs) which provide up to 25 L of working volume and integrates to the pH+dO2 sensor for combined pH, DO, and temperature monitoring.
Designed to create optimal culture conditions
Thermo Scientific HyPerforma Rocker BPC is designed for use with the Thermo Scientific HyPerforma Rocker Bioreactor to create optimal culture conditions for rocked cell cultures. It has multiple ports, allowing the user to optimize the supply of gases, media, and buffers.
They are manufactured using a multi-layered, polyolefin-based, slip agent free film and are equipped with a puck sensor that unites the single-use sensors for DO, pH, and temperature in a unique configuration designed to maintain contact with the culture even at low volumes.
Key features
Designed to create optimal culture conditions
Thermo Scientific HyPerforma Rocker BPC is designed for use with the Thermo Scientific HyPerforma Rocker Bioreactor to create optimal culture conditions for rocked cell cultures. It has multiple ports, allowing the user to optimize the supply of gases, media, and buffers.
They are manufactured using a multi-layered, polyolefin-based, slip agent free film and are equipped with a puck sensor that unites the single-use sensors for DO, pH, and temperature in a unique configuration designed to maintain contact with the culture even at low volumes.
Key features
Controllers that provide ease of transferability and scale-up throughout your process with flexibility and open-architecture.
Thermo Scientific TruBio, TruChrom, and TruPur automation software platforms are capable of controlling bioreactor systems from research to commercial manufacturing.
A range of intuitive process sensors to help you monitor processes, reduce failures, and gain efficiencies.
Available in standard or custom configurations, covering a range of applications, sizes, and connection systems.
Next-generation centrifuge-ready BioProcess Containers provide a single-use, sterilized, closed system for cell harvesting in BioProcessing.
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: