Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Reliable and efficient centrifuges
Thermo Scientific clinical centrifuges are designed to deliver exceptional performance and precision, ensuring optimal workflow efficiency for a wide range of common clinical applications. Our benchtop centrifuges provide a compact footprint with capacities from 120 mL to 4 L and a comprehensive rotor portfolio from 1.5 mL to 50 mL sample tubes, microplates, and 10 mL standard syringes. With innovative technologies and a flexible design, they help simplify the clinical sample preparation process.
Designed for routine clinical and life science applications, the Thermo Scientific Medifuge Small Bench Centrifuge offers a compact footprint and features the unique Thermo Scientific DualSpin rotor with its hybrid design. This allows to quickly change between fixed angle and swinging buckets using the same rotor.
Microcentrifuges |
Small Bench Centrifuges |
Compact General Purpose Centrifuges |
High-Capacity General Purpose Centrifuges |
Description |
|||
A sleek, remarkably small footprint provides outstanding processing power, with a rugged design for years of dependability.
|
Compact centrifuges for effective and efficient processing of clinical samples, including platelet-poor plasma tests.
|
For clinical laboratories requiring mid-level throughput in a small footprint, these 1.6L compact benchtop centrifuges are a great solution. Outfitted with AutoLock to quickly and easily remove rotors to maximize versatility and facilitate convenient cleaning.
|
Achieve 4L capacity in a 3L footprint in this high-capacity centrifuge, available in benchtop or floor-standing options. Outfitted with AutoLock to quickly and easily remove rotors to maximize versatility and facilitate convenient cleaning.
|
Models | |||
Floor standing model:
|
|
Floor standing models: |
Clinical Chemistry/ Hematology |
Pathology | Clinical Microbiology |
Molecular Biology |
Hematology | Clinical Immunology |
Pediatrics | |||
Thermo Scientific Centrifuges |
RBC/ Coagulation Studies |
Urinalysis and Platelet Poor Plasma |
TB Sputum |
Chlamydia Trachomatis |
DNA/ RNA Isolation |
Hemotocrit | Leucocytes | ELISA/ RIA |
Blood Analysis |
Medifuge centrifuge | |||||||||
Microcentrifuges | |||||||||
Small bench centrifuges | |||||||||
Compact general purpose centrifuges | |||||||||
High-capacity general purpose centrifuges |
Thermo Scientific Large Capacity Centrifuges provide medium to high throughput processing with a variety of swinging bucket rotors, so you can use your validated protocols to process a range of products in blood bags or tubes. Our products are certified in accordance with the EU 2017/745 Medical Device Regulation.
Large capacity centrifuges
For regular- to medium-throughput blood processing needs, the Sorvall BP 8 / Cryofuge 8 centrifuge provides 6 x 550mL or 8 x 550mL blood bag capacity, utilizing the same bucket system. The Sorvall BP 16 / Cryofuge 16 centrifuge is designed for high-throughput blood processing with its 12 x 550mL capacity and is upgradeable to 16 x 550mL blood bags with the same bucket selections. Our large capacity centrifuges offer flexibility to meet your needs with a range of buckets and adapters to spin several types of tubes.
Order now
Clinical Centrifuge Solutions