Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Thermo Scientific Custom Peptide synthesis service offers numerous options for synthesis platforms, purity levels, modifications, and formats to meet a variety of research needs. We synthesize peptides of 2–110 amino acids using the latest Fmoc solid-phase technology, and purify our products using HPLC. Synthesis scales range from 0.1 mg to 1 kg+, and purities from crude to >98% based on the service.
Standard Peptides | Alexa Fluor Dye-Labeled Peptide Synthesis Service | PEPotec Immuno Peptide Libraries | PEPotec SRM Peptide Libraries | HeavyPeptide AQUA Peptide Synthesis Service | |
---|---|---|---|---|---|
Primary application | Basic research or biomarker discovery | Fluorescence imaging | Epitope mapping, vaccine development | Discovery | Quantitative mass spectrometry |
Key features | Over 400 modifications available | High performing dyes | High throughput, low toxicity | High throughput | High quality and concentration precision |
Peptide length (amino acids) | 2–110* | 6–70* | 6–25 | 6–25 | 6–30* |
Scale | 1 mg–1 kg+ | 1–10 mg | 1 mg | >0.1 mg | Multiple scales available |
Purity | Crude to 98% | Crude to 98% | Crude | Crude | 95–97% |
QC | MS and HPLC | MS and HPLC | MS | MS check or MS analysis | MS, analytical HPLC, AAA |
Delivery time | 2–3 weeks | 2–3 weeks | 4–5 weeks | 2–3 weeks | 5–7 weeks standard AQUA 2–3 weeks AQUA express** |
* Please inquire about shorter or longer peptide lengths.
**AQUA Express requests are available for up to 10 sequences on a single order and must be approved for express turnaround time.
Type | Name | Abbreviation or alternative name | Position within sequence | Graphical representation of available positions |
---|---|---|---|---|
Misc | 1-Adamantanecarbonyl - | 1-Adamantanecarbonyl-NH | N-term | ■□□□...□□□□ |
Misc | 2,4,6-Trinitrophenyl - TNP | TNP-NH | N-term | ■□□□...□□□□ |
Misc | 2-Furoyl - | 2-Furoyl-NH | N-term | ■□□□...□□□□ |
Misc | 2-Naphtoic acid - | Naphtyl-NH | N-term | ■□□□...□□□□ |
Misc | 3-(Maleimido)propionyl - | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | 3,5-Diiodo-Tyrosine - | [Y(I)2] | Internal | ■■■■...■■■■ |
Misc | 3-Chloro-Tyrosine - | [Y(Cl)] | Internal | ■■■■...■■■■ |
Misc | 3-nitro-2-pyridylthio-L-Cys - | [C(Npys)] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Indole - | [3-Nitro-Indole] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Pyrrol - | [3-Nitro-Pyrrol] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Tyrosine - | [Y(NO2)] | Internal | ■■■■...■■■■ |
Misc | 4-Methylthio-2-oxobutanoic ac. - | [H3C-S-CH2-CH2-CO-CO-NH] | N-term | ■□□□...□□□□ |
Misc | 4-nitrobenzo-2-oxa-1,3-diazole - NBD | NBD-NH | N-term | ■□□□...□□□□ |
Misc | 4-Pentynoic acid - | 4-Pentynoic acid-NH | N-term | ■□□□...□□□□ |
Misc | 4-Phenylazophenol - O-PAP | CO-PAP | C-term | □□□□...□□□■ |
Misc | 6-Bromo-D/L-Tryptophan - | [D/L-W(Br)] | Internal | ■■■■...■■■■ |
Misc | 6-Cloro-Tryptophan - | [W(Cl)] | Internal | ■■■■...■■■■ |
Misc | Acetylation - | Acetyl-NH | N-term | ■□□□...□□□□ |
Misc | Aldehyde - | CHO | C-term | □□□□...□□□■ |
Misc | Amidation - | CONH2 | C-term | □□□□...□□□■ |
Misc | Amino nitrophenyl prop. acid - | [Amino nitrophenyl prop. acid] | Internal | ■■■■...■■■■ |
Misc | Amino-nitrophenyl propion. ac. - ANP-linker | [ANP] | Internal | ■■■■...■■■■ |
Misc | Aminooxyacetyl - | H2N-O-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Benzoyl - Bz | Benzoyl-NH | N-term | ■□□□...□□□□ |
Misc | Benzyloxycarbonyl - Cbz, Z | Z-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - | Biotin-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - Ethylenediamin-Biotin | CO-NH-CH2-CH2-NH-Biotin | C-term | □□□□...□□□■ |
Misc | Biotin-SS-linker - | Biotin-SS-NH | N-term | ■□□□...□□□□ |
Misc | Bis(aminoethyl)ethylene glycol - | CO-NH-(CH2-CH2-O)2-CH2-CH2-NH2 | C-term | □□□□...□□□■ |
Misc | Brom acetic acid - | BrCH2CO-NH | N-term | ■□□□...□□□□ |
Misc | Chlormethylketon - CMK | CMK | C-term | □□□□...□□□■ |
Misc | Cys(4-Methoxytrityl) - Cys(MMT) | [C(Mmt)] | Internal | ■■■■...■■■■ |
Misc | Cys(Acetamidomethyl) - Cys(Acm) | [C(Acm)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carbamidomethyl) - Cys(CAM) / Cys(CH2CONH2) | [C(Cam)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carboxymethyl) - Cys(CMC) / Cys(CH2COOH) | [C(Cmc)] | Internal | ■■■■...■■■■ |
Misc | Cys(di(palmitoyloxy)-propyl) - Cys(Pam2) | [C(Pam2)] | Internal | ■■■■...■■■■ |
Misc | Cys(Farnesyl) - | [C(Farnesyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(HRP) - Cys(horseradish peroxidase) | [C(HRP)] | Internal | ■■■■...■■■■ |
Misc | Cys(MTSL) - | [C(MTSL)] | Internal | ■■■■...■■■■ |
Misc | Cys(Myristyl) - Cys(CH2(CH2)12CH3) | [C(Myristyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(Palmitoyl) - | [C(Palm)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Butyl) - | [C(S-But)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Ethyl) - | [C(S-Et)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isobutyl) - | [C(S-Isobut)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isopropyl) - | [C(S-Isoprop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Methyl) - | [C(S-Me)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Pentyl) - | [C(S-Pentyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Propyl) - | [C(S-Prop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-tert-Butyl) - | [C(S-tert-But] | Internal | ■■■■...■■■■ |
Misc | Cysteamide - | Cysteamide | C-term | □□□□...□□□■ |
Misc | Decanoyl - | Decanoyl-NH | N-term | ■□□□...□□□□ |
Misc | Desthiobiotin - | Desthiobiotin-NH | N-term | ■□□□...□□□□ |
Misc | Digoxigenin - | Digoxigenin-NH | N-term | ■□□□...□□□□ |
Misc | DOTA - | [DOTA] | N-term | ■□□□...□□□□ |
Misc | Ethylamide - N-Ethyl | CONH(C2H5) | C-term | □□□□...□□□■ |
Misc | Ethylester - O-Ethyl | COOEt | C-term | □□□□...□□□■ |
Misc | Fmoc - | Fmoc-NH | N-term | ■□□□...□□□□ |
Misc | Formyl - | Formyl-NH | N-term | ■□□□...□□□□ |
Misc | Glu(Arg) - Arg at side chain of Glu | [Glu(Arg)] | Internal | ■■■■...■■■■ |
Misc | Homoserine lactone - | [Homoser lactone] | C-term | □□□□...□□□■ |
Misc | HRP - | CO-NH -HRP | C-term | □□□□...□□□■ |
Misc | HRP - | HRP-NH | N-term | ■□□□...□□□□ |
Misc | Hydrazide - | CO-NH -NH2 | C-term | □□□□...□□□■ |
Misc | Hydrazine-succinyl - | H2N-NH-CO-CH2-CH2-CO-NH- | N-term | ■□□□...□□□□ |
Misc | Iodoacetyl - | IH2C-CO- | N-term | ■□□□...□□□□ |
Misc | Lys(2-Aminooxy acetic acid) - Lys(Aoa) | [K(Aoa)] | Internal | ■■■■...■■■■ |
Misc | Lys(Acetyl) - | [K(Ac)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-Biotin) - | [K(Ahx-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-SS-Biotin) - | [K(Ahx-SS-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Biotin) - | [K(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Boc) - | [K(Boc)] | Internal | ■■■■...■■■■ |
Misc | Lys(Butyryl) - | [K(Butyryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Desthiobiotin) - | [K(Desthiobiotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Digoxigenin] - | [K(Digoxigenin)] | Internal | ■■■■...■■■■ |
Misc | Lys(DST-PEG 2-Biotin) - | [K(DST-PEG 2-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ferrocene) - | [K(Ferrocene)] | Internal | ■■■■...■■■■ |
Misc | Lys(Formyl) - | [K(Formyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(GG) - Ubiquitin | [K(GG)] | Internal | ■■■■...■■■■ |
Misc | Lys(GGRLR) - | [K(GGRLR)] | Internal | ■■■■...■■■■ |
Misc | Lys(Glutaryl) - | [K(Glutaryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Lipoyl) - | [K(Lipoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(NIP) - | [K(NIP)] | Internal | ■■■■...■■■■ |
Misc | Lys(Palmitoyl) - | [K(Palmitoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Propionyl) - | [K(Propionyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Pyrofolate) - | [K(Pyrofolate)] | Internal | ■■■■...■■■■ |
Misc | Lys(Stearyl) - | [K(Stearyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Succinyl) - | [K(Succinyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(TFA) - Lysine(Trifluoroacetyl) | [K(TFA)] | Internal | ■■■■...■■■■ |
Misc | Maleimide - via SMCC | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | Methanethiosulfonate - MTS | MTS-NH | N-term | ■□□□...□□□□ |
Misc | Methoxysuccinyl - | H3C-O-CO-CH2-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Methylamide - N-Methyl | CO-NH(CH3) | C-term | □□□□...□□□■ |
Misc | Methylester - O-Methyl | COOMe | C-term | □□□□...□□□■ |
Misc | Myristoyl - | Myristoyl-NH | N-term | ■□□□...□□□□ |
Misc | N-Hydroxysuccinimid ester - NHS | CO-NHS | C-term | □□□□...□□□■ |
Misc | Orn(Biotin) - | [Orn(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Palmitoyl - | Palmitoyl-NH | N-term | ■□□□...□□□□ |
Misc | para-Bromo-D-Phenylalanine - | [D-F(4-Br)] | Internal | ■■■■...■■■■ |
Misc | para-Chloro-Phenylalanine - | [F(4-Cl)] | Internal | ■■■■...■■■■ |
Misc | para-Iodo-Phenylalanine - | [F(I)] | Internal | ■■■■...■■■■ |
Misc | para-Nitro-Phenylalanine - | [F(NO2)] | Internal | ■■■■...■■■■ |
Misc | Phenylacetic acid - Paa | Paa-NH | N-term | ■□□□...□□□□ |
Misc | Photo-Leucine - | [Photo-L] | Internal | ■■■■...■■■■ |
Misc | Photo-Methionine - | [Photo-M] | Internal | ■■■■...■■■■ |
Misc | p-tosyl-Gly - | [p-tosyl-G] | N-term | ■□□□...□□□□ |
Misc | S-Acetylmercaptosuccinyl - SAMSA | SAMSA-NH | N-term | ■□□□...□□□□ |
Misc | Ser(Acetyl) - | [S(Ac)] | Internal | ■■■■...■■■■ |
Misc | Ser(Octanoyl) - | [S(Oct)] | Internal | ■■■■...■■■■ |
Misc | Stearyl - | Stearyl-NH | N-term | ■□□□...□□□□ |
Misc | Succinyl - | Succinyl-NH | N-term | ■□□□...□□□□ |
Misc | t-Butylester - | COOtBu | C-term | □□□□...□□□■ |
Misc | tert. Butoxycarbonyl - Boc | Boc-NH | N-term | ■□□□...□□□□ |
Misc | Thiobenzyl - | CO-S(Bzl) | C-term | □□□□...□□□■ |
Misc | Thiopropionyl - Tpa | Tpa-NH | N-term | ■□□□...□□□□ |
Misc | trans-Cinnamoyl - | Cinnamoyl-NH | N-term | ■□□□...□□□□ |
Misc | Tyr(Palmitoyl) - | [Y(Palm)] | Internal | ■■■■...■■■■ |
Dye | (2,4-Dinitrophenyl)-Glycine - (2,4-DNP)-Gly | DNP-NH-G | N-term | ■□□□...□□□□ |
Dye | 2,4-Dinitrophenyl - DNP | CO-DNP | C-term | □□□□...□□□■ |
Dye | 2,4-Dinitrophenyl - DNP | DNP-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Carboxyfluorescein - 5/6-FLUO | 5/6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Tetramethylrhodamine - 5/6-TAMRA | 5/6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 5-Carboxyfluorescein - 5-FLUO | 5-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5-Tetramethylrhodamine - 5-TAMRA | 5-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 6-Carboxyfluorescein - 6-FLUO | 6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 6-Tetramethylrhodamine - 6-TAMRA | 6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 7-Amino-4-methylcoumarin - AMC | CO-AMC | C-term | □□□□...□□□■ |
Dye | Ala(2-Bacd) - benzo[b]acridin-12(5H)-on-2-yl | [Ala(2-Bacd)] | Internal | ■■■■...■■■■ |
Dye | Amino-trifluoromethyl-coumarin - AFC | CO-AFC | C-term | □□□□...□□□■ |
Dye | ATTO 647N - | ATTO 647N-NH | N-term | ■□□□...□□□□ |
Dye | Cy7 - | Cy7-NH | N-term | ■□□□...□□□□ |
Dye | Cys(5/6-TAMRA) - | [C(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(5-TAMRA) - | [C(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(6-TAMRA) - | [C(6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(acetamido-fluorescein) - | [C(acetamido-fluorescein)] | Internal | ■■■■...■■■■ |
Dye | Cys(AEDANS) - | [C(AEDANS)] | Internal | ■■■■...■■■■ |
Dye | Cys(Allophycocyanine) - | [C(Allophycocyanine)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 555) - | [C(DY 555)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 647P1) - | [C(DY 647P1)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 649) - | [C(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Cys(DYQ660) - | [Cys(DYQ660)] | Internal | ■■■■...■■■■ |
Dye | Dabcyl - | Dabcyl-NH | N-term | ■□□□...□□□□ |
Dye | Dabsyl - | Dabsyl-NH | N-term | ■□□□...□□□□ |
Dye | Dansyl - | CO-Dansyl | C-term | □□□□...□□□■ |
Dye | Dansyl - | Dansyl-NH | N-term | ■□□□...□□□□ |
Dye | DAP(DNP) - DNP at Diaminopropionic acid | [DAP(DNP)] | Internal | ■■■■...■■■■ |
Dye | DAP(DY 647) - DY 647 at Diaminopropionic ac. | [DPA(DY-647)] | Internal | ■■■■...■■■■ |
Dye | DAP(DyLight 488) - DyLight 488 at Diaminoprop. ac | [DPA(DY 488)] | Internal | ■■■■...■■■■ |
Dye | DAP(NMA) - NMA at Diaminopropionic acid | [DAP(NMA)] | Internal | ■■■■...■■■■ |
Dye | Dinitrophenyl-Ethylenediamine - EDDnp | CO-EDDnp | C-term | □□□□...□□□■ |
Dye | D-Lys(5/6-Fluorescein) - | [D-K(5/6-Fluorescein)] | Internal | ■■■■...■■■■ |
Dye | D-Lys(5/6-TAMRA) - | [D-Lys(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | DY 495 - | DY 495-NH | N-term | ■□□□...□□□□ |
Dye | DY 505 - | DY 505-NH | N-term | ■□□□...□□□□ |
Dye | DY 530 - | DY 530-NH | N-term | ■□□□...□□□□ |
Dye | DY 547 - | DY 547-NH | N-term | ■□□□...□□□□ |
Dye | DY 549 - | DY 549-NH | N-term | ■□□□...□□□□ |
Dye | DY 630 - | DY630-NH | N-term | ■□□□...□□□□ |
Dye | DY 633 - | DY 633-NH | N-term | ■□□□...□□□□ |
Dye | DY 635 - | DY 635-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - | DY 647-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - aminomodified | CO-DY 647 | C-term | □□□□...□□□■ |
Dye | DY 649 - | DY 649-NH | N-term | ■□□□...□□□□ |
Dye | DY 649P1 - aminomodified | CO-DY 649P1 | N-term | ■□□□...□□□□ |
Dye | DY 650 - | DY650-NH | Internal | ■■■■...■■■■ |
Dye | DY 680 - | DY 680-NH | N-term | ■□□□...□□□□ |
Dye | DY 682 - | DY 682-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | DY 750-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | CO-DY 750 | C-term | □□□□...□□□■ |
Dye | DY 751 - | DY 751-NH | N-term | ■□□□...□□□□ |
Dye | DY 800 - | DY 800-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 488 - | DY 488-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 504Q - | DyLight 504Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 543Q - | DyLight 543Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 554-R1 - | DyLight 554-R1-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 683Q - | DyLight 683Q-NH | N-term | ■□□□...□□□□ |
Dye | DYQ 660 - | DYQ 660-NH | N-term | ■□□□...□□□□ |
Dye | EDANS - | CO-EDANS | C-term | □□□□...□□□■ |
Dye | Glu(EDANS) - | [E(EDANS)] | Internal | ■■■■...■■■■ |
Dye | His(DNP) - | [H(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(2-(N-methylamino)benzoyl) - Lys(NMA) | [Lys(NMA)] | Internal | ■■■■...■■■■ |
Dye | Lys(2,4-DNP) - | [K(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-Fluorescein) - | [K(5/6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-TAMRA) - | [K(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-Fluorescein) - | [K(5-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-TAMRA) - | [K(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(6-Fluorescein) - | [K(6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(Ahx-(5-Fluorescein)) - | [K(Ahx-(5-Fluo))] | Internal | ■■■■...■■■■ |
Dye | Lys(ATTO 655) - | [K(ATTO 655)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dabcyl) - | [K(Dabcyl)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dde) - | [Lys(Dde)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 405) - | [K(DY 405)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 495) - | [K(DY 495)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 530) - | [K(DY 530)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 547) - | [K(DY 547)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 590) - | [K(DY 590)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 630) - | [K(DY 630)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 635) - | [K(DY 635)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 647) - | [K(DY 647)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 650) - | [Lys(DY 650)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 800) - | [K(DY 800)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 488) - | [K(DY 488)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 633) - Lys(DY633) | [K(DyLight 633)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 649) - | [K(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Lys(DYQ 3) - | [K(DYQ 3)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOC) - (7-hydroxycoumarin-3-yl)formyl | [Lys(HOC)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOMCA) - | [Lys(HOMCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MCA) - | [K(MCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MOC) - (7-methoxycoumarin-3-yl)formyl | Lys(MOC) | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine B) - | [K(Rhodamine B)] | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine Green) - | [K(Rhodamine Green)] | Internal | ■■■■...■■■■ |
Dye | Methoxycoumarin acetic acid - MCA | CO-MCA | C-term | □□□□...□□□■ |
Dye | Methoxycoumarin acetic acid - MCA | MCA-NH | N-term | ■□□□...□□□□ |
Dye | Ornithine(Coumarin343) - | [Orn(C343)] | Internal | ■■■■...■■■■ |
Dye | para-Nitroanilide - pNA | CO-pNA | C-term | □□□□...□□□■ |
Dye | para-Nitrophenylester - | COONp | C-term | □□□□...□□□■ |
Dye | QSY21 - | QSY21-NH | N-term | ■□□□...□□□□ |
Dye | Rhodamine 110 - | CO-Rhod 110 | C-term | □□□□...□□□■ |
Dye | Rhodamine B - | Rhodamin B-NH | N-term | ■□□□...□□□□ |
Dye | R-Phycoerythrin - | CO- R-Phycoerythrin | C-term | □□□□...□□□■ |
Dye | Tide Quencher 2 - TQ2 | TQ2-NH | N-term | ■□□□...□□□□ |
Linkers | 12-Aminododecanoic acid - Ado | [Ado] | Internal | ■■■■...■■■■ |
Linkers | 5-Aminopentanoic acid - Aminovaleric acid (Ava) | [Ava] | Internal | ■■■■...■■■■ |
Linkers | 6-Aminohexanoic acid (Ahx) - Aminocaproic acid (Aca) | [Ahx] | Internal | ■■■■...■■■■ |
Linkers | 8-Aminooctanoic acid - Aoc | [Aoc] | Internal | ■■■■...■■■■ |
Linkers | Cys(PEG 40000) - | [C(PEG 40000)] | Internal | ■■■■...■■■■ |
Linkers | Methyl-PEG12 - MS(PEG)12 | MS(PEG)12-NH | N-term | ■□□□...□□□□ |
Linkers | Methyl-PEG24 - MS(PEG)24 | MS(PEG)24-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 12 (40 atoms) - Amino-PEG-acid | [PEG 12] | Internal | ■■■■...■■■■ |
Linkers | PEG 2 (9 atoms) - 8-Amino-3,6-dioxaoctanoic acid | [PEG 2] | Internal | ■■■■...■■■■ |
Linkers | PEG 20,000 - | PEG 20,000-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 2000 - Amino-PEG-acid | [PEG 2000] | Internal | ■■■■...■■■■ |
Linkers | PEG 3 (13 atoms) - Amino-trioxadodecanoic acid | [PEG 3] | Internal | ■■■■...■■■■ |
Linkers | PEG 4 (16 atoms) - Amino-PEG-acid | [PEG 4] | Internal | ■■■■...■■■■ |
Linkers | PEG 5 (19 atoms) - Amino-PEG-acid | [PEG 5] | Internal | ■■■■...■■■■ |
Linkers | PEG 5000 - | CO-PEG 5000 | C-term | □□□□...□□□■ |
Linkers | PEG 5000 - Amino-PEG-acid | [PEG 5000] | Internal | ■■■■...■■■■ |
Linkers | PEG 6 (22 atoms) - Amino-PEG-acid | [PEG 6] | Internal | ■■■■...■■■■ |
Linkers | PEG 8 (28 atoms) - Amino-PEG-acid | [PEG 8] | Internal | ■■■■...■■■■ |
Linkers | PEG-4 Azide - | N3-PEG4-NH | N-term | ■□□□...□□□□ |
Special AAs | 2-(4'-Pentenyl)alanine - | [A(pentenyl)] | Internal | ■■■■...■■■■ |
Special AAs | 2,3-Diaminopropionic acid - DAP | [DAP] | Internal | ■■■■...■■■■ |
Special AAs | 2-Amino-4-thiazolinic acid - | [2-Amino-4-thiazolinyl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Indanyl-Glycine - Igl | [Igl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Methyl-D-Tryptophan - | [D-W(2-Me)] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-Alanine - 2-Nal | [2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-D-Alanine - D-2-Nal | [D-2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 3,3-Diphenyl-Alanine - | [A(F2)] | Internal | ■■■■...■■■■ |
Special AAs | 3,4-Dihydroxy-Phenylalanine - | [F(OH)2] | Internal | ■■■■...■■■■ |
Special AAs | 5,5-Dimethyl-Proline - | [P(Me2)] | Internal | ■■■■...■■■■ |
Special AAs | 5-Hydroxy-Tryptophan - | [W(OH)] | Internal | ■■■■...■■■■ |
Special AAs | Allyl-Glycine - 2-Aminopent-4-enoic acid | [G(allyl)] | Internal | ■■■■...■■■■ |
Special AAs | alpha-Allylalanine - | [Allyl-A] | Internal | ■■■■...■■■■ |
Special AAs | alpha-aminoadipic acid - Aad | [Aad] | Internal | ■■■■...■■■■ |
Special AAs | alpha-t-butylglycine - | [G(tBu)] | Internal | ■■■■...■■■■ |
Special AAs | Aminobenzoic acid - Abz | [Abz] | Internal | ■■■■...■■■■ |
Special AAs | Aminobutyric acid - Abu | [Abu] | Internal | ■■■■...■■■■ |
Special AAs | Aminocyclohexyl carbox. acid - Ac6c | [Ac6c] | Internal | ■■■■...■■■■ |
Special AAs | Aminoisobutyric acid - Aib | 0 | Internal | ■■■■...■■■■ |
Special AAs | Asp beta-methyl ester - Asp(OMe) | [D(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Asp(Isopropylamide) - | [D(Isopropylamide)] | Internal | ■■■■...■■■■ |
Special AAs | Azetidine-2-carboxylic acid - Aze | [Aze] | Internal | ■■■■...■■■■ |
Special AAs | Canavanine - Cav | [Cav] | Internal | ■■■■...■■■■ |
Special AAs | Citrulline - | [Cit] | Internal | ■■■■...■■■■ |
Special AAs | Cyclopentyl-Glycine - | [G(cyclopentyl)] | Internal | ■■■■...■■■■ |
Special AAs | Cysteic acid - Cya | [Cya] | Internal | ■■■■...■■■■ |
Special AAs | Cysteine sulfonate - | [C(SO3H)] | Internal | ■■■■...■■■■ |
Special AAs | D-2-Indanyl-Glycine - D-Igl | [D-Igl] | Internal | ■■■■...■■■■ |
Special AAs | D-4-Hydroxyphenly-Glycine - D-Hpg | [D-Hpg] | Internal | ■■■■...■■■■ |
Special AAs | D-Alanine - | *A | Internal | ■■■■...■■■■ |
Special AAs | D-Amino Acid - | [D-aa] | Internal | ■■■■...■■■■ |
Special AAs | D-Arginine - | *R | Internal | ■■■■...■■■■ |
Special AAs | D-Asparagine - | *N | Internal | ■■■■...■■■■ |
Special AAs | D-Aspartic acid - | *D | Internal | ■■■■...■■■■ |
Special AAs | D-Cysteine - | *C | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamic acid - | *E | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamine - | *Q | Internal | ■■■■...■■■■ |
Special AAs | D-Histidine - | *H | Internal | ■■■■...■■■■ |
Special AAs | D-Homocysteine - | [D-Homocys] | Internal | ■■■■...■■■■ |
Special AAs | D-Homoserine - | [D-Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Diaminotrioxatridecan suc. ac. - TTDS | [TTDS] | Internal | ■■■■...■■■■ |
Special AAs | D-Isoleucine - | *I | Internal | ■■■■...■■■■ |
Special AAs | D-Leucine - | *L | Internal | ■■■■...■■■■ |
Special AAs | D-Lysine - | *K | Internal | ■■■■...■■■■ |
Special AAs | D-Methionine - | *M | Internal | ■■■■...■■■■ |
Special AAs | D-Ornithine - | [D-Orn] | Internal | ■■■■...■■■■ |
Special AAs | D-Phenylalanine - | *F | Internal | ■■■■...■■■■ |
Special AAs | D-Proline - | *P | Internal | ■■■■...■■■■ |
Special AAs | D-Serine - | *S | Internal | ■■■■...■■■■ |
Special AAs | D-Threonine - | *T | Internal | ■■■■...■■■■ |
Special AAs | D-Tryptophan - | *W | Internal | ■■■■...■■■■ |
Special AAs | D-Tyrosine - | *Y | Internal | ■■■■...■■■■ |
Special AAs | D-Valine - | *V | Internal | ■■■■...■■■■ |
Special AAs | Formylglycine - | [G(CHO)] | Internal | ■■■■...■■■■ |
Special AAs | gamma-Carboxyglutamic acid - Gla | [Gla] | Internal | ■■■■...■■■■ |
Special AAs | Gamma-Glu, amidated - Gla, amidation at side chain | [Gla amide] | Internal | ■■■■...■■■■ |
Special AAs | Glu gamma-methyl ester - Glu(OMe) | [E(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Homo-Citrulline - | [Homocit] | Internal | ■■■■...■■■■ |
Special AAs | Homophenylalanine - | [HomoF] | Internal | ■■■■...■■■■ |
Special AAs | Homoserine - | [Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Hydroxy-Proline - Hyp | [Hyp] | Internal | ■■■■...■■■■ |
Special AAs | Hyposine - Hpu | [Hpu] | Internal | ■■■■...■■■■ |
Special AAs | Isoaspartic acid - | 0 | Internal | ■■■■...■■■■ |
Special AAs | Kynurenine - Kyn | [Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptoacetic acid - Thioglycolic acid | [Mercaptoacetyl] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptopropionic acid - Mpa | [Mpa] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfone - | [M(O2)] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfoxide - | [M(O)] | Internal | ■■■■...■■■■ |
Special AAs | N-Formylkynurenine - | [Formyl-Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Norleucine - | [Nle] | Internal | ■■■■...■■■■ |
Special AAs | Norvaline - 2-Aminopentanoic acid, Nva | [Nva] | Internal | ■■■■...■■■■ |
Special AAs | Octahydroindole carbox. acid - | [Oic] | Internal | ■■■■...■■■■ |
Special AAs | Ornitine - | [Orn] | Internal | ■■■■...■■■■ |
Special AAs | para-Acetylphenylalanine - pAF | [para-Ac-Phe] | Internal | ■■■■...■■■■ |
Special AAs | Phenylglycine - Phg | [Phg] | Internal | ■■■■...■■■■ |
Special AAs | Pyroglutamic acid - Pyr | [Pyr] | Internal | ■■■■...■■■■ |
Special AAs | Selenocysteine - Sec | [Sec] | Internal | ■■■■...■■■■ |
Special AAs | β-Alanine - | [β-A] | Internal | ■■■■...■■■■ |
Special AAs | β-Cyclohexyl-Alanine - Cha | [Cha] | Internal | ■■■■...■■■■ |
Special AAs | β-cyclohexyl-D-Alanine - D-Cha | [D-Cha] | Internal | ■■■■...■■■■ |
Special AAs | Tetrahydroisquinol.carb.acid - Tic | [Tic] | Internal | ■■■■...■■■■ |
Special AAs | Thiazolidine-4-carboxylic acid - Thz | [Thz] | Internal | ■■■■...■■■■ |
Special AAs | γ-Glutamic acid - | [γ-E] | Internal | ■■■■...■■■■ |
PTM | 1-Methyl-Tryptophan - | [W(1-Me)] | Internal | ■■■■...■■■■ |
PTM | 2,6-Dimethyl-Tyrosine - | [Y(Me2)] | Internal | ■■■■...■■■■ |
PTM | 2-Methylproline - | [P(2-Me)] | Internal | ■■■■...■■■■ |
PTM | 2-Methyl-Tryptophan - | [2-Me-Trp] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (asym.) - | [R(Me)2-asym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (sym.) - | [R(Me)2-sym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Cysteine - Penicillamine | [Pen] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Lysine - | [K(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Phenylalanine - | [F(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Valine - | [V(Me)2] | Internal | ■■■■...■■■■ |
PTM | Methyl-Arginine - | [R(Me)] | Internal | ■■■■...■■■■ |
PTM | Methyl-Lysine - | [K(Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Alanine - | [Ala(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Glycine - | [G(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Leucine - | [L(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Phenylalanine - | [F(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Tyrosine - | [Y(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Valine - | [V(N-Me)] | Internal | ■■■■...■■■■ |
PTM | Phosphono-Phenylalanine - | [F(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Serine - | [S(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Threonin - | [T(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Tyrosin - | [Y(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | β-GalNAc-Threonine - | [T(β-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Asparagine - | [N(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Serine - | [S(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Threonine - | [T(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Serine - | [S(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Threonine - | [T(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Tyrosine - | [Y(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Arginine - | [R(Me3)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Lysine - | [K(Me)3] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Serine - | [S(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Threonine - | [T(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Serine - | [S(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Threonine - | [T(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Aspartic acid - | [D(α-Me)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Glutamic acid - | [E(α-Me)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (13C3;15N) - | [A(13C3; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (D3) - | [A(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6) - | [R(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6;15N4) - | [R(13C6; 15N4)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asn (13C4;15N2) - | [N(13C4; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asp (13C4;15N - | [D(13C4; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gln (13C5;15N2) - | [Q(13C5;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Glu (13C5;15N) - | [E(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gly (13C2;15N) - | [G(13C2; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy His (15N3) - | [His(15N3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ile (13C6;15N) - | [I(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C1) - | [L(13C1)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6) - | [L(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6;15N) - | [L(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D10) - | [L(D10)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D3) - | [L(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6) - | [K(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6;15N2) - | [K(13C6; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Phe (13C9;15N) - | [F(13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Pro (13C5;15N) - | [P(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (13C3;15N) - | [S (13C3;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (D3) - | [S(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Thr(13C4;15N) - | [T(13C4;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Trp (13C11;15N2) - | [W (13C11;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Tyr (13C9;15N) - | [Y (13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5) - | [V(13C5)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5;15N) - | [V(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | L-tyrosine-phenyl-d4 - deuterated Tyr | [Tyr(D4)] | Internal | ■■■■...■■■■ |
Isotopic | TMT6-126 - | TMT6-126-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-127 - | TMT6-127-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-128 - | TMT6-128-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-129 - | TMT6-129-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-130 - | TMT6-130-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-131 - | TMT6-131-NH | N-term | ■□□□...□□□□ |
Immunization | Blue Carrier Protein - | CO-NH -Blue Carrier Protein | C-term | □□□□...□□□■ |
Immunization | BSA - | CO-BSA | C-term | □□□□...□□□■ |
Immunization | BSA via Glutaraldehyde - | BSA-NH | N-term | ■□□□...□□□□ |
Immunization | Cys(BSA) - | [C(BSA)] | Internal | ■■■■...■■■■ |
Immunization | Cys(KLH) - | [C(KLH)] | Internal | ■■■■...■■■■ |
Immunization | Cys(OVA) - | [C(OVA)] | Internal | ■■■■...■■■■ |
Immunization | KLH - | CO-KLH | C-term | □□□□...□□□■ |
Immunization | KLH via Glutaraldehyde - | KLH-NH | N-term | ■□□□...□□□□ |
Immunization | Lys(BSA) - | [K(BSA] | Internal | ■■■■...■■■■ |
Immunization | Lys(KLH) - | [K(KLH] | Internal | ■■■■...■■■■ |
Immunization | MAP 16 - Crude 8-20 aa, at C-term. only | MAP 16 | C-term | □□□□...□□□■ |
Immunization | MAP 4 - Crude 8-20 aa, at C-term. only | MAP 4 | C-term | □□□□...□□□■ |
Immunization | MAP 8 - Crude 8-20 aa, at C-term. only | MAP 8 | C-term | □□□□...□□□■ |
Immunization | OVA - | CO-OVA | C-term | □□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | 3-Azido-Alanine - Aza | [Aza] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | 5-Azidopentanoyl - 5-Azidovalerianoyl | 5-Azidopentanoyl-NH | N-term | ■□□□...□□□□ |
Cyclization/Dimerization/Special Synthesis | 6-Azido-Norleucine - Nle(N3) | [Nle(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azidohomoalanine - Aha/ Dab(N3) | [Aha] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azido-Lysine - | [K(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (C-C), disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (D-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (E-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (K-C-terminus) - side chain of K to C-terminus | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (NH-CO), homodetic - | [ ] | N-term | ■□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | Double disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | D-Propargylglycine - D-Pra | [D-Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Hetero-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Homo-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Mixed amino acids - | [X] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | para-Azido-Phenylalanine - | [F(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylalanine - alpha-Me-L-Ala(Pentynyl)-OH | [Propargylala] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylglycine - Pra | [Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Asymmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Symmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Site directed disulfide bridge - Cyclisation (C-C) | [Cys cyclo(S-S)] | Internal | ■■■■...■■■■ |
A. The shelf life is >24 months at –20°C for lyophilized peptides protected by argon in the original non-open container. However, it is sequence dependent and may be shorter for some sequences.
A. Unless otherwise requested, all peptides are delivered as lyophilized trifluoroacetic salts with free amino and carboxy termini. N-terminal acetylation or C-terminal amidation is available for a small fee but must be requested when ordering your peptide(s).
A. The ability to synthesize the peptide and its solubility, stability, and overall charge are discussed in the Protein Biology Learning Center under the Peptide Design.
A. All the information required to get an accurate quote is included in the Quote Request form.
Standard Peptides | Alexa Fluor Dye-Labeled Peptide Synthesis Service | PEPotec Immuno Peptide Libraries | PEPotec SRM Peptide Libraries | HeavyPeptide AQUA Peptide Synthesis Service | |
---|---|---|---|---|---|
Primary application | Basic research or biomarker discovery | Fluorescence imaging | Epitope mapping, vaccine development | Discovery | Quantitative mass spectrometry |
Key features | Over 400 modifications available | High performing dyes | High throughput, low toxicity | High throughput | High quality and concentration precision |
Peptide length (amino acids) | 2–110* | 6–70* | 6–25 | 6–25 | 6–30* |
Scale | 1 mg–1 kg+ | 1–10 mg | 1 mg | >0.1 mg | Multiple scales available |
Purity | Crude to 98% | Crude to 98% | Crude | Crude | 95–97% |
QC | MS and HPLC | MS and HPLC | MS | MS check or MS analysis | MS, analytical HPLC, AAA |
Delivery time | 2–3 weeks | 2–3 weeks | 4–5 weeks | 2–3 weeks | 5–7 weeks standard AQUA 2–3 weeks AQUA express** |
* Please inquire about shorter or longer peptide lengths.
**AQUA Express requests are available for up to 10 sequences on a single order and must be approved for express turnaround time.
Type | Name | Abbreviation or alternative name | Position within sequence | Graphical representation of available positions |
---|---|---|---|---|
Misc | 1-Adamantanecarbonyl - | 1-Adamantanecarbonyl-NH | N-term | ■□□□...□□□□ |
Misc | 2,4,6-Trinitrophenyl - TNP | TNP-NH | N-term | ■□□□...□□□□ |
Misc | 2-Furoyl - | 2-Furoyl-NH | N-term | ■□□□...□□□□ |
Misc | 2-Naphtoic acid - | Naphtyl-NH | N-term | ■□□□...□□□□ |
Misc | 3-(Maleimido)propionyl - | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | 3,5-Diiodo-Tyrosine - | [Y(I)2] | Internal | ■■■■...■■■■ |
Misc | 3-Chloro-Tyrosine - | [Y(Cl)] | Internal | ■■■■...■■■■ |
Misc | 3-nitro-2-pyridylthio-L-Cys - | [C(Npys)] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Indole - | [3-Nitro-Indole] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Pyrrol - | [3-Nitro-Pyrrol] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Tyrosine - | [Y(NO2)] | Internal | ■■■■...■■■■ |
Misc | 4-Methylthio-2-oxobutanoic ac. - | [H3C-S-CH2-CH2-CO-CO-NH] | N-term | ■□□□...□□□□ |
Misc | 4-nitrobenzo-2-oxa-1,3-diazole - NBD | NBD-NH | N-term | ■□□□...□□□□ |
Misc | 4-Pentynoic acid - | 4-Pentynoic acid-NH | N-term | ■□□□...□□□□ |
Misc | 4-Phenylazophenol - O-PAP | CO-PAP | C-term | □□□□...□□□■ |
Misc | 6-Bromo-D/L-Tryptophan - | [D/L-W(Br)] | Internal | ■■■■...■■■■ |
Misc | 6-Cloro-Tryptophan - | [W(Cl)] | Internal | ■■■■...■■■■ |
Misc | Acetylation - | Acetyl-NH | N-term | ■□□□...□□□□ |
Misc | Aldehyde - | CHO | C-term | □□□□...□□□■ |
Misc | Amidation - | CONH2 | C-term | □□□□...□□□■ |
Misc | Amino nitrophenyl prop. acid - | [Amino nitrophenyl prop. acid] | Internal | ■■■■...■■■■ |
Misc | Amino-nitrophenyl propion. ac. - ANP-linker | [ANP] | Internal | ■■■■...■■■■ |
Misc | Aminooxyacetyl - | H2N-O-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Benzoyl - Bz | Benzoyl-NH | N-term | ■□□□...□□□□ |
Misc | Benzyloxycarbonyl - Cbz, Z | Z-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - | Biotin-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - Ethylenediamin-Biotin | CO-NH-CH2-CH2-NH-Biotin | C-term | □□□□...□□□■ |
Misc | Biotin-SS-linker - | Biotin-SS-NH | N-term | ■□□□...□□□□ |
Misc | Bis(aminoethyl)ethylene glycol - | CO-NH-(CH2-CH2-O)2-CH2-CH2-NH2 | C-term | □□□□...□□□■ |
Misc | Brom acetic acid - | BrCH2CO-NH | N-term | ■□□□...□□□□ |
Misc | Chlormethylketon - CMK | CMK | C-term | □□□□...□□□■ |
Misc | Cys(4-Methoxytrityl) - Cys(MMT) | [C(Mmt)] | Internal | ■■■■...■■■■ |
Misc | Cys(Acetamidomethyl) - Cys(Acm) | [C(Acm)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carbamidomethyl) - Cys(CAM) / Cys(CH2CONH2) | [C(Cam)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carboxymethyl) - Cys(CMC) / Cys(CH2COOH) | [C(Cmc)] | Internal | ■■■■...■■■■ |
Misc | Cys(di(palmitoyloxy)-propyl) - Cys(Pam2) | [C(Pam2)] | Internal | ■■■■...■■■■ |
Misc | Cys(Farnesyl) - | [C(Farnesyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(HRP) - Cys(horseradish peroxidase) | [C(HRP)] | Internal | ■■■■...■■■■ |
Misc | Cys(MTSL) - | [C(MTSL)] | Internal | ■■■■...■■■■ |
Misc | Cys(Myristyl) - Cys(CH2(CH2)12CH3) | [C(Myristyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(Palmitoyl) - | [C(Palm)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Butyl) - | [C(S-But)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Ethyl) - | [C(S-Et)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isobutyl) - | [C(S-Isobut)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isopropyl) - | [C(S-Isoprop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Methyl) - | [C(S-Me)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Pentyl) - | [C(S-Pentyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Propyl) - | [C(S-Prop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-tert-Butyl) - | [C(S-tert-But] | Internal | ■■■■...■■■■ |
Misc | Cysteamide - | Cysteamide | C-term | □□□□...□□□■ |
Misc | Decanoyl - | Decanoyl-NH | N-term | ■□□□...□□□□ |
Misc | Desthiobiotin - | Desthiobiotin-NH | N-term | ■□□□...□□□□ |
Misc | Digoxigenin - | Digoxigenin-NH | N-term | ■□□□...□□□□ |
Misc | DOTA - | [DOTA] | N-term | ■□□□...□□□□ |
Misc | Ethylamide - N-Ethyl | CONH(C2H5) | C-term | □□□□...□□□■ |
Misc | Ethylester - O-Ethyl | COOEt | C-term | □□□□...□□□■ |
Misc | Fmoc - | Fmoc-NH | N-term | ■□□□...□□□□ |
Misc | Formyl - | Formyl-NH | N-term | ■□□□...□□□□ |
Misc | Glu(Arg) - Arg at side chain of Glu | [Glu(Arg)] | Internal | ■■■■...■■■■ |
Misc | Homoserine lactone - | [Homoser lactone] | C-term | □□□□...□□□■ |
Misc | HRP - | CO-NH -HRP | C-term | □□□□...□□□■ |
Misc | HRP - | HRP-NH | N-term | ■□□□...□□□□ |
Misc | Hydrazide - | CO-NH -NH2 | C-term | □□□□...□□□■ |
Misc | Hydrazine-succinyl - | H2N-NH-CO-CH2-CH2-CO-NH- | N-term | ■□□□...□□□□ |
Misc | Iodoacetyl - | IH2C-CO- | N-term | ■□□□...□□□□ |
Misc | Lys(2-Aminooxy acetic acid) - Lys(Aoa) | [K(Aoa)] | Internal | ■■■■...■■■■ |
Misc | Lys(Acetyl) - | [K(Ac)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-Biotin) - | [K(Ahx-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-SS-Biotin) - | [K(Ahx-SS-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Biotin) - | [K(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Boc) - | [K(Boc)] | Internal | ■■■■...■■■■ |
Misc | Lys(Butyryl) - | [K(Butyryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Desthiobiotin) - | [K(Desthiobiotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Digoxigenin] - | [K(Digoxigenin)] | Internal | ■■■■...■■■■ |
Misc | Lys(DST-PEG 2-Biotin) - | [K(DST-PEG 2-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ferrocene) - | [K(Ferrocene)] | Internal | ■■■■...■■■■ |
Misc | Lys(Formyl) - | [K(Formyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(GG) - Ubiquitin | [K(GG)] | Internal | ■■■■...■■■■ |
Misc | Lys(GGRLR) - | [K(GGRLR)] | Internal | ■■■■...■■■■ |
Misc | Lys(Glutaryl) - | [K(Glutaryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Lipoyl) - | [K(Lipoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(NIP) - | [K(NIP)] | Internal | ■■■■...■■■■ |
Misc | Lys(Palmitoyl) - | [K(Palmitoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Propionyl) - | [K(Propionyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Pyrofolate) - | [K(Pyrofolate)] | Internal | ■■■■...■■■■ |
Misc | Lys(Stearyl) - | [K(Stearyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Succinyl) - | [K(Succinyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(TFA) - Lysine(Trifluoroacetyl) | [K(TFA)] | Internal | ■■■■...■■■■ |
Misc | Maleimide - via SMCC | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | Methanethiosulfonate - MTS | MTS-NH | N-term | ■□□□...□□□□ |
Misc | Methoxysuccinyl - | H3C-O-CO-CH2-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Methylamide - N-Methyl | CO-NH(CH3) | C-term | □□□□...□□□■ |
Misc | Methylester - O-Methyl | COOMe | C-term | □□□□...□□□■ |
Misc | Myristoyl - | Myristoyl-NH | N-term | ■□□□...□□□□ |
Misc | N-Hydroxysuccinimid ester - NHS | CO-NHS | C-term | □□□□...□□□■ |
Misc | Orn(Biotin) - | [Orn(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Palmitoyl - | Palmitoyl-NH | N-term | ■□□□...□□□□ |
Misc | para-Bromo-D-Phenylalanine - | [D-F(4-Br)] | Internal | ■■■■...■■■■ |
Misc | para-Chloro-Phenylalanine - | [F(4-Cl)] | Internal | ■■■■...■■■■ |
Misc | para-Iodo-Phenylalanine - | [F(I)] | Internal | ■■■■...■■■■ |
Misc | para-Nitro-Phenylalanine - | [F(NO2)] | Internal | ■■■■...■■■■ |
Misc | Phenylacetic acid - Paa | Paa-NH | N-term | ■□□□...□□□□ |
Misc | Photo-Leucine - | [Photo-L] | Internal | ■■■■...■■■■ |
Misc | Photo-Methionine - | [Photo-M] | Internal | ■■■■...■■■■ |
Misc | p-tosyl-Gly - | [p-tosyl-G] | N-term | ■□□□...□□□□ |
Misc | S-Acetylmercaptosuccinyl - SAMSA | SAMSA-NH | N-term | ■□□□...□□□□ |
Misc | Ser(Acetyl) - | [S(Ac)] | Internal | ■■■■...■■■■ |
Misc | Ser(Octanoyl) - | [S(Oct)] | Internal | ■■■■...■■■■ |
Misc | Stearyl - | Stearyl-NH | N-term | ■□□□...□□□□ |
Misc | Succinyl - | Succinyl-NH | N-term | ■□□□...□□□□ |
Misc | t-Butylester - | COOtBu | C-term | □□□□...□□□■ |
Misc | tert. Butoxycarbonyl - Boc | Boc-NH | N-term | ■□□□...□□□□ |
Misc | Thiobenzyl - | CO-S(Bzl) | C-term | □□□□...□□□■ |
Misc | Thiopropionyl - Tpa | Tpa-NH | N-term | ■□□□...□□□□ |
Misc | trans-Cinnamoyl - | Cinnamoyl-NH | N-term | ■□□□...□□□□ |
Misc | Tyr(Palmitoyl) - | [Y(Palm)] | Internal | ■■■■...■■■■ |
Dye | (2,4-Dinitrophenyl)-Glycine - (2,4-DNP)-Gly | DNP-NH-G | N-term | ■□□□...□□□□ |
Dye | 2,4-Dinitrophenyl - DNP | CO-DNP | C-term | □□□□...□□□■ |
Dye | 2,4-Dinitrophenyl - DNP | DNP-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Carboxyfluorescein - 5/6-FLUO | 5/6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Tetramethylrhodamine - 5/6-TAMRA | 5/6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 5-Carboxyfluorescein - 5-FLUO | 5-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5-Tetramethylrhodamine - 5-TAMRA | 5-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 6-Carboxyfluorescein - 6-FLUO | 6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 6-Tetramethylrhodamine - 6-TAMRA | 6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 7-Amino-4-methylcoumarin - AMC | CO-AMC | C-term | □□□□...□□□■ |
Dye | Ala(2-Bacd) - benzo[b]acridin-12(5H)-on-2-yl | [Ala(2-Bacd)] | Internal | ■■■■...■■■■ |
Dye | Amino-trifluoromethyl-coumarin - AFC | CO-AFC | C-term | □□□□...□□□■ |
Dye | ATTO 647N - | ATTO 647N-NH | N-term | ■□□□...□□□□ |
Dye | Cy7 - | Cy7-NH | N-term | ■□□□...□□□□ |
Dye | Cys(5/6-TAMRA) - | [C(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(5-TAMRA) - | [C(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(6-TAMRA) - | [C(6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(acetamido-fluorescein) - | [C(acetamido-fluorescein)] | Internal | ■■■■...■■■■ |
Dye | Cys(AEDANS) - | [C(AEDANS)] | Internal | ■■■■...■■■■ |
Dye | Cys(Allophycocyanine) - | [C(Allophycocyanine)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 555) - | [C(DY 555)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 647P1) - | [C(DY 647P1)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 649) - | [C(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Cys(DYQ660) - | [Cys(DYQ660)] | Internal | ■■■■...■■■■ |
Dye | Dabcyl - | Dabcyl-NH | N-term | ■□□□...□□□□ |
Dye | Dabsyl - | Dabsyl-NH | N-term | ■□□□...□□□□ |
Dye | Dansyl - | CO-Dansyl | C-term | □□□□...□□□■ |
Dye | Dansyl - | Dansyl-NH | N-term | ■□□□...□□□□ |
Dye | DAP(DNP) - DNP at Diaminopropionic acid | [DAP(DNP)] | Internal | ■■■■...■■■■ |
Dye | DAP(DY 647) - DY 647 at Diaminopropionic ac. | [DPA(DY-647)] | Internal | ■■■■...■■■■ |
Dye | DAP(DyLight 488) - DyLight 488 at Diaminoprop. ac | [DPA(DY 488)] | Internal | ■■■■...■■■■ |
Dye | DAP(NMA) - NMA at Diaminopropionic acid | [DAP(NMA)] | Internal | ■■■■...■■■■ |
Dye | Dinitrophenyl-Ethylenediamine - EDDnp | CO-EDDnp | C-term | □□□□...□□□■ |
Dye | D-Lys(5/6-Fluorescein) - | [D-K(5/6-Fluorescein)] | Internal | ■■■■...■■■■ |
Dye | D-Lys(5/6-TAMRA) - | [D-Lys(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | DY 495 - | DY 495-NH | N-term | ■□□□...□□□□ |
Dye | DY 505 - | DY 505-NH | N-term | ■□□□...□□□□ |
Dye | DY 530 - | DY 530-NH | N-term | ■□□□...□□□□ |
Dye | DY 547 - | DY 547-NH | N-term | ■□□□...□□□□ |
Dye | DY 549 - | DY 549-NH | N-term | ■□□□...□□□□ |
Dye | DY 630 - | DY630-NH | N-term | ■□□□...□□□□ |
Dye | DY 633 - | DY 633-NH | N-term | ■□□□...□□□□ |
Dye | DY 635 - | DY 635-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - | DY 647-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - aminomodified | CO-DY 647 | C-term | □□□□...□□□■ |
Dye | DY 649 - | DY 649-NH | N-term | ■□□□...□□□□ |
Dye | DY 649P1 - aminomodified | CO-DY 649P1 | N-term | ■□□□...□□□□ |
Dye | DY 650 - | DY650-NH | Internal | ■■■■...■■■■ |
Dye | DY 680 - | DY 680-NH | N-term | ■□□□...□□□□ |
Dye | DY 682 - | DY 682-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | DY 750-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | CO-DY 750 | C-term | □□□□...□□□■ |
Dye | DY 751 - | DY 751-NH | N-term | ■□□□...□□□□ |
Dye | DY 800 - | DY 800-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 488 - | DY 488-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 504Q - | DyLight 504Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 543Q - | DyLight 543Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 554-R1 - | DyLight 554-R1-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 683Q - | DyLight 683Q-NH | N-term | ■□□□...□□□□ |
Dye | DYQ 660 - | DYQ 660-NH | N-term | ■□□□...□□□□ |
Dye | EDANS - | CO-EDANS | C-term | □□□□...□□□■ |
Dye | Glu(EDANS) - | [E(EDANS)] | Internal | ■■■■...■■■■ |
Dye | His(DNP) - | [H(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(2-(N-methylamino)benzoyl) - Lys(NMA) | [Lys(NMA)] | Internal | ■■■■...■■■■ |
Dye | Lys(2,4-DNP) - | [K(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-Fluorescein) - | [K(5/6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-TAMRA) - | [K(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-Fluorescein) - | [K(5-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-TAMRA) - | [K(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(6-Fluorescein) - | [K(6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(Ahx-(5-Fluorescein)) - | [K(Ahx-(5-Fluo))] | Internal | ■■■■...■■■■ |
Dye | Lys(ATTO 655) - | [K(ATTO 655)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dabcyl) - | [K(Dabcyl)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dde) - | [Lys(Dde)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 405) - | [K(DY 405)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 495) - | [K(DY 495)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 530) - | [K(DY 530)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 547) - | [K(DY 547)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 590) - | [K(DY 590)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 630) - | [K(DY 630)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 635) - | [K(DY 635)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 647) - | [K(DY 647)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 650) - | [Lys(DY 650)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 800) - | [K(DY 800)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 488) - | [K(DY 488)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 633) - Lys(DY633) | [K(DyLight 633)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 649) - | [K(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Lys(DYQ 3) - | [K(DYQ 3)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOC) - (7-hydroxycoumarin-3-yl)formyl | [Lys(HOC)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOMCA) - | [Lys(HOMCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MCA) - | [K(MCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MOC) - (7-methoxycoumarin-3-yl)formyl | Lys(MOC) | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine B) - | [K(Rhodamine B)] | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine Green) - | [K(Rhodamine Green)] | Internal | ■■■■...■■■■ |
Dye | Methoxycoumarin acetic acid - MCA | CO-MCA | C-term | □□□□...□□□■ |
Dye | Methoxycoumarin acetic acid - MCA | MCA-NH | N-term | ■□□□...□□□□ |
Dye | Ornithine(Coumarin343) - | [Orn(C343)] | Internal | ■■■■...■■■■ |
Dye | para-Nitroanilide - pNA | CO-pNA | C-term | □□□□...□□□■ |
Dye | para-Nitrophenylester - | COONp | C-term | □□□□...□□□■ |
Dye | QSY21 - | QSY21-NH | N-term | ■□□□...□□□□ |
Dye | Rhodamine 110 - | CO-Rhod 110 | C-term | □□□□...□□□■ |
Dye | Rhodamine B - | Rhodamin B-NH | N-term | ■□□□...□□□□ |
Dye | R-Phycoerythrin - | CO- R-Phycoerythrin | C-term | □□□□...□□□■ |
Dye | Tide Quencher 2 - TQ2 | TQ2-NH | N-term | ■□□□...□□□□ |
Linkers | 12-Aminododecanoic acid - Ado | [Ado] | Internal | ■■■■...■■■■ |
Linkers | 5-Aminopentanoic acid - Aminovaleric acid (Ava) | [Ava] | Internal | ■■■■...■■■■ |
Linkers | 6-Aminohexanoic acid (Ahx) - Aminocaproic acid (Aca) | [Ahx] | Internal | ■■■■...■■■■ |
Linkers | 8-Aminooctanoic acid - Aoc | [Aoc] | Internal | ■■■■...■■■■ |
Linkers | Cys(PEG 40000) - | [C(PEG 40000)] | Internal | ■■■■...■■■■ |
Linkers | Methyl-PEG12 - MS(PEG)12 | MS(PEG)12-NH | N-term | ■□□□...□□□□ |
Linkers | Methyl-PEG24 - MS(PEG)24 | MS(PEG)24-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 12 (40 atoms) - Amino-PEG-acid | [PEG 12] | Internal | ■■■■...■■■■ |
Linkers | PEG 2 (9 atoms) - 8-Amino-3,6-dioxaoctanoic acid | [PEG 2] | Internal | ■■■■...■■■■ |
Linkers | PEG 20,000 - | PEG 20,000-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 2000 - Amino-PEG-acid | [PEG 2000] | Internal | ■■■■...■■■■ |
Linkers | PEG 3 (13 atoms) - Amino-trioxadodecanoic acid | [PEG 3] | Internal | ■■■■...■■■■ |
Linkers | PEG 4 (16 atoms) - Amino-PEG-acid | [PEG 4] | Internal | ■■■■...■■■■ |
Linkers | PEG 5 (19 atoms) - Amino-PEG-acid | [PEG 5] | Internal | ■■■■...■■■■ |
Linkers | PEG 5000 - | CO-PEG 5000 | C-term | □□□□...□□□■ |
Linkers | PEG 5000 - Amino-PEG-acid | [PEG 5000] | Internal | ■■■■...■■■■ |
Linkers | PEG 6 (22 atoms) - Amino-PEG-acid | [PEG 6] | Internal | ■■■■...■■■■ |
Linkers | PEG 8 (28 atoms) - Amino-PEG-acid | [PEG 8] | Internal | ■■■■...■■■■ |
Linkers | PEG-4 Azide - | N3-PEG4-NH | N-term | ■□□□...□□□□ |
Special AAs | 2-(4'-Pentenyl)alanine - | [A(pentenyl)] | Internal | ■■■■...■■■■ |
Special AAs | 2,3-Diaminopropionic acid - DAP | [DAP] | Internal | ■■■■...■■■■ |
Special AAs | 2-Amino-4-thiazolinic acid - | [2-Amino-4-thiazolinyl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Indanyl-Glycine - Igl | [Igl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Methyl-D-Tryptophan - | [D-W(2-Me)] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-Alanine - 2-Nal | [2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-D-Alanine - D-2-Nal | [D-2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 3,3-Diphenyl-Alanine - | [A(F2)] | Internal | ■■■■...■■■■ |
Special AAs | 3,4-Dihydroxy-Phenylalanine - | [F(OH)2] | Internal | ■■■■...■■■■ |
Special AAs | 5,5-Dimethyl-Proline - | [P(Me2)] | Internal | ■■■■...■■■■ |
Special AAs | 5-Hydroxy-Tryptophan - | [W(OH)] | Internal | ■■■■...■■■■ |
Special AAs | Allyl-Glycine - 2-Aminopent-4-enoic acid | [G(allyl)] | Internal | ■■■■...■■■■ |
Special AAs | alpha-Allylalanine - | [Allyl-A] | Internal | ■■■■...■■■■ |
Special AAs | alpha-aminoadipic acid - Aad | [Aad] | Internal | ■■■■...■■■■ |
Special AAs | alpha-t-butylglycine - | [G(tBu)] | Internal | ■■■■...■■■■ |
Special AAs | Aminobenzoic acid - Abz | [Abz] | Internal | ■■■■...■■■■ |
Special AAs | Aminobutyric acid - Abu | [Abu] | Internal | ■■■■...■■■■ |
Special AAs | Aminocyclohexyl carbox. acid - Ac6c | [Ac6c] | Internal | ■■■■...■■■■ |
Special AAs | Aminoisobutyric acid - Aib | 0 | Internal | ■■■■...■■■■ |
Special AAs | Asp beta-methyl ester - Asp(OMe) | [D(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Asp(Isopropylamide) - | [D(Isopropylamide)] | Internal | ■■■■...■■■■ |
Special AAs | Azetidine-2-carboxylic acid - Aze | [Aze] | Internal | ■■■■...■■■■ |
Special AAs | Canavanine - Cav | [Cav] | Internal | ■■■■...■■■■ |
Special AAs | Citrulline - | [Cit] | Internal | ■■■■...■■■■ |
Special AAs | Cyclopentyl-Glycine - | [G(cyclopentyl)] | Internal | ■■■■...■■■■ |
Special AAs | Cysteic acid - Cya | [Cya] | Internal | ■■■■...■■■■ |
Special AAs | Cysteine sulfonate - | [C(SO3H)] | Internal | ■■■■...■■■■ |
Special AAs | D-2-Indanyl-Glycine - D-Igl | [D-Igl] | Internal | ■■■■...■■■■ |
Special AAs | D-4-Hydroxyphenly-Glycine - D-Hpg | [D-Hpg] | Internal | ■■■■...■■■■ |
Special AAs | D-Alanine - | *A | Internal | ■■■■...■■■■ |
Special AAs | D-Amino Acid - | [D-aa] | Internal | ■■■■...■■■■ |
Special AAs | D-Arginine - | *R | Internal | ■■■■...■■■■ |
Special AAs | D-Asparagine - | *N | Internal | ■■■■...■■■■ |
Special AAs | D-Aspartic acid - | *D | Internal | ■■■■...■■■■ |
Special AAs | D-Cysteine - | *C | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamic acid - | *E | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamine - | *Q | Internal | ■■■■...■■■■ |
Special AAs | D-Histidine - | *H | Internal | ■■■■...■■■■ |
Special AAs | D-Homocysteine - | [D-Homocys] | Internal | ■■■■...■■■■ |
Special AAs | D-Homoserine - | [D-Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Diaminotrioxatridecan suc. ac. - TTDS | [TTDS] | Internal | ■■■■...■■■■ |
Special AAs | D-Isoleucine - | *I | Internal | ■■■■...■■■■ |
Special AAs | D-Leucine - | *L | Internal | ■■■■...■■■■ |
Special AAs | D-Lysine - | *K | Internal | ■■■■...■■■■ |
Special AAs | D-Methionine - | *M | Internal | ■■■■...■■■■ |
Special AAs | D-Ornithine - | [D-Orn] | Internal | ■■■■...■■■■ |
Special AAs | D-Phenylalanine - | *F | Internal | ■■■■...■■■■ |
Special AAs | D-Proline - | *P | Internal | ■■■■...■■■■ |
Special AAs | D-Serine - | *S | Internal | ■■■■...■■■■ |
Special AAs | D-Threonine - | *T | Internal | ■■■■...■■■■ |
Special AAs | D-Tryptophan - | *W | Internal | ■■■■...■■■■ |
Special AAs | D-Tyrosine - | *Y | Internal | ■■■■...■■■■ |
Special AAs | D-Valine - | *V | Internal | ■■■■...■■■■ |
Special AAs | Formylglycine - | [G(CHO)] | Internal | ■■■■...■■■■ |
Special AAs | gamma-Carboxyglutamic acid - Gla | [Gla] | Internal | ■■■■...■■■■ |
Special AAs | Gamma-Glu, amidated - Gla, amidation at side chain | [Gla amide] | Internal | ■■■■...■■■■ |
Special AAs | Glu gamma-methyl ester - Glu(OMe) | [E(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Homo-Citrulline - | [Homocit] | Internal | ■■■■...■■■■ |
Special AAs | Homophenylalanine - | [HomoF] | Internal | ■■■■...■■■■ |
Special AAs | Homoserine - | [Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Hydroxy-Proline - Hyp | [Hyp] | Internal | ■■■■...■■■■ |
Special AAs | Hyposine - Hpu | [Hpu] | Internal | ■■■■...■■■■ |
Special AAs | Isoaspartic acid - | 0 | Internal | ■■■■...■■■■ |
Special AAs | Kynurenine - Kyn | [Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptoacetic acid - Thioglycolic acid | [Mercaptoacetyl] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptopropionic acid - Mpa | [Mpa] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfone - | [M(O2)] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfoxide - | [M(O)] | Internal | ■■■■...■■■■ |
Special AAs | N-Formylkynurenine - | [Formyl-Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Norleucine - | [Nle] | Internal | ■■■■...■■■■ |
Special AAs | Norvaline - 2-Aminopentanoic acid, Nva | [Nva] | Internal | ■■■■...■■■■ |
Special AAs | Octahydroindole carbox. acid - | [Oic] | Internal | ■■■■...■■■■ |
Special AAs | Ornitine - | [Orn] | Internal | ■■■■...■■■■ |
Special AAs | para-Acetylphenylalanine - pAF | [para-Ac-Phe] | Internal | ■■■■...■■■■ |
Special AAs | Phenylglycine - Phg | [Phg] | Internal | ■■■■...■■■■ |
Special AAs | Pyroglutamic acid - Pyr | [Pyr] | Internal | ■■■■...■■■■ |
Special AAs | Selenocysteine - Sec | [Sec] | Internal | ■■■■...■■■■ |
Special AAs | β-Alanine - | [β-A] | Internal | ■■■■...■■■■ |
Special AAs | β-Cyclohexyl-Alanine - Cha | [Cha] | Internal | ■■■■...■■■■ |
Special AAs | β-cyclohexyl-D-Alanine - D-Cha | [D-Cha] | Internal | ■■■■...■■■■ |
Special AAs | Tetrahydroisquinol.carb.acid - Tic | [Tic] | Internal | ■■■■...■■■■ |
Special AAs | Thiazolidine-4-carboxylic acid - Thz | [Thz] | Internal | ■■■■...■■■■ |
Special AAs | γ-Glutamic acid - | [γ-E] | Internal | ■■■■...■■■■ |
PTM | 1-Methyl-Tryptophan - | [W(1-Me)] | Internal | ■■■■...■■■■ |
PTM | 2,6-Dimethyl-Tyrosine - | [Y(Me2)] | Internal | ■■■■...■■■■ |
PTM | 2-Methylproline - | [P(2-Me)] | Internal | ■■■■...■■■■ |
PTM | 2-Methyl-Tryptophan - | [2-Me-Trp] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (asym.) - | [R(Me)2-asym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (sym.) - | [R(Me)2-sym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Cysteine - Penicillamine | [Pen] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Lysine - | [K(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Phenylalanine - | [F(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Valine - | [V(Me)2] | Internal | ■■■■...■■■■ |
PTM | Methyl-Arginine - | [R(Me)] | Internal | ■■■■...■■■■ |
PTM | Methyl-Lysine - | [K(Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Alanine - | [Ala(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Glycine - | [G(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Leucine - | [L(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Phenylalanine - | [F(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Tyrosine - | [Y(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Valine - | [V(N-Me)] | Internal | ■■■■...■■■■ |
PTM | Phosphono-Phenylalanine - | [F(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Serine - | [S(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Threonin - | [T(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Tyrosin - | [Y(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | β-GalNAc-Threonine - | [T(β-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Asparagine - | [N(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Serine - | [S(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Threonine - | [T(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Serine - | [S(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Threonine - | [T(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Tyrosine - | [Y(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Arginine - | [R(Me3)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Lysine - | [K(Me)3] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Serine - | [S(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Threonine - | [T(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Serine - | [S(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Threonine - | [T(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Aspartic acid - | [D(α-Me)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Glutamic acid - | [E(α-Me)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (13C3;15N) - | [A(13C3; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (D3) - | [A(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6) - | [R(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6;15N4) - | [R(13C6; 15N4)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asn (13C4;15N2) - | [N(13C4; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asp (13C4;15N - | [D(13C4; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gln (13C5;15N2) - | [Q(13C5;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Glu (13C5;15N) - | [E(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gly (13C2;15N) - | [G(13C2; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy His (15N3) - | [His(15N3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ile (13C6;15N) - | [I(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C1) - | [L(13C1)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6) - | [L(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6;15N) - | [L(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D10) - | [L(D10)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D3) - | [L(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6) - | [K(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6;15N2) - | [K(13C6; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Phe (13C9;15N) - | [F(13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Pro (13C5;15N) - | [P(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (13C3;15N) - | [S (13C3;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (D3) - | [S(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Thr(13C4;15N) - | [T(13C4;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Trp (13C11;15N2) - | [W (13C11;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Tyr (13C9;15N) - | [Y (13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5) - | [V(13C5)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5;15N) - | [V(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | L-tyrosine-phenyl-d4 - deuterated Tyr | [Tyr(D4)] | Internal | ■■■■...■■■■ |
Isotopic | TMT6-126 - | TMT6-126-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-127 - | TMT6-127-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-128 - | TMT6-128-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-129 - | TMT6-129-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-130 - | TMT6-130-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-131 - | TMT6-131-NH | N-term | ■□□□...□□□□ |
Immunization | Blue Carrier Protein - | CO-NH -Blue Carrier Protein | C-term | □□□□...□□□■ |
Immunization | BSA - | CO-BSA | C-term | □□□□...□□□■ |
Immunization | BSA via Glutaraldehyde - | BSA-NH | N-term | ■□□□...□□□□ |
Immunization | Cys(BSA) - | [C(BSA)] | Internal | ■■■■...■■■■ |
Immunization | Cys(KLH) - | [C(KLH)] | Internal | ■■■■...■■■■ |
Immunization | Cys(OVA) - | [C(OVA)] | Internal | ■■■■...■■■■ |
Immunization | KLH - | CO-KLH | C-term | □□□□...□□□■ |
Immunization | KLH via Glutaraldehyde - | KLH-NH | N-term | ■□□□...□□□□ |
Immunization | Lys(BSA) - | [K(BSA] | Internal | ■■■■...■■■■ |
Immunization | Lys(KLH) - | [K(KLH] | Internal | ■■■■...■■■■ |
Immunization | MAP 16 - Crude 8-20 aa, at C-term. only | MAP 16 | C-term | □□□□...□□□■ |
Immunization | MAP 4 - Crude 8-20 aa, at C-term. only | MAP 4 | C-term | □□□□...□□□■ |
Immunization | MAP 8 - Crude 8-20 aa, at C-term. only | MAP 8 | C-term | □□□□...□□□■ |
Immunization | OVA - | CO-OVA | C-term | □□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | 3-Azido-Alanine - Aza | [Aza] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | 5-Azidopentanoyl - 5-Azidovalerianoyl | 5-Azidopentanoyl-NH | N-term | ■□□□...□□□□ |
Cyclization/Dimerization/Special Synthesis | 6-Azido-Norleucine - Nle(N3) | [Nle(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azidohomoalanine - Aha/ Dab(N3) | [Aha] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azido-Lysine - | [K(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (C-C), disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (D-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (E-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (K-C-terminus) - side chain of K to C-terminus | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (NH-CO), homodetic - | [ ] | N-term | ■□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | Double disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | D-Propargylglycine - D-Pra | [D-Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Hetero-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Homo-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Mixed amino acids - | [X] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | para-Azido-Phenylalanine - | [F(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylalanine - alpha-Me-L-Ala(Pentynyl)-OH | [Propargylala] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylglycine - Pra | [Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Asymmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Symmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Site directed disulfide bridge - Cyclisation (C-C) | [Cys cyclo(S-S)] | Internal | ■■■■...■■■■ |
A. The shelf life is >24 months at –20°C for lyophilized peptides protected by argon in the original non-open container. However, it is sequence dependent and may be shorter for some sequences.
A. Unless otherwise requested, all peptides are delivered as lyophilized trifluoroacetic salts with free amino and carboxy termini. N-terminal acetylation or C-terminal amidation is available for a small fee but must be requested when ordering your peptide(s).
A. The ability to synthesize the peptide and its solubility, stability, and overall charge are discussed in the Protein Biology Learning Center under the Peptide Design.
A. All the information required to get an accurate quote is included in the Quote Request form.
For Research Use Only. Not for use in diagnostic procedures.