Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Simultaneous detection of expression of five genes in a whole-mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes. Red: sog labeled using aminoallyl UTP (Cat. No. A21663, A32765) and Alexa Fluor® 647 succinimidyl ester (Cat. No. A20006, A20106). Green: ind labeled with DNP, followed by rabbit anti–dinitrophenyl-KLH IgG antibody (Cat. No. A6430) prelabeled with the Zenon® Alexa Fluor® 555 Rabbit IgG Labeling Kit (Cat. No. Z25305). Blue: en labeled with biotin and detected with HRP–streptavidin and Alexa Fluor® 405 tyramide (TSA™ Kit #39, T30952). Yellow: wg labeled with digoxigenin and detected with sheep anti–digoxigenin IgG antibody and Alexa Fluor® 594 Donkey Anti–Sheep IgG antibody (Cat. No. A11016). Magenta: msh labeled with fluorescein and detected with mouse anti–fluorescein/Oregon Green® IgG2a antibody (Cat. No. A6421) and Alexa Fluor® 488 Goat Anti–Mouse IgG antibody (Cat. No. A11001, A11029). Image contributed by Dave Kosman and Ethan Bier, University of California, San Diego.
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Endothelial cells. Go ›
Labeled paint probes hybridized to human metaphase chromosomes Go ›
Simultaneous detection of three gene targets in a whole-mount Drosophila embryo by fluorescence in situ hybridization. Go ›
Immunocytochemistry using Alpha-Tubulin Monoclonal Antibody, Mouse Go ›
Tyramide signal amplification of immunofluorescent staining in mouse brain sections. Go ›
Fixed and permeabilized bovine pulmonary artery endothelial cells stained with Mouse anti-ATP Synthase Subunit IF1 Monoclonal Antibody. Go ›
Fixed and permeabilized HeLa cells stained with Alexa Fluor® 350 phalloidin and Mouse anti ATP Synthase Subunit IF1 Monoclonal Antibody to label actin filaments and the mitochondria, respectively. Go ›