Scale your COVID-19 testing with our high-throughput molecular diagnostic test kits
The Applied Biosystems TaqPath COVID-19 HT Kit and the TaqPath COVID-19, Flu A/B, RSV HT Kit are intended for use with the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit, High-Throughput, and are compatible with the Amplitude Platform to enable a high-throughput workflow for the analysis of 1,000 to 8,000 COVID-19 specimens in 24 hours. Each kit contains 20,000 reactions to maximize specimen throughput and help laboratories quickly scale COVID-19 diagnostic testing to volumes needed to meet demand. 
TaqPath COVID-19 HT Kit
TaqPath COVID-19 HT Kit
Create peace of mind with automated COVID-19 testing that is highly sensitive and specific. This test uses a multi-target design that includes an S-gene target. If a sample with the 69-70del S-gene mutation is tested using the TaqPath COVID-19 HT Kit, it will result in an S-gene dropout. The European CDC and WHO have noted this pattern of detection may help with early identification of the Omicron variant (B.1.1.529).
What is included in the kit:
  • TaqPath COVID‑19 Module 1 MS2 Phage Control—an internal process control for RNA extraction
  • TaqPath COVID‑19 Module 2 Assay Kit—multiplexed assays that contain three primer/probe sets specific to three different SARS-CoV-2 genomic regions (ORF1ab, N gene, and S gene) and primers/probes for bacteriophage MS2
  • TaqPath COVID‑19 Module 2 IVT RNA Control—an RNA control that contains targets specific to the SARS-CoV-2 genomic regions targeted by the assays
  • TaqPath COVID‑19 Module 2 Control Dilution Buffer
  • TaqPath HT Module 2 1-Step Multiplex Master Mix (No ROX)
  • TaqPath HT Module 2 Empty 5 mL Mixing Tubes
  • HT Water for Dilution
Download flyer
covid_flu_kit_4 (1)
TaqPath COVID-19, Flu A/B, RSV HT Kit
The kit contains the reagents and controls for a real-time reverse transcription polymerase chain reaction (RT-PCR) test intended for the qualitative detection of nucleic acid from SARS-CoV-2, influenza A/B, and/or Respiratory Syncytial Virus (RSV) in nasopharyngeal, nasal, and oropharyngeal swabs from individuals suspected of having a respiratory tract infection.
What is included in the kit:
  • TaqPath COVID-19 Module 1 MS2 Phage Control—an internal process control for RNA extraction 
  • TaqPath COVID-19, Flu A/B, RSV Module 2 Assay Kit—multiplexed assays that contain primer and probe sets specific to the following targets
    • Two SARS-CoV-2 targets (N gene and S gene)
    • Two influenza targets (influenza A and influenza B)
    • Two RSV targets (RSV A and RSV B)
    • Bacteriophage MS2
  • TaqPath COVID-19, Flu A/B, RSV Module 2 IVT RNA Control—an RNA control that contains targets specific to the SARS-CoV-2, influenza A/B, and RSV A/B genomic regions targeted by the assays
  • TaqPath COVID-19 Module 2 control Dilution Buffer
  • TaqPath HT Module 2 1-Step Multiplex Master Mix (No ROX)
  • TaqPath HT Module 2 Empty 5 mL Mixing Tubes
  • HT Water for dilution
Download flyer
Contact us
For In Vitro Diagnostic Use.
Regulatory requirements vary by country; products may not be available in your geographic area.