Search Thermo Fisher Scientific
Bovine pulmonary artery endothelial (BPAEC) cells were initially stained with the reactive oxygen species (ROS) indicator, 6-carboxy-2',7'-dichlorodihydrofluorescein diacetate, di(acetoxymethyl ester) (C2938). After a 30-minute incubation, the cells were washed and then incubated simultaneously with FM 5-95 (T23360) and Hoechst 33342 (H1399, H3570, H21492) in phosphate-buffered saline (PBS) for an additional five minutes before washing and mounting in PBS. The red-fluorescent FM 5-95 appears to stain both the plasma membrane and early endosomes; the green-fluorescent, oxidized carboxydichlorofluorescein localizes to the cytoplasm; and the blue-fluorescent Hoechst 33342 dye stains the nucleus.
Oxidative stress analysis in BPAE and Raw cells using CellROX® Deep Red Reagent (Cat. No. C10422) Go ›
BPAE detection of oxidative stress using MitoTracker® Green (Cat. No. M7514) and CellROX® Deep Red Reagent (Cat. No. C10422) Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®. Go ›
Immunofluorescent Staining using Mouse HuC/HuD Neuronal Protein Monoclonal Antibody, Biotin-XX Conjugate. Go ›