Product References
Bone Marrow Mesenchymal Stem Cell-Derived Dermcidin-Containing Migrasomes enhance LC3-Associated Phagocytosis of Pulmonary Macrophages and Protect against Post-Stroke Pneumonia.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Li T,Su X,Lu P,Kang X,Hu M,Li C,Wang S,Lu D,Shen S,Huang H,Liu Y,Deng X,Cai W,Wei L,Lu Z
Tue Aug 01 00:00:00 EDT 2023
Thioredoxin interacting protein protects mice from fasting induced liver steatosis by activating ER stress and its downstream signaling pathways.
Scientific reports
Miyahara H,Hasegawa K,Yashiro M,Ohara T,Fujisawa M,Yoshimura T,Matsukawa A,Tsukahara H
Mon Mar 21 00:00:00 EDT 2022
Phosphorylated STYK1 restrains the inhibitory role of EGFR in autophagy initiation and EGFR-TKIs sensitivity.
Cell insight
Zhou C,Dong X,Wang M,Qian X,Hu M,Liang K,Liang Y,Zhang R,Huang Y,Lyu H,Xiao S,Tang Y,Ali DW,Michalak M,Chen XZ,Tang J
Mon Aug 01 00:00:00 EDT 2022
Phosphorylated STYK1 restrains the inhibitory role of EGFR in autophagy initiation and EGFR-TKIs sensitivity.
Cell insight
Zhou C,Dong X,Wang M,Qian X,Hu M,Liang K,Liang Y,Zhang R,Huang Y,Lyu H,Xiao S,Tang Y,Ali DW,Michalak M,Chen XZ,Tang J
Mon Aug 01 00:00:00 EDT 2022
Phosphorylated STYK1 restrains the inhibitory role of EGFR in autophagy initiation and EGFR-TKIs sensitivity.
Cell insight
Zhou C,Dong X,Wang M,Qian X,Hu M,Liang K,Liang Y,Zhang R,Huang Y,Lyu H,Xiao S,Tang Y,Ali DW,Michalak M,Chen XZ,Tang J
Mon Aug 01 00:00:00 EDT 2022
Targeting CRL4 suppresses chemoresistant ovarian cancer growth by inducing mitophagy.
Signal transduction and targeted therapy
Meng Y,Qiu L,Zeng X,Hu X,Zhang Y,Wan X,Mao X,Wu J,Xu Y,Xiong Q,Chen Z,Zhang B,Han J
Fri Dec 09 00:00:00 EST 2022
Betulinic acid-nucleoside hybrid prevents acute alcohol -induced liver damage by promoting anti-oxidative stress and autophagy.
European journal of pharmacology
Zheng LY,Zou X,Wang YL,Zou M,Ma F,Wang N,Li JW,Wang MS,Hung HY,Wang Q
Wed Jan 05 00:00:00 EST 2022
Protective autophagy attenuates soft substrate-induced apoptosis through ROS/JNK signaling pathway in breast cancer cells.
Free radical biology & medicine
Chen Y,Li P,Peng Y,Xie X,Zhang Y,Jiang Y,Li T,Qin X,Li S,Yang H,Wu C,Zheng C,Zhu J,You F,Liu Y
Fri Aug 20 00:00:00 EDT 2021
Autophagic elimination of ribosomes during spermiogenesis provides energy for flagellar motility.
Developmental cell
Lei Y,Zhang X,Xu Q,Liu S,Li C,Jiang H,Lin H,Kong E,Liu J,Qi S,Li H,Xu W,Lu K
Mon Aug 23 00:00:00 EDT 2021
IKKβ activation promotes amphisome formation and extracellular vesicle secretion in tumor cells.
Biochimica et biophysica acta. Molecular cell research
Peng X,Yang L,Ma Y,Li X,Yang S,Li Y,Wu B,Tang S,Zhang F,Zhang B,Liu J,Li H
Fri Jan 01 00:00:00 EST 2021
MicroRNA-375 Targets ATG14 to Inhibit Autophagy and Sensitize Hepatocellular Carcinoma Cells to Sorafenib.
OncoTargets and therapy
Yang S,Wang M,Yang L,Li Y,Ma Y,Peng X,Li X,Li B,Jin H,Li H
Thu Apr 14 00:00:00 EDT 2022
LncRNA PVT1 promotes gemcitabine resistance of pancreatic cancer via activating Wnt/β-catenin and autophagy pathway through modulating the miR-619-5p/Pygo2 and miR-619-5p/ATG14 axes.
Molecular cancer
Zhou C,Yi C,Yi Y,Qin W,Yan Y,Dong X,Zhang X,Huang Y,Zhang R,Wei J,Ali DW,Michalak M,Chen XZ,Tang J
Wed Jul 29 00:00:00 EDT 2020
miR-29c-3p inhibits autophagy and cisplatin resistance in ovarian cancer by regulating FOXP1/ATG14 pathway.
Cell cycle (Georgetown, Tex.)
Hu Z,Cai M,Zhang Y,Tao L,Guo R
Wed Jan 01 00:00:00 EST 2020
Aberrant miR-1246 expression promotes radioresistance in non-small cell lung cancer: a potential prognostic biomarker and radiotherapy sensitization target.
American journal of cancer research
Fan L,Wang J,Cao Q,Ding X,Li B
Mon Nov 13 00:00:00 EST 2023
STYK1 promotes autophagy through enhancing the assembly of autophagy-specific class III phosphatidylinositol 3-kinase complex I.
Autophagy
Zhou C,Qian X,Hu M,Zhang R,Liu N,Huang Y,Yang J,Zhang J,Bai H,Yang Y,Wang Y,Ali D,Michalak M,Chen XZ,Tang J
Thu Oct 01 00:00:00 EDT 2020
Sequential formation of different layers of dystrophic neurites in Alzheimer's brains.
Molecular psychiatry
Sharoar MG,Hu X,Ma XM,Zhu X,Yan R
Sun Sep 01 00:00:00 EDT 2019
Spatiotemporal Control of ULK1 Activation by NDP52 and TBK1 during Selective Autophagy.
Molecular cell
Vargas JNS,Wang C,Bunker E,Hao L,Maric D,Schiavo G,Randow F,Youle RJ
Thu Apr 18 00:00:00 EDT 2019
Spatiotemporal Control of ULK1 Activation by NDP52 and TBK1 during Selective Autophagy.
Molecular cell
Vargas JNS,Wang C,Bunker E,Hao L,Maric D,Schiavo G,Randow F,Youle RJ
Thu Apr 18 00:00:00 EDT 2019
miR199a-5p inhibits hepatic insulin sensitivity via suppression of ATG14-mediated autophagy.
Cell death & disease
Li B,Wu X,Chen H,Zhuang C,Zhang Z,Yao S,Cai D,Ning G,Su Q
Wed Mar 14 00:00:00 EDT 2018
miR199a-5p inhibits hepatic insulin sensitivity via suppression of ATG14-mediated autophagy.
Cell death & disease
Li B,Wu X,Chen H,Zhuang C,Zhang Z,Yao S,Cai D,Ning G,Su Q
Wed Mar 14 00:00:00 EDT 2018
RUBCN/rubicon and EGFR regulate lysosomal degradative processes in the retinal pigment epithelium (RPE) of the eye.
Autophagy
Muniz-Feliciano L,Doggett TA,Zhou Z,Ferguson TA
Fri Jul 05 00:00:00 EDT 2019
RUBCN/rubicon and EGFR regulate lysosomal degradative processes in the retinal pigment epithelium (RPE) of the eye.
Autophagy
Muniz-Feliciano L,Doggett TA,Zhou Z,Ferguson TA
Fri Jul 05 00:00:00 EDT 2019
Sensitization of tamoxifen-resistant breast cancer cells by Z-ligustilide through inhibiting autophagy and accumulating DNA damages.
Oncotarget
Qi H,Jiang Z,Wang C,Yang Y,Li L,He H,Yu Z
Tue Apr 25 00:00:00 EDT 2017
Sensitization of tamoxifen-resistant breast cancer cells by Z-ligustilide through inhibiting autophagy and accumulating DNA damages.
Oncotarget
Qi H,Jiang Z,Wang C,Yang Y,Li L,He H,Yu Z
Tue Apr 25 00:00:00 EDT 2017
Simultaneous inhibition of the ubiquitin-proteasome system and autophagy enhances apoptosis induced by ER stress aggravators in human pancreatic cancer cells.
Autophagy
Li X,Zhu F,Jiang J,Sun C,Zhong Q,Shen M,Wang X,Tian R,Shi C,Xu M,Peng F,Guo X,Hu J,Ye D,Wang M,Qin R
Thu Sep 01 00:00:00 EDT 2016
Simultaneous inhibition of the ubiquitin-proteasome system and autophagy enhances apoptosis induced by ER stress aggravators in human pancreatic cancer cells.
Autophagy
Li X,Zhu F,Jiang J,Sun C,Zhong Q,Shen M,Wang X,Tian R,Shi C,Xu M,Peng F,Guo X,Hu J,Ye D,Wang M,Qin R
Thu Sep 01 00:00:00 EDT 2016
Lentivirus-mediated Bos taurus bta-miR-29b overexpression interferes with bovine viral diarrhoea virus replication and viral infection-related autophagy by directly targeting ATG14 and ATG9A in Madin-Darby bovine kidney cells.
The Journal of general virology
Fu Q,Shi H,Ni W,Shi M,Meng L,Zhang H,Ren Y,Guo F,Wang P,Qiao J,Jia B,Chen C
Thu Jan 01 00:00:00 EST 2015
Lentivirus-mediated Bos taurus bta-miR-29b overexpression interferes with bovine viral diarrhoea virus replication and viral infection-related autophagy by directly targeting ATG14 and ATG9A in Madin-Darby bovine kidney cells.
The Journal of general virology
Fu Q,Shi H,Ni W,Shi M,Meng L,Zhang H,Ren Y,Guo F,Wang P,Qiao J,Jia B,Chen C
Thu Jan 01 00:00:00 EST 2015
Bovine viral diarrhea virus infection induces autophagy in MDBK cells.
Journal of microbiology (Seoul, Korea)
Fu Q,Shi H,Ren Y,Guo F,Ni W,Qiao J,Wang P,Zhang H,Chen C
Tue Jul 01 00:00:00 EDT 2014
Bovine viral diarrhea virus infection induces autophagy in MDBK cells.
Journal of microbiology (Seoul, Korea)
Fu Q,Shi H,Ren Y,Guo F,Ni W,Qiao J,Wang P,Zhang H,Chen C
Tue Jul 01 00:00:00 EDT 2014