Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: The DJR1 monoclonal antibody reacts with human DR4, also known as TRAIL-R1. DR4 binds to TRAIL, activates NF-kappaB, and induces TRAIL-mediated apoptosis. DR4 is expressed by a variety of human tumor cells and at very low level on human leukocytes.
Expression has also been found inside the cell on some cell lines (Zhang et al). This has been confirmed with intracellular staining of Jurkat cell line. Please contact tech@ebioscience.com for more information.
Applications Reported: The DJR1 antibody has been reported for use in flow cytometric analysis.
Applications Tested: The DJR1 antibody has been tested by flow cytometric analysis of human DR4-transfected cells. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Filtration: 0.2 µm post-manufacturing filtered.
TRAIL-R1 (CD261, DR4) is a type I transmembrane protein, also called TRAIL receptor 1. The ligand for this DR4 death receptor has been identified and termed TRAIL, which is a member of the TNF family. DR4, as many other receptors (Fas, TNFR1, etc.), mediates apoptosis and NF kappaB activation in many cells and tissues. Apoptosis, a programmed cell death, is a operating process in normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by coupled of certain cytokines (TNF family - TNF, Fas ligand) and their death domain containing receptors (TNFR1, Fas receptor).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD261; cytotoxic TRAIL receptor; Death receptor 4; MGC9365; sCD261; soluble CD261; soluble TRAIL; sTRAIL; TNF-related apoptosis-inducing ligand receptor 1; TRAIL R1; TRAIL receptor 1; TRAIL-R1; Tumor necrosis factor receptor superfamily member 10A; tumor necrosis factor receptor superfamily member 10a variant 2; tumor necrosis factor receptor superfamily, member 10a
Gene Aliases: APO2; CD261; DR4; TNFRSF10A; TRAILR-1; TRAILR1
UniProt ID: (Human) O00220
Entrez Gene ID: (Human) 8797
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support