Product References
Type H vessel/platelet-derived growth factor receptor β+ perivascular cell disintegration is involved in vascular injury and bone loss in radiation-induced bone damage.
Cell proliferation
Li J,Chen X,Ren L,Chen X,Wu T,Wang Y,Ren X,Cheng B,Xia J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2023
β3 adrenergic agonism: A novel pathway which improves right ventricular-pulmonary arterial hemodynamics in pulmonary arterial hypertension.
Physiological reports
Karimi Galougahi K,Zhang Y,Kienzle V,Liu CC,Quek LE,Patel S,Lau E,Cordina RL,Figtree GA,Celermajer DS
14-0311-82 was used in Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation to examine the efficacy of β3 AR agonism as a novel pathway in experimental PAH. In hypoxia (5 weeks) and Sugen hypoxia (hypoxia for 5 weeks + SU5416 injection) models of PAH, and examined the effects of the selective β3 AR agonist CL316243.
Sun Jan 01 00:00:00 EST 2023
Targeting myocardial equilibrative nucleoside transporter ENT1 provides cardioprotection by enhancing myeloid Adora2b signaling.
JCI insight
Ruan W,Li J,Choi S,Ma X,Liang Y,Nair R,Yuan X,Mills TW,Eltzschig HK
14-0311-82 was used in Flow cytometry/Cell sorting to reveal a previously unrecognized role for myocyte-specific ENT1 in cardioprotection by enhancing myeloid-dependent Adora2b signaling during reperfusion.
Thu Jun 08 00:00:00 EDT 2023
The Wnt/TCF7L1 transcriptional repressor axis drives primitive endoderm formation by antagonizing naive and formative pluripotency.
Nature communications
Athanasouli P,Balli M,De Jaime-Soguero A,Boel A,Papanikolaou S,van der Veer BK,Janiszewski A,Vanhessche T,Francis A,El Laithy Y,Nigro AL,Aulicino F,Koh KP,Pasque V,Cosma MP,Verfaillie C,Zwijsen A,Heindryckx B,Nikolaou C,Lluis F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Mar 03 00:00:00 EST 2023
Isolation and in vitro characterization of murine young-adult long bone skeletal progenitors.
Frontiers in endocrinology
Loopmans S,Stockmans I,Carmeliet G,Stegen S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Aug 19 00:00:00 EDT 2022
Synovial Fibroblast Sialylation Regulates Cell Migration and Activation of Inflammatory Pathways in Arthritogenesis.
Frontiers in immunology
Wang Y,Pan P,Khan A,Çil Ç,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Apr 08 00:00:00 EDT 2022
Macrophage IL-1β promotes arteriogenesis by autocrine STAT3- and NF-κB-mediated transcription of pro-angiogenic VEGF-A.
Cell reports
Mantsounga CS,Lee C,Neverson J,Sharma S,Healy A,Berus JM,Parry C,Ceneri NM,López-Giráldez F,Chun HJ,Lu Q,Sellke F,Choudhary G,Morrison AR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Tue Feb 01 00:00:00 EST 2022
Lamin A/C-dependent chromatin architecture safeguards naïve pluripotency to prevent aberrant cardiovascular cell fate and function.
Nature communications
Wang Y,Elsherbiny A,Kessler L,Cordero J,Shi H,Serke H,Lityagina O,Trogisch FA,Mohammadi MM,El-Battrawy I,Backs J,Wieland T,Heineke J,Dobreva G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Nov 04 00:00:00 EDT 2022
Spatial Transcriptomics-Based Identification of Molecular Markers for Nanomedicine Distribution in Tumor Tissue.
Small methods
Park J,Choi J,Lee JE,Choi H,Im HJ
14-0311-82 was used in Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation to explore the factor candidates that are associated with nanomedicine tumor uptake, a molecular marker identification platform is developed by integrating microscopic fluorescence images of a nanomedicine distribution with spatial transcriptomics information.
Tue Nov 01 00:00:00 EDT 2022
Proteoglycan 4 (PRG4) treatment enhances wound closure and tissue regeneration.
NPJ Regenerative medicine
Krawetz RJ,Abubacker S,Leonard C,Masson AO,Shah S,Narendran N,Tailor P,Regmi SC,Labit E,Ninkovic N,Corpuz JM,Ito K,Underhill TM,Salo PT,Schmidt TA,Biernaskie JA
14-0311-82 was used in Immunohistochemistry-immunofluorescence to propose that rhPRG4 may represent a potential therapy to mitigate scar and improve wound healing.
Fri Jun 24 00:00:00 EDT 2022
Fibroblast activation protein activated antifibrotic peptide delivery attenuates fibrosis in mouse models of liver fibrosis.
Nature communications
Lee J,Byun J,Shim G,Oh YK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Mon Mar 21 00:00:00 EDT 2022
Age-related changes in the local milieu of inflamed tissues cause aberrant neutrophil trafficking and subsequent remote organ damage.
Immunity
Barkaway A,Rolas L,Joulia R,Bodkin J,Lenn T,Owen-Woods C,Reglero-Real N,Stein M,Vázquez-Martínez L,Girbl T,Poston RN,Golding M,Saleeb RS,Thiriot A,von Andrian UH,Duchene J,Voisin MB,Bishop CL,Voehringer D,Roers A,Rot A,Lämmermann T,Nourshargh S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Tue Jul 13 00:00:00 EDT 2021
Age-related changes in the local milieu of inflamed tissues cause aberrant neutrophil trafficking and subsequent remote organ damage.
Immunity
Barkaway A,Rolas L,Joulia R,Bodkin J,Lenn T,Owen-Woods C,Reglero-Real N,Stein M,Vázquez-Martínez L,Girbl T,Poston RN,Golding M,Saleeb RS,Thiriot A,von Andrian UH,Duchene J,Voisin MB,Bishop CL,Voehringer D,Roers A,Rot A,Lämmermann T,Nourshargh S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Tue Jul 13 00:00:00 EDT 2021
Fli1+ cells transcriptional analysis reveals an Lmo2-Prdm16 axis in angiogenesis.
Proceedings of the National Academy of Sciences of the United States of America
Matrone G,Xia B,Chen K,Denvir MA,Baker AH,Cooke JP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Tue Aug 03 00:00:00 EDT 2021
Photoreceptor protection by mesenchymal stem cell transplantation identifies exosomal MiR-21 as a therapeutic for retinal degeneration.
Cell death and differentiation
Deng CL,Hu CB,Ling ST,Zhao N,Bao LH,Zhou F,Xiong YC,Chen T,Sui BD,Yu XR,Hu CH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
Anti-adipogenic signals at the onset of obesity-related inflammation in white adipose tissue.
Cellular and molecular life sciences : CMLS
Caputo T,Tran VDT,Bararpour N,Winkler C,Aguileta G,Trang KB,Giordano Attianese GMP,Wilson A,Thomas A,Pagni M,Guex N,Desvergne B,Gilardi F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
SORLA is required for insulin-induced expansion of the adipocyte precursor pool in visceral fat.
The Journal of cell biology
Schmidt V,Horváth C,Dong H,Blüher M,Qvist P,Wolfrum C,Willnow TE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Dec 06 00:00:00 EST 2021
Galectin-3 Enhances Vascular Endothelial Growth Factor-A Receptor 2 Activity in the Presence of Vascular Endothelial Growth Factor.
Frontiers in cell and developmental biology
Cano I,Hu Z,AbuSamra DB,Saint-Geniez M,Ng YSE,Argüeso P,D'Amore PA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Western Blot
Sat Jul 16 00:00:00 EDT 2022
Tendon and motor phenotypes in the mouse model of recessive osteogenesis imperfecta.
eLife
Grol MW,Haelterman NA,Lim J,Munivez EM,Archer M,Hudson DM,Tufa SF,Keene DR,Lei K,Park D,Kuzawa CD,Ambrose CG,Eyre DR,Lee BH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed May 26 00:00:00 EDT 2021
The antitumoral effects of chemerin are independent from leukocyte recruitment and mediated by inhibition of neoangiogenesis.
Oncotarget
Dubois-Vedrenne I,Al Delbany D,De Henau O,Robert V,Vernimmen M,Langa F,Lefort A,Libert F,Wittamer V,Parmentier M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Tue Sep 14 00:00:00 EDT 2021
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed Sep 08 00:00:00 EDT 2021
The IRE1/XBP1 signaling axis promotes skeletal muscle regeneration through a cell non-autonomous mechanism.
eLife
Roy A,Tomaz da Silva M,Bhat R,Bohnert KR,Iwawaki T,Kumar A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Nov 23 00:00:00 EST 2021
Interleukin-11-expressing fibroblasts have a unique gene signature correlated with poor prognosis of colorectal cancer.
Nature communications
Nishina T,Deguchi Y,Ohshima D,Takeda W,Ohtsuka M,Shichino S,Ueha S,Yamazaki S,Kawauchi M,Nakamura E,Nishiyama C,Kojima Y,Adachi-Akahane S,Hasegawa M,Nakayama M,Oshima M,Yagita H,Shibuya K,Mikami T,Inohara N,Matsushima K,Tada N,Nakano H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Blood angiopoietin-2 predicts liver angiogenesis and fibrosis in hepatitis C patients.
BMC gastroenterology
Osawa Y,Yoshio S,Aoki Y,Korenaga M,Imamura M,Oide T,Okawara M,Kawai H,Tsutsui Y,Yoshida Y,Yoshikawa S,Mori T,Yamazoe T,Kanto T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry (Frozen)
Mon Feb 08 00:00:00 EST 2021
Therapeutic Potential of Mesenchymal Stem Cells in a Pre-Clinical Model of Diabetic Kidney Disease and Obesity.
International journal of molecular sciences
Sávio-Silva C,Soinski-Sousa PE,Simplício-Filho A,Bastos RMC,Beyerstedt S,Rangel ÉB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Feb 04 00:00:00 EST 2021
Protocols for isolation and characterization of mouse placental hemogenic endothelial cells.
STAR protocols
Liang G,Huang B,Wang F,Liu F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Dec 17 00:00:00 EST 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Tue May 11 00:00:00 EDT 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Tue May 11 00:00:00 EDT 2021
Loss of α2-6 sialylation promotes the transformation of synovial fibroblasts into a pro-inflammatory phenotype in arthritis.
Nature communications
Wang Y,Khan A,Antonopoulos A,Bouché L,Buckley CD,Filer A,Raza K,Li KP,Tolusso B,Gremese E,Kurowska-Stolarska M,Alivernini S,Dell A,Haslam SM,Pineda MA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Apr 20 00:00:00 EDT 2021
Computed tomography imaging of macrophage phagocytic activity in abdominal aortic aneurysm.
Theranostics
Toczek J,Boodagh P,Sanzida N,Ghim M,Salarian M,Gona K,Kukreja G,Rajendran S,Wei L,Han J,Zhang J,Jung JJ,Graham M,Liu X,Sadeghi MM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Fri Jul 30 00:00:00 EDT 2021
Mice with induced pulmonary morbidities display severe lung inflammation and mortality following exposure to SARS-CoV-2.
JCI insight
Falach R,Bar-On L,Lazar S,Kadar T,Mazor O,Aftalion M,Gur D,Evgy Y,Shifman O,Aminov T,Israeli O,Cohen-Gihon I,Zaide G,Gutman H,Vagima Y,Makdasi E,Stein D,Rosenfeld R,Alcalay R,Zahavy E,Levy H,Glinert I,Ben-Shmuel A,Israely T,Melamed S,Politi B,Achdout H,Yitzhaki S,Kronman C,Sabo T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Jun 22 00:00:00 EDT 2021
SPARC regulation of PMN clearance protects from pristane-induced lupus and rheumatoid arthritis.
iScience
Sangaletti S,Botti L,Gulino A,Lecis D,Bassani B,Portararo P,Milani M,Cancila V,De Cecco L,Dugo M,Tripodo C,Colombo MP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Deficiency of Dietary Fiber Modulates Gut Microbiota Composition, Neutrophil Recruitment and Worsens Experimental Colitis.
Frontiers in immunology
Shen S,Prame Kumar K,Wen SW,Shim R,Wanrooy BJ,Stanley D,Moore RJ,Van TTH,Robert R,Hickey MJ,Wong CHY
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Mon Jul 05 00:00:00 EDT 2021
Protocols for endothelial cell isolation from mouse tissues: brain, choroid, lung, and muscle.
STAR protocols
Conchinha NV,Sokol L,Teuwen LA,Veys K,Dumas SJ,Meta E,García-Caballero M,Geldhof V,Chen R,Treps L,Borri M,de Zeeuw P,Falkenberg KD,Dubois C,Parys M,de Rooij LPMH,Rohlenova K,Goveia J,Schoonjans L,Dewerchin M,Eelen G,Li X,Kalucka J,Carmeliet P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Sep 17 00:00:00 EDT 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Meningeal lymphatic vessels regulate brain tumor drainage and immunity.
Cell research
Hu X,Deng Q,Ma L,Li Q,Chen Y,Liao Y,Zhou F,Zhang C,Shao L,Feng J,He T,Ning W,Kong Y,Huo Y,He A,Liu B,Zhang J,Adams R,He Y,Tang F,Bian X,Luo J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Platelets docking to VWF prevent leaks during leukocyte extravasation by stimulating Tie-2.
Blood
Braun LJ,Stegmeyer RI,Schäfer K,Volkery S,Currie SM,Kempe B,Nottebaum AF,Vestweber D
14-0311-82 was used in Immunocytochemistry to identify the receptor system preventing plasma leakage during leukocyte extravasation.
Thu Jul 30 00:00:00 EDT 2020
Methylation in pericytes after acute injury promotes chronic kidney disease.
The Journal of clinical investigation
Chou YH,Pan SY,Shao YH,Shih HM,Wei SY,Lai CF,Chiang WC,Schrimpf C,Yang KC,Lai LC,Chen YM,Chu TS,Lin SL
14-0311 was used in Immunohistochemistry-immunofluorescence to show chronic kidney disease can be prevented by inhibiting hypermethylation of pericytes after acute kidney injury.
Tue Sep 01 00:00:00 EDT 2020
Therapy-Induced Senescence Drives Bone Loss.
Cancer research
Yao Z,Murali B,Ren Q,Luo X,Faget DV,Cole T,Ricci B,Thotala D,Monahan J,van Deursen JM,Baker D,Faccio R,Schwarz JK,Stewart SA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
HSPA12B Secreted by Tumor-Associated Endothelial Cells Might Induce M2 Polarization of Macrophages via Activating PI3K/Akt/mTOR Signaling.
OncoTargets and therapy
Zhou J,Zhang A,Fan L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Sun Apr 17 00:00:00 EDT 2022
Defining the role of pulmonary endothelial cell heterogeneity in the response to acute lung injury.
eLife
Niethamer TK,Stabler CT,Leach JP,Zepp JA,Morley MP,Babu A,Zhou S,Morrisey EE
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Feb 24 00:00:00 EST 2020
Enhancer remodeling promotes tumor-initiating activity in NRF2-activated non-small cell lung cancers.
Nature communications
Okazaki K,Anzawa H,Liu Z,Ota N,Kitamura H,Onodera Y,Alam MM,Matsumaru D,Suzuki T,Katsuoka F,Tadaka S,Motoike I,Watanabe M,Hayasaka K,Sakurada A,Okada Y,Yamamoto M,Suzuki T,Kinoshita K,Sekine H,Motohashi H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Nov 20 00:00:00 EST 2020
Snail promotes the generation of vascular endothelium by breast cancer cells.
Cell death & disease
Chang Z,Zhang Y,Liu J,Zheng Y,Li H,Kong Y,Li P,Peng H,Shi Y,Cao B,Ran F,Chen Y,Song Y,Ye Q,Ding L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Jun 15 00:00:00 EDT 2020
The armed oncolytic adenovirus ZD55-IL-24 eradicates melanoma by turning the tumor cells from the self-state into the nonself-state besides direct killing.
Cell death & disease
Hu HJ,Liang X,Li HL,Du CM,Hao JL,Wang HY,Gu JF,Ni AM,Sun LY,Xiao J,Hu JQ,Yuan H,Dai YS,Jin XT,Zhang KJ,Liu XY
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Mon Nov 30 00:00:00 EST 2020
Exosomes derived from GDNF-modified human adipose mesenchymal stem cells ameliorate peritubular capillary loss in tubulointerstitial fibrosis by activating the SIRT1/eNOS signaling pathway.
Theranostics
Chen L,Wang Y,Li S,Zuo B,Zhang X,Wang F,Sun D
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Mon May 24 00:00:00 EDT 2021
Exosomes derived from GDNF-modified human adipose mesenchymal stem cells ameliorate peritubular capillary loss in tubulointerstitial fibrosis by activating the SIRT1/eNOS signaling pathway.
Theranostics
Chen L,Wang Y,Li S,Zuo B,Zhang X,Wang F,Sun D
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Mon May 24 00:00:00 EDT 2021
Sierra: discovery of differential transcript usage from polyA-captured single-cell RNA-seq data.
Genome biology
Patrick R,Humphreys DT,Janbandhu V,Oshlack A,Ho JWK,Harvey RP,Lo KK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed Jul 08 00:00:00 EDT 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Dec 17 00:00:00 EST 2020
Myh11+ microvascular mural cells and derived mesenchymal stem cells promote retinal fibrosis.
Scientific reports
Ray HC,Corliss BA,Bruce AC,Kesting S,Dey P,Mansour J,Seaman SA,Smolko CM,Mathews C,Dey BK,Owens GK,Peirce SM,Yates PA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Fri Sep 25 00:00:00 EDT 2020
Myh11+ microvascular mural cells and derived mesenchymal stem cells promote retinal fibrosis.
Scientific reports
Ray HC,Corliss BA,Bruce AC,Kesting S,Dey P,Mansour J,Seaman SA,Smolko CM,Mathews C,Dey BK,Owens GK,Peirce SM,Yates PA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Fri Sep 25 00:00:00 EDT 2020
Diaphanous-related formin mDia2 regulates beta2 integrins to control hematopoietic stem and progenitor cell engraftment.
Nature communications
Mei Y,Han X,Liu Y,Yang J,Sumagin R,Ji P
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Tue Jun 23 00:00:00 EDT 2020
M2-like, dermal macrophages are maintained via IL-4/CCL24-mediated cooperative interaction with eosinophils in cutaneous leishmaniasis.
Science immunology
Lee SH,Chaves MM,Kamenyeva O,Gazzinelli-Guimaraes PH,Kang B,Pessenda G,Passelli K,Tacchini-Cottier F,Kabat J,Jacobsen EA,Nutman TB,Sacks DL
14-0311-82 was used in In vivo experiments to show that, in infected mice lacking IL-4/13 from eosinophils, dermal tissue-resident macrophages shifted to a proinflammatory state, their numbers declined, and disease was attenuated.
Fri Apr 10 00:00:00 EDT 2020
Disseminated Melanoma Cells Transdifferentiate into Endothelial Cells in Intravascular Niches at Metastatic Sites.
Cell reports
Li X,Karras P,Torres R,Rambow F,van den Oord J,Marine JC,Kos L
14-0311-82 was used in Flow Cytometry to observe tumour cell plasticity in melanoma cells.
Tue Jun 16 00:00:00 EDT 2020
Apelin inhibition prevents resistance and metastasis associated with anti-angiogenic therapy.
EMBO molecular medicine
Uribesalgo I,Hoffmann D,Zhang Y,Kavirayani A,Lazovic J,Berta J,Novatchkova M,Pai TP,Wimmer RA,László V,Schramek D,Karim R,Tortola L,Deswal S,Haas L,Zuber J,Szűcs M,Kuba K,Dome B,Cao Y,Haubner BJ,Penninger JM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Thu Aug 01 00:00:00 EDT 2019
Pericyte transplantation improves skeletal muscle recovery following hindlimb immobilization.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Munroe M,Dvoretskiy S,Lopez A,Leong J,Dyle MC,Kong H,Adams CM,Boppart MD
14-0311 was used in Immunocytochemistry-Immunofluorescence to provide the first evidence that pericytes effectively rehabilitate skeletal muscle mass following disuse atrophy.
Sat Jun 01 00:00:00 EDT 2019
Vascular endothelial growth factor pathway promotes osseointegration and CD31hiEMCNhi endothelium expansion in a mouse tibial implant model: an animal study.
The bone & joint journal
Ji G,Xu R,Niu Y,Li N,Ivashkiv L,Bostrom MPG,Greenblatt MB,Yang X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
Cell-specific ablation of Hsp47 defines the collagen-producing cells in the injured heart.
JCI insight
Khalil H,Kanisicak O,Vagnozzi RJ,Johansen AK,Maliken BD,Prasad V,Boyer JG,Brody MJ,Schips T,Kilian KK,Correll RN,Kawasaki K,Nagata K,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Aug 08 00:00:00 EDT 2019
Single-cell expression profiling reveals dynamic flux of cardiac stromal, vascular and immune cells in health and injury.
eLife
Farbehi N,Patrick R,Dorison A,Xaymardan M,Janbandhu V,Wystub-Lis K,Ho JW,Nordon RE,Harvey RP
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Mar 26 00:00:00 EDT 2019
PDGFRα+ stromal adipocyte progenitors transition into epithelial cells during lobulo-alveologenesis in the murine mammary gland.
Nature communications
Joshi PA,Waterhouse PD,Kasaian K,Fang H,Gulyaeva O,Sul HS,Boutros PC,Khokha R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Apr 15 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
Dual microglia effects on blood brain barrier permeability induced by systemic inflammation.
Nature communications
Haruwaka K,Ikegami A,Tachibana Y,Ohno N,Konishi H,Hashimoto A,Matsumoto M,Kato D,Ono R,Kiyama H,Moorhouse AJ,Nabekura J,Wake H
14-0311-82 was used in Flow Cytometry to show microglia play a dual role in maintaining BBB integrity with implications for elucidating how systemic immune-activation impacts neural functions.
Fri Dec 20 00:00:00 EST 2019
Dual microglia effects on blood brain barrier permeability induced by systemic inflammation.
Nature communications
Haruwaka K,Ikegami A,Tachibana Y,Ohno N,Konishi H,Hashimoto A,Matsumoto M,Kato D,Ono R,Kiyama H,Moorhouse AJ,Nabekura J,Wake H
14-0311-82 was used in Flow Cytometry to show microglia play a dual role in maintaining BBB integrity with implications for elucidating how systemic immune-activation impacts neural functions.
Fri Dec 20 00:00:00 EST 2019
Metastatic cancers promote cachexia through ZIP14 upregulation in skeletal muscle.
Nature medicine
Wang G,Biswas AK,Ma W,Kandpal M,Coker C,Grandgenett PM,Hollingsworth MA,Jain R,Tanji K,Lόpez-Pintado S,Borczuk A,Hebert D,Jenkitkasemwong S,Hojyo S,Davuluri RV,Knutson MD,Fukada T,Acharyya S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Targeting skeletal endothelium to ameliorate bone loss.
Nature medicine
Xu R,Yallowitz A,Qin A,Wu Z,Shin DY,Kim JM,Debnath S,Ji G,Bostrom MP,Yang X,Zhang C,Dong H,Kermani P,Lalani S,Li N,Liu Y,Poulos MG,Wach A,Zhang Y,Inoue K,Di Lorenzo A,Zhao B,Butler JM,Shim JH,Glimcher LH,Greenblatt MB
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Ubiquitin ligase RNF8 suppresses Notch signaling to regulate mammary development and tumorigenesis.
The Journal of clinical investigation
Li L,Guturi KKN,Gautreau B,Patel PS,Saad A,Morii M,Mateo F,Palomero L,Barbour H,Gomez A,Ng D,Kotlyar M,Pastrello C,Jackson HW,Khokha R,Jurisica I,Affar EB,Raught B,Sanchez O,Alaoui-Jamali M,Pujana MA,Hakem A,Hakem R
14-0311 was used in Flow cytometry/Cell sorting to study the correlation between low expression of full-length RNF8 and poor prognosis for breast cancer patients.
Mon Oct 01 00:00:00 EDT 2018
Targeted delivery of immune therapeutics to lymph nodes prolongs cardiac allograft survival.
The Journal of clinical investigation
Bahmani B,Uehara M,Jiang L,Ordikhani F,Banouni N,Ichimura T,Solhjou Z,Furtmüller GJ,Brandacher G,Alvarez D,von Andrian UH,Uchimura K,Xu Q,Vohra I,Yilmam OA,Haik Y,Azzi J,Kasinath V,Bromberg JS,McGrath MM,Abdi R
14-0311 was used in Immunohistochemistry to synthesize a nanoparticle (NP) as a carrier of anti-CD3, and optimized the conjugation strategy to coat the NP surface with MECA79 mAb (MECA79-anti-CD3-NP) to enhance LN accumulation.
Thu Nov 01 00:00:00 EDT 2018
IL-7-dependent maintenance of ILC3s is required for normal entry of lymphocytes into lymph nodes.
The Journal of experimental medicine
Yang J,Cornelissen F,Papazian N,Reijmers RM,Llorian M,Cupedo T,Coles M,Seddon B
14-0311 was used in Flow cytometry/Cell sorting to investigate whether continued IL-7 expression was required for the maintenance of fully developed, adult lymph nodes.
Mon Apr 02 00:00:00 EDT 2018
IL-7-dependent maintenance of ILC3s is required for normal entry of lymphocytes into lymph nodes.
The Journal of experimental medicine
Yang J,Cornelissen F,Papazian N,Reijmers RM,Llorian M,Cupedo T,Coles M,Seddon B
14-0311 was used in Flow cytometry/Cell sorting to investigate whether continued IL-7 expression was required for the maintenance of fully developed, adult lymph nodes.
Mon Apr 02 00:00:00 EDT 2018
Indian Hedgehog Suppresses a Stromal Cell-Driven Intestinal Immune Response.
Cellular and molecular gastroenterology and hepatology
Westendorp BF,Büller NVJA,Karpus ON,van Dop WA,Koster J,Versteeg R,Koelink PJ,Snel CY,Meisner S,Roelofs JJTH,Uhmann A,Ver Loren van Themaat E,Heijmans J,Hahn H,Muncan V,Wildenberg ME,van den Brink GR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Mar 31 00:00:00 EDT 2022
Mammary molecular portraits reveal lineage-specific features and progenitor cell vulnerabilities.
The Journal of cell biology
Casey AE,Sinha A,Singhania R,Livingstone J,Waterhouse P,Tharmapalan P,Cruickshank J,Shehata M,Drysdale E,Fang H,Kim H,Isserlin R,Bailey S,Medina T,Deblois G,Shiah YJ,Barsyte-Lovejoy D,Hofer S,Bader G,Lupien M,Arrowsmith C,Knapp S,De Carvalho D,Berman H,Boutros PC,Kislinger T,Khokha R
14-0311 was used in Flow cytometry/Cell sorting to investigate the normal cell composition of the mammary epithelium by profiling primary cell epigenomes, transcriptomes and proteomes.
Mon Aug 06 00:00:00 EDT 2018
Therapeutic Impact of Nanoparticle Therapy Targeting Tumor-Associated Macrophages.
Molecular cancer therapeutics
Penn CA,Yang K,Zong H,Lim JY,Cole A,Yang D,Baker J,Goonewardena SN,Buckanovich RJ
14-0311 was used in Immunohistochemistry to demonstrate here that ovarian tumour-associated macrophages express high levels of the folate receptor-2 and can be selectively targeted using G5-dendrimer nanoparticles using methotrexate as both a ligand and a toxin.
Mon Jan 01 00:00:00 EST 2018
Modelling glioblastoma tumour-host cell interactions using adult brain organotypic slice co-culture.
Disease models & mechanisms
Marques-Torrejon MA,Gangoso E,Pollard SM
14-0311 was used in Immunocytochemistry to explore the use of whole adult brain coronal organotypic slices as an alternative model for glioblastoma tumour-host cell interactions.
Thu Feb 22 00:00:00 EST 2018
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
Collagenase-based Single Cell Isolation of Primary Murine Brain Endothelial Cells Using Flow Cytometry.
Bio-protocol
Czupalla CJ,Yousef H,Wyss-Coray T,Butcher EC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in ChIP assay
Tue Nov 20 00:00:00 EST 2018
Collagenase-based Single Cell Isolation of Primary Murine Brain Endothelial Cells Using Flow Cytometry.
Bio-protocol
Czupalla CJ,Yousef H,Wyss-Coray T,Butcher EC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in ChIP assay
Tue Nov 20 00:00:00 EST 2018
Stem cell factor is selectively secreted by arterial endothelial cells in bone marrow.
Nature communications
Xu C,Gao X,Wei Q,Nakahara F,Zimmerman SE,Mar J,Frenette PS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Jun 22 00:00:00 EDT 2018
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Dec 11 00:00:00 EST 2018
Impairment of Angiogenesis by Fatty Acid Synthase Inhibition Involves mTOR Malonylation.
Cell metabolism
Bruning U,Morales-Rodriguez F,Kalucka J,Goveia J,Taverna F,Queiroz KCS,Dubois C,Cantelmo AR,Chen R,Loroch S,Timmerman E,Caixeta V,Bloch K,Conradi LC,Treps L,Staes A,Gevaert K,Tee A,Dewerchin M,Semenkovich CF,Impens F,Schilling B,Verdin E,Swinnen JV,Meier JL,Kulkarni RA,Sickmann A,Ghesquière B,Schoonjans L,Li X,Mazzone M,Carmeliet P
14-0311-82 was used in Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Misc to report that fatty acid synthase knockdown (FASNKD) in ECs impedes vessel sprouting by reducing proliferation.
Tue Dec 04 00:00:00 EST 2018
Tumour ischaemia by interferon-γ resembles physiological blood vessel regression.
Nature
Kammertoens T,Friese C,Arina A,Idel C,Briesemeister D,Rothe M,Ivanov A,Szymborska A,Patone G,Kunz S,Sommermeyer D,Engels B,Leisegang M,Textor A,Fehling HJ,Fruttiger M,Lohoff M,Herrmann A,Yu H,Weichselbaum R,Uckert W,Hübner N,Gerhardt H,Beule D,Schreiber H,Blankenstein T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu May 04 00:00:00 EDT 2017
Increased Vascular Permeability in the Bone Marrow Microenvironment Contributes to Disease Progression and Drug Response in Acute Myeloid Leukemia.
Cancer cell
Passaro D,Di Tullio A,Abarrategi A,Rouault-Pierre K,Foster K,Ariza-McNaughton L,Montaner B,Chakravarty P,Bhaw L,Diana G,Lassailly F,Gribben J,Bonnet D
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Sep 11 00:00:00 EDT 2017
Gli1+ Mesenchymal Stromal Cells Are a Key Driver of Bone Marrow Fibrosis and an Important Cellular Therapeutic Target.
Cell stem cell
Schneider RK,Mullally A,Dugourd A,Peisker F,Hoogenboezem R,Van Strien PMH,Bindels EM,Heckl D,Büsche G,Fleck D,Müller-Newen G,Wongboonsin J,Ventura Ferreira M,Puelles VG,Saez-Rodriguez J,Ebert BL,Humphreys BD,Kramann R
14-0311 was used in Immunohistochemistry-immunofluorescence to study the cellular origin of bone marrow fibrosis.
Thu Jun 01 00:00:00 EDT 2017
VEGF165b Modulates Endothelial VEGFR1-STAT3 Signaling Pathway and Angiogenesis in Human and Experimental Peripheral Arterial Disease.
Circulation research
Ganta VC,Choi M,Kutateladze A,Annex BH
14031182 was used in flow cytometry to find that that vascular endothelial growth factor 165b prevents activation of vascular endothelial growth factor receptor 1-STAT3 signaling by vascular endothelial growth factor 165a
Fri Jan 20 00:00:00 EST 2017
Preexisting endothelial cells mediate cardiac neovascularization after injury.
The Journal of clinical investigation
He L,Huang X,Kanisicak O,Li Y,Wang Y,Li Y,Pu W,Liu Q,Zhang H,Tian X,Zhao H,Liu X,Zhang S,Nie Y,Hu S,Miao X,Wang QD,Wang F,Chen T,Xu Q,Lui KO,Molkentin JD,Zhou B
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Gli1+ Pericyte Loss Induces Capillary Rarefaction and Proximal Tubular Injury.
Journal of the American Society of Nephrology : JASN
Kramann R,Wongboonsin J,Chang-Panesso M,Machado FG,Humphreys BD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Wed Mar 01 00:00:00 EST 2017
Gli1+ Pericyte Loss Induces Capillary Rarefaction and Proximal Tubular Injury.
Journal of the American Society of Nephrology : JASN
Kramann R,Wongboonsin J,Chang-Panesso M,Machado FG,Humphreys BD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Wed Mar 01 00:00:00 EST 2017
CD34+ mesenchymal cells are a major component of the intestinal stem cells niche at homeostasis and after injury.
Proceedings of the National Academy of Sciences of the United States of America
Stzepourginski I,Nigro G,Jacob JM,Dulauroy S,Sansonetti PJ,Eberl G,Peduto L
14031182 was used in flow cytometry to discuss the relationship between CD34+ Gp38+ mesenchymal cells and Lgr5+ intestinal epithelial stem cells
Tue Jan 24 00:00:00 EST 2017
A population of hematopoietic stem cells derives from GATA4-expressing progenitors located in the placenta and lateral mesoderm of mice.
Haematologica
Cañete A,Carmona R,Ariza L,Sánchez MJ,Rojas A,Muñoz-Chápuli R
14031182 was used in flow cytometry to utilize a G2-Gata4Cre;R26REYFP mouse line to track the developmental fate of the G2-Gata4 cell lineage
Sat Apr 01 00:00:00 EDT 2017
Defective germinal center B-cell response and reduced arthritic pathology in microRNA-29a-deficient mice.
Cellular and molecular life sciences : CMLS
van Nieuwenhuijze A,Dooley J,Humblet-Baron S,Sreenivasan J,Koenders M,Schlenner SM,Linterman M,Liston A
14031182 was used in flow cytometry to evaluate miR-29a in B cells as a potential therapeutic target in arthritis
Thu Jun 01 00:00:00 EDT 2017
Obesity reversibly depletes the basal cell population and enhances mammary epithelial cell estrogen receptor alpha expression and progenitor activity.
Breast cancer research : BCR
Chamberlin T,D'Amato JV,Arendt LM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Wed Nov 29 00:00:00 EST 2017
Obesity reversibly depletes the basal cell population and enhances mammary epithelial cell estrogen receptor alpha expression and progenitor activity.
Breast cancer research : BCR
Chamberlin T,D'Amato JV,Arendt LM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Wed Nov 29 00:00:00 EST 2017
Mesenchymal Stem Cell Therapy Protects Lungs from Radiation-Induced Endothelial Cell Loss by Restoring Superoxide Dismutase 1 Expression.
Antioxidants & redox signaling
Klein D,Steens J,Wiesemann A,Schulz F,Kaschani F,Röck K,Yamaguchi M,Wirsdörfer F,Kaiser M,Fischer JW,Stuschke M,Jendrossek V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Apr 10 00:00:00 EDT 2017
Characterization of transgene expression and pDNA distribution of the suctioned kidney in mice.
Drug delivery
Oyama N,Fuchigami Y,Fumoto S,Sato M,Hagimori M,Shimizu K,Kawakami S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry (Frozen)
Wed Nov 01 00:00:00 EDT 2017
Characterization of transgene expression and pDNA distribution of the suctioned kidney in mice.
Drug delivery
Oyama N,Fuchigami Y,Fumoto S,Sato M,Hagimori M,Shimizu K,Kawakami S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry (Frozen)
Wed Nov 01 00:00:00 EDT 2017
Characterization of transgene expression and pDNA distribution of the suctioned kidney in mice.
Drug delivery
Oyama N,Fuchigami Y,Fumoto S,Sato M,Hagimori M,Shimizu K,Kawakami S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry (Frozen)
Wed Nov 01 00:00:00 EDT 2017
Cornea lymphatics drive the CD8+ T-cell response to herpes simplex virus-1.
Immunology and cell biology
Gurung HR,Carr MM,Carr DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
β3-Adrenergic Regulation of EPC Features Through Manipulation of the Bone Marrow MSC Niche.
Journal of cellular biochemistry
Vafaei R,Nassiri SM,Siavashi V
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
Vector-mediated Tum-5 expression in neovascular endothelial cells for treating hepatocellular carcinoma.
Experimental and therapeutic medicine
Li C,Guan X,Sun B,Ma M,Wang P,Gai X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Sat Apr 01 00:00:00 EDT 2017
Endogenous TNFα orchestrates the trafficking of neutrophils into and within lymphatic vessels during acute inflammation.
Scientific reports
Arokiasamy S,Zakian C,Dilliway J,Wang W,Nourshargh S,Voisin MB
14-0311 was used in Immunofluorescence to demonstrate a new role for TNFα as a key regulator of neutrophil trafficking into and within the lymphatic system.
Mon Mar 13 00:00:00 EDT 2017
PKN1 Directs Polarized RAB21 Vesicle Trafficking via RPH3A and Is Important for Neutrophil Adhesion and Ischemia-Reperfusion Injury.
Cell reports
Yuan Q,Ren C,Xu W,Petri B,Zhang J,Zhang Y,Kubes P,Wu D,Tang W
14-0311 was used in Flow cytometry/Cell sorting to characterize a mechanism for protein polarization in neutrophils and identify a potential protein kinase target for therapeutic intervention in reperfusion-related tissue injury.
Tue Jun 20 00:00:00 EDT 2017
RIPK1/RIPK3 promotes vascular permeability to allow tumor cell extravasation independent of its necroptotic function.
Cell death & disease
Hänggi K,Vasilikos L,Valls AF,Yerbes R,Knop J,Spilgies LM,Rieck K,Misra T,Bertin J,Gough PJ,Schmidt T,de Almodòvar CR,Wong WW
14-0311 was used in Flow cytometry/Cell sorting to suggest an alternative function for RIPK1/RIPK3 in vascular permeability leading to decreased number of metastasis.
Thu Feb 02 00:00:00 EST 2017
Dynamic Patterns of Clonal Evolution in Tumor Vasculature Underlie Alterations in Lymphocyte-Endothelial Recognition to Foster Tumor Immune Escape.
Cancer research
Corey DM,Rinkevich Y,Weissman IL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Mar 15 00:00:00 EDT 2016
Endothelial cell survival during angiogenesis requires the pro-survival protein MCL1.
Cell death and differentiation
Watson EC,Whitehead L,Adams RH,Dewson G,Coultas L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Group 2 innate lymphoid cells utilize the IRF4-IL-9 module to coordinate epithelial cell maintenance of lung homeostasis.
Mucosal immunology
Mohapatra A,Van Dyken SJ,Schneider C,Nussbaum JC,Liang HE,Locksley RM
14-0311 was used in Flow cytometry/Cell sorting to study how ILC2s link alveolar function to regulation of airway flow, revealing a key interaction between resident lymphoid and structural cells that might underlie similar organisational hierarchies in other organs.
Fri Jan 01 00:00:00 EST 2016
A20 Deficiency in Lung Epithelial Cells Protects against Influenza A Virus Infection.
PLoS pathogens
Maelfait J,Roose K,Vereecke L,Mc Guire C,Sze M,Schuijs MJ,Willart M,Ibañez LI,Hammad H,Lambrecht BN,Beyaert R,Saelens X,van Loo G
14-0311 was used in Flow cytometry/Cell sorting to investigate the function of A20 in club cells in antiviral immunity, showing that deficiency in A20 protects against influenza virus infection.
Fri Jan 01 00:00:00 EST 2016
Lack of VEGFR2 signaling causes maldevelopment of the intestinal microvasculature and facilitates necrotizing enterocolitis in neonatal mice.
American journal of physiology. Gastrointestinal and liver physiology
Yan X,Managlia E,Liu SX,Tan XD,Wang X,Marek C,De Plaen IG
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Sun May 01 00:00:00 EDT 2016
A Prolactin Family Paralog Regulates Placental Adaptations to a Physiological Stressor.
Biology of reproduction
Bu P,Alam SM,Dhakal P,Vivian JL,Soares MJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Sun May 01 00:00:00 EDT 2016
Laminin regulates PDGFRβ(+) cell stemness and muscle development.
Nature communications
Yao Y,Norris EH,Mason CE,Strickland S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Tue May 03 00:00:00 EDT 2016
Activated c-Kit receptor in the heart promotes cardiac repair and regeneration after injury.
Cell death & disease
Di Siena S,Gimmelli R,Nori SL,Barbagallo F,Campolo F,Dolci S,Rossi P,Venneri MA,Giannetta E,Gianfrilli D,Feigenbaum L,Lenzi A,Naro F,Cianflone E,Mancuso T,Torella D,Isidori AM,Pellegrini M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Thu Jul 28 00:00:00 EDT 2016
Endothelial to mesenchymal transition is common in atherosclerotic lesions and is associated with plaque instability.
Nature communications
Evrard SM,Lecce L,Michelis KC,Nomura-Kitabayashi A,Pandey G,Purushothaman KR,d'Escamard V,Li JR,Hadri L,Fujitani K,Moreno PR,Benard L,Rimmele P,Cohain A,Mecham B,Randolph GJ,Nabel EG,Hajjar R,Fuster V,Boehm M,Kovacic JC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Jun 24 00:00:00 EDT 2016
Endothelial to mesenchymal transition is common in atherosclerotic lesions and is associated with plaque instability.
Nature communications
Evrard SM,Lecce L,Michelis KC,Nomura-Kitabayashi A,Pandey G,Purushothaman KR,d'Escamard V,Li JR,Hadri L,Fujitani K,Moreno PR,Benard L,Rimmele P,Cohain A,Mecham B,Randolph GJ,Nabel EG,Hajjar R,Fuster V,Boehm M,Kovacic JC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Jun 24 00:00:00 EDT 2016
Genetic lineage tracing defines myofibroblast origin and function in the injured heart.
Nature communications
Kanisicak O,Khalil H,Ivey MJ,Karch J,Maliken BD,Correll RN,Brody MJ,J Lin SC,Aronow BJ,Tallquist MD,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Fri Jul 22 00:00:00 EDT 2016
Genetic lineage tracing defines myofibroblast origin and function in the injured heart.
Nature communications
Kanisicak O,Khalil H,Ivey MJ,Karch J,Maliken BD,Correll RN,Brody MJ,J Lin SC,Aronow BJ,Tallquist MD,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunocytochemistry
Fri Jul 22 00:00:00 EDT 2016
ADAM17 substrate release in proximal tubule drives kidney fibrosis.
JCI insight
Kefaloyianni E,Muthu ML,Kaeppler J,Sun X,Sabbisetti V,Chalaris A,Rose-John S,Wong E,Sagi I,Waikar SS,Rennke H,Humphreys BD,Bonventre JV,Herrlich A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Thu Aug 18 00:00:00 EDT 2016
Leukemia Inhibitory Factor Enhances Endogenous Cardiomyocyte Regeneration after Myocardial Infarction.
PloS one
Kanda M,Nagai T,Takahashi T,Liu ML,Kondou N,Naito AT,Akazawa H,Sashida G,Iwama A,Komuro I,Kobayashi Y
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Thu Jul 06 00:00:00 EDT 2017
Leukemia Inhibitory Factor Enhances Endogenous Cardiomyocyte Regeneration after Myocardial Infarction.
PloS one
Kanda M,Nagai T,Takahashi T,Liu ML,Kondou N,Naito AT,Akazawa H,Sashida G,Iwama A,Komuro I,Kobayashi Y
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Thu Jul 06 00:00:00 EDT 2017
A cell-autonomous tumour suppressor role of RAF1 in hepatocarcinogenesis.
Nature communications
Jeric I,Maurer G,Cavallo AL,Raguz J,Desideri E,Tarkowski B,Parrini M,Fischer I,Zatloukal K,Baccarini M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed Dec 21 00:00:00 EST 2016
Structural and functional features of central nervous system lymphatic vessels.
Nature
Louveau A,Smirnov I,Keyes TJ,Eccles JD,Rouhani SJ,Peske JD,Derecki NC,Castle D,Mandell JW,Lee KS,Harris TH,Kipnis J
14-0311 was used in Immunohistochemistry-wholemount to investigate the mechanisms governing the entrance and exit of immune cells from the CNS.
Thu Jul 16 00:00:00 EDT 2015
Self-renewing diploid Axin2(+) cells fuel homeostatic renewal of the liver.
Nature
Wang B,Zhao L,Fish M,Logan CY,Nusse R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Aug 13 00:00:00 EDT 2015
Inhibition of the TNF Family Cytokine RANKL Prevents Autoimmune Inflammation in the Central Nervous System.
Immunity
Guerrini MM,Okamoto K,Komatsu N,Sawa S,Danks L,Penninger JM,Nakashima T,Takayanagi H
14-0311-82 was used in Flow Cytometry to show that receptor activator of nuclear factor-κB (NF-κB) ligand (RANKL) on T cells regulates C-C type chemokine ligand 20 (CCL20) production by astrocytes and T cell localization in the CNS.
Tue Dec 15 00:00:00 EST 2015
Host microbiota constantly control maturation and function of microglia in the CNS.
Nature neuroscience
Erny D,Hrabě de Angelis AL,Jaitin D,Wieghofer P,Staszewski O,David E,Keren-Shaul H,Mahlakoiv T,Jakobshagen K,Buch T,Schwierzeck V,Utermöhlen O,Chun E,Garrett WS,McCoy KD,Diefenbach A,Staeheli P,Stecher B,Amit I,Prinz M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Hepatocellular carcinoma originates from hepatocytes and not from the progenitor/biliary compartment.
The Journal of clinical investigation
Mu X,Español-Suñer R,Mederacke I,Affò S,Manco R,Sempoux C,Lemaigre FP,Adili A,Yuan D,Weber A,Unger K,Heikenwälder M,Leclercq IA,Schwabe RF
14-0311 was used in Immunohistochemistry to study the cell of origin for hepatocellular carcinoma in mice.
Thu Oct 01 00:00:00 EDT 2015
Transcriptional Pathways in cPGI2-Induced Adipocyte Progenitor Activation for Browning.
Frontiers in endocrinology
Bayindir I,Babaeikelishomi R,Kocanova S,Sousa IS,Lerch S,Hardt O,Wild S,Bosio A,Bystricky K,Herzig S,Vegiopoulos A
14-0311 was used in Magnetic cell separation to develop models for the mechanistic investigation of progenitor-mediated beige/brite adipocyte differentiation.
Tue Sep 08 00:00:00 EDT 2015
Vascular endothelial growth factor c/vascular endothelial growth factor receptor 3 signaling regulates chemokine gradients and lymphocyte migration from tissues to lymphatics.
Transplantation
Iwami D,Brinkman CC,Bromberg JS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Conditional immortalization of primary adipocyte precursor cells.
Adipocyte
Church C,Brown M,Rodeheffer MS
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Aug 10 00:00:00 EDT 2015
Platelet aggregation but not activation and degranulation during the acute post-ischemic reperfusion phase in livers with no underlying disease.
Journal of clinical and translational research
van Golen RF,Stevens KM,Colarusso P,Jaeschke H,Heger M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Thu Mar 31 00:00:00 EDT 2022
Effector lymphocyte-induced lymph node-like vasculature enables naive T-cell entry into tumours and enhanced anti-tumour immunity.
Nature communications
Peske JD,Thompson ED,Gemta L,Baylis RA,Fu YX,Engelhard VH
14031182 was used in flow cytometry and immunohistochemistry to find that lymph node-like vasculature in melanoma and lung carcinoma murine models is both a consequence of and key contributor to anti-tumor immunity
Wed May 13 00:00:00 EDT 2015
Targeted Gene Correction in Osteopetrotic-Induced Pluripotent Stem Cells for the Generation of Functional Osteoclasts.
Stem cell reports
Neri T,Muggeo S,Paulis M,Caldana ME,Crisafulli L,Strina D,Focarelli ML,Faggioli F,Recordati C,Scaramuzza S,Scanziani E,Mantero S,Buracchi C,Sobacchi C,Lombardo A,Naldini L,Vezzoni P,Villa A,Ficara F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Oct 13 00:00:00 EDT 2015
Roles of lymphatic endothelial cells expressing peripheral tissue antigens in CD4 T-cell tolerance induction.
Nature communications
Rouhani SJ,Eccles JD,Riccardi P,Peske JD,Tewalt EF,Cohen JN,Liblau R,Mäkinen T,Engelhard VH
14-0311 was used in Flow cytometry/Cell sorting to demonstrate that when β-galactosidase (β-gal) is expressed in LECs, β-gal-specific CD8 T cells undergo deletion via the PD-1/PD-L1 and LAG-3/MHC-II pathways.
Fri Apr 10 00:00:00 EDT 2015
Ginger-derived nanoparticles protect against alcohol-induced liver damage.
Journal of extracellular vesicles
Zhuang X,Deng ZB,Mu J,Zhang L,Yan J,Miller D,Feng W,McClain CJ,Zhang HG
14-0311 was used in Immunohistochemistry to discover that orally given ginger nanoparticles provided protection against alcohol-induced liver damage in mice.
Fri Nov 27 00:00:00 EST 2015
Rejuvenation of the muscle stem cell population restores strength to injured aged muscles.
Nature medicine
Cosgrove BD,Gilbert PM,Porpiglia E,Mourkioti F,Lee SP,Corbel SY,Llewellyn ME,Delp SL,Blau HM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Sat Mar 01 00:00:00 EST 2014
c-kit+ cells minimally contribute cardiomyocytes to the heart.
Nature
van Berlo JH,Kanisicak O,Maillet M,Vagnozzi RJ,Karch J,Lin SC,Middleton RC,Marbán E,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu May 15 00:00:00 EDT 2014
c-kit+ cells minimally contribute cardiomyocytes to the heart.
Nature
van Berlo JH,Kanisicak O,Maillet M,Vagnozzi RJ,Karch J,Lin SC,Middleton RC,Marbán E,Molkentin JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu May 15 00:00:00 EDT 2014
Mesenchymal-endothelial transition contributes to cardiac neovascularization.
Nature
Ubil E,Duan J,Pillai IC,Rosa-Garrido M,Wu Y,Bargiacchi F,Lu Y,Stanbouly S,Huang J,Rojas M,Vondriska TM,Stefani E,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Thu Oct 30 00:00:00 EDT 2014
Mesenchymal-endothelial transition contributes to cardiac neovascularization.
Nature
Ubil E,Duan J,Pillai IC,Rosa-Garrido M,Wu Y,Bargiacchi F,Lu Y,Stanbouly S,Huang J,Rojas M,Vondriska TM,Stefani E,Deb A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Thu Oct 30 00:00:00 EDT 2014
Sex steroid blockade enhances thymopoiesis by modulating Notch signaling.
The Journal of experimental medicine
Velardi E,Tsai JJ,Holland AM,Wertheimer T,Yu VW,Zakrzewski JL,Tuckett AZ,Singer NV,West ML,Smith OM,Young LF,Kreines FM,Levy ER,Boyd RL,Scadden DT,Dudakov JA,van den Brink MR
14-0311 was used in Flow cytometry/Cell sorting to reveal a novel mechanism underlying improved thymic regeneration upon sex steroid ablation.
Mon Nov 17 00:00:00 EST 2014
TGF-β-activated kinase 1 is crucial in podocyte differentiation and glomerular capillary formation.
Journal of the American Society of Nephrology : JASN
Kim SI,Lee SY,Wang Z,Ding Y,Haque N,Zhang J,Zhou J,Choi ME
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Mon Sep 01 00:00:00 EDT 2014
TGF-β-activated kinase 1 is crucial in podocyte differentiation and glomerular capillary formation.
Journal of the American Society of Nephrology : JASN
Kim SI,Lee SY,Wang Z,Ding Y,Haque N,Zhang J,Zhou J,Choi ME
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Mon Sep 01 00:00:00 EDT 2014
Fluorescence microangiography for quantitative assessment of peritubular capillary changes after AKI in mice.
Journal of the American Society of Nephrology : JASN
Kramann R,Tanaka M,Humphreys BD
14-0311 was used in Immunohistochemistry-immunofluorescence to adapt and validate a fluorescence microangiography approach for use with mice to visualise, analyse, and quantitate peritubular capillary dynamics after acute kidney injury.
Mon Sep 01 00:00:00 EDT 2014
p53Ψ is a transcriptionally inactive p53 isoform able to reprogram cells toward a metastatic-like state.
Proceedings of the National Academy of Sciences of the United States of America
Senturk S,Yao Z,Camiolo M,Stiles B,Rathod T,Walsh AM,Nemajerova A,Lazzara MJ,Altorki NK,Krainer A,Moll UM,Lowe SW,Cartegni L,Sordella R
14-0311 was used in Flow cytometry/Cell sorting to describe a unique mode of p53 regulation involving alternative splicing of the TP53 gene.
Tue Aug 12 00:00:00 EDT 2014
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Mar 13 00:00:00 EDT 2014
Pharmacological mobilization of endogenous stem cells significantly promotes skin regeneration after full-thickness excision: the synergistic activity of AMD3100 and tacrolimus.
The Journal of investigative dermatology
Lin Q,Wesson RN,Maeda H,Wang Y,Cui Z,Liu JO,Cameron AM,Gao B,Montgomery RA,Williams GM,Sun Z
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Mon Sep 01 00:00:00 EDT 2014
Pharmacological mobilization of endogenous stem cells significantly promotes skin regeneration after full-thickness excision: the synergistic activity of AMD3100 and tacrolimus.
The Journal of investigative dermatology
Lin Q,Wesson RN,Maeda H,Wang Y,Cui Z,Liu JO,Cameron AM,Gao B,Montgomery RA,Williams GM,Sun Z
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Mon Sep 01 00:00:00 EDT 2014
Characterization of Cre recombinase activity for in vivo targeting of adipocyte precursor cells.
Stem cell reports
Krueger KC,Costa MJ,Du H,Feldman BJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Tue Dec 09 00:00:00 EST 2014
The Wilms' tumour suppressor Wt1 is a major regulator of tumour angiogenesis and progression.
Nature communications
Wagner KD,Cherfils-Vicini J,Hosen N,Hohenstein P,Gilson E,Hastie ND,Michiels JF,Wagner N
14-0311 was used in Flow cytometry/Cell sorting to show that inducible knockout of Wt1 is sufficient to cause regression of tumour vascularisation and enhanced anti-tumour immune responses in mice.
Tue Dec 16 00:00:00 EST 2014
Perivascular mast cells dynamically probe cutaneous blood vessels to capture immunoglobulin E.
Immunity
Cheng LE,Hartmann K,Roers A,Krummel MF,Locksley RM
14-0311 was used in Flow cytometry/Cell sorting to investigate the mechanism by which perivascular mast cells acquire IgE from the blood.
Thu Jan 24 00:00:00 EST 2013
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Fri Feb 01 00:00:00 EST 2013
miR-126 and miR-126* repress recruitment of mesenchymal stem cells and inflammatory monocytes to inhibit breast cancer metastasis.
Nature cell biology
Zhang Y,Yang P,Sun T,Li D,Xu X,Rui Y,Li C,Chong M,Ibrahim T,Mercatali L,Amadori D,Lu X,Xie D,Li QJ,Wang XF
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
The expression of Sox17 identifies and regulates haemogenic endothelium.
Nature cell biology
Clarke RL,Yzaguirre AD,Yashiro-Ohtani Y,Bondue A,Blanpain C,Pear WS,Speck NA,Keller G
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013
Blockade of NOX2 and STIM1 signaling limits lipopolysaccharide-induced vascular inflammation.
The Journal of clinical investigation
Gandhirajan RK,Meng S,Chandramoorthy HC,Mallilankaraman K,Mancarella S,Gao H,Razmpour R,Yang XF,Houser SR,Chen J,Koch WJ,Wang H,Soboloff J,Gill DL,Madesh M
14-0311 was used in Immunohistochemistry to elucidate the molecular mechanisms behind the development of LPS-induced acute lung injury.
Fri Feb 01 00:00:00 EST 2013
Wnt4/β-catenin signaling in medullary kidney myofibroblasts.
Journal of the American Society of Nephrology : JASN
DiRocco DP,Kobayashi A,Taketo MM,McMahon AP,Humphreys BD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Sun Sep 01 00:00:00 EDT 2013
Etv2 rescues Flk1 mutant embryoid bodies.
Genesis (New York, N.Y. : 2000)
Rasmussen TL,Martin CM,Walter CA,Shi X,Perlingeiro R,Koyano-Nakagawa N,Garry DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2013
Angiopoietins modulate endothelial adaptation, glomerular and podocyte hypertrophy after uninephrectomy.
PloS one
Chiang WC,Lai CF,Su CT,Peng WH,Wu CF,Chang FC,Chen YT,Lin SL,Chen YM,Wu KD,Lu KS,Tsai TJ
14-0311 was used in Immunohistochemistry to demonstrate that angiopoietin 1 or 2 may play an important role in the development of glomerular hypertrophy after uninephrectomy.
Wed Oct 08 00:00:00 EDT 2014
Endothelial cells provide an instructive niche for the differentiation and functional polarization of M2-like macrophages.
Blood
He H,Xu J,Warren CM,Duan D,Li X,Wu L,Iruela-Arispe ML
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Oct 11 00:00:00 EDT 2012
Endothelial cells provide an instructive niche for the differentiation and functional polarization of M2-like macrophages.
Blood
He H,Xu J,Warren CM,Duan D,Li X,Wu L,Iruela-Arispe ML
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Oct 11 00:00:00 EDT 2012
Lymphatic endothelial cells induce tolerance via PD-L1 and lack of costimulation leading to high-level PD-1 expression on CD8 T cells.
Blood
Tewalt EF,Cohen JN,Rouhani SJ,Guidi CJ,Qiao H,Fahl SP,Conaway MR,Bender TP,Tung KS,Vella AT,Adler AJ,Chen L,Engelhard VH
14-0311 was used in Flow cytometry/Cell sorting to demonstrate that LECs mediate deletion only via programmed cell death-1 (PD-1) ligand 1.
Thu Dec 06 00:00:00 EST 2012
Lymphatic endothelial cells induce tolerance via PD-L1 and lack of costimulation leading to high-level PD-1 expression on CD8 T cells.
Blood
Tewalt EF,Cohen JN,Rouhani SJ,Guidi CJ,Qiao H,Fahl SP,Conaway MR,Bender TP,Tung KS,Vella AT,Adler AJ,Chen L,Engelhard VH
14-0311 was used in Flow cytometry/Cell sorting to demonstrate that LECs mediate deletion only via programmed cell death-1 (PD-1) ligand 1.
Thu Dec 06 00:00:00 EST 2012
Lymphotoxin-beta receptor blockade induces inflammation and fibrosis in tolerized cardiac allografts.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Nakayama Y,Bromberg JS
14-0311 was used in Flow cytometry/Cell sorting to study the effect of LTβRIg treatment in a murine cardiac allograft model.
Sat Sep 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Wed Aug 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunofluorescence
Wed Aug 01 00:00:00 EDT 2012
The transcriptional programme controlled by Runx1 during early embryonic blood development.
Developmental biology
Tanaka Y,Joshi A,Wilson NK,Kinston S,Nishikawa S,Göttgens B
14-0311 was used in Flow cytometry/Cell sorting to analyse the transcriptional programme controlled by a key haematopoietic regulator during the process of early blood cell specification.
Fri Jun 15 00:00:00 EDT 2012
Primed T cell responses to chemokines are regulated by the immunoglobulin-like molecule CD31.
PloS one
Kishore M,Ma L,Cornish G,Nourshargh S,Marelli-Berg FM
14-0311 was used in Functional assays to study the effects of CD31-mediated signals on T cell chemokinesis.
Wed Dec 12 00:00:00 EST 2012
Characterization and fate of telomerase-expressing epithelia during kidney repair.
Journal of the American Society of Nephrology : JASN
Song J,Czerniak S,Wang T,Ying W,Carlone DL,Breault DT,Humphreys BD
14-0311 was used in Immunofluorescence to elucidate the role of telomere length for kidney repair.
Thu Dec 01 00:00:00 EST 2011
Intramuscular VEGF activates an SDF1-dependent progenitor cell cascade and an SDF1-independent muscle paracrine cascade for cardiac repair.
American journal of physiology. Heart and circulatory physiology
Zisa D,Shabbir A,Mastri M,Taylor T,Aleksic I,McDaniel M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011
Heparan sulfate regulates VEGF165- and VEGF121-mediated vascular hyperpermeability.
The Journal of biological chemistry
Xu D,Fuster MM,Lawrence R,Esko JD
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Fri Jan 07 00:00:00 EST 2011
A chemotactic gradient sequestered on endothelial heparan sulfate induces directional intraluminal crawling of neutrophils.
Blood
Massena S,Christoffersson G,Hjertström E,Zcharia E,Vlodavsky I,Ausmees N,Rolny C,Li JP,Phillipson M
14-0311 was used in Immunofluorescence to determine whether a chemokine gradient was formed intravascularly and influenced intraluminal neutrophil transmigration.
Thu Sep 16 00:00:00 EDT 2010
Minimally invasive assessment of tumor angiogenesis by fine needle aspiration and flow cytometry.
Angiogenesis
Sottnik JL,Guth AM,Mitchell LA,Dow SW
14-0311 was used in Immunohistochemistry on frozen tissues to develop an optimised method for studying tumour angiogenesis.
Wed Sep 01 00:00:00 EDT 2010
Auf1/Hnrnpd-deficient mice develop pruritic inflammatory skin disease.
The Journal of investigative dermatology
Sadri N,Schneider RJ
Sun Mar 01 00:00:00 EST 2009
Heart failure therapy mediated by the trophic activities of bone marrow mesenchymal stem cells: a noninvasive therapeutic regimen.
American journal of physiology. Heart and circulatory physiology
Shabbir A,Zisa D,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2009
Intramuscular VEGF repairs the failing heart: role of host-derived growth factors and mobilization of progenitor cells.
American journal of physiology. Regulatory, integrative and comparative physiology
Zisa D,Shabbir A,Mastri M,Suzuki G,Lee T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2009
Resident vascular progenitor cells: an emerging role for non-terminally differentiated vessel-resident cells in vascular biology.
Stem cell research
Kovacic JC,Boehm M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Thu Jan 01 00:00:00 EST 2009
Resident vascular progenitor cells: an emerging role for non-terminally differentiated vessel-resident cells in vascular biology.
Stem cell research
Kovacic JC,Boehm M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 14-0311-82) in Immunohistochemistry
Thu Jan 01 00:00:00 EST 2009