Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
CHMP5 belongs to the chromatin-modifying protein/charged multivesicular body protein family. These proteins are components of ESCRT-III, a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies. Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apoptosis-related protein PNAS-2; Charged multivesicular body protein 5; chromatin modifying protein 5; Chromatin-modifying protein 5; HGNC:26942; hVps60; likely ortholog of H. sapiens SNF7 domain containing 2; SNF7 domain containing 2; SNF7 domain-containing protein 2; Vacuolar protein sorting-associated protein 60; Vps60
Gene Aliases: 2210412K09Rik; AW545668; C9orf83; CGI-34; CHMP5; HSPC177; PNAS-114; PNAS-2; RGD1305968; SNF7DC2; Vps60
UniProt ID: (Human) Q9NZZ3, (Mouse) Q9D7S9, (Rat) Q4QQV8
Entrez Gene ID: (Human) 51510, (Mouse) 76959, (Rat) 297995
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support