Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can recognize both CYFIP1 and CYFIP2.
Immunogen sequence: CYNGSTNRF VRTAIPFTQE PQRDKPANVQ PYYLYGSKPL NIAYSHIYSS YRNFVGPPHF KTICRLLGYQ GIAVVMEELL KIVKSLLQGT ILQYVKTLIE VMPKICRLPR HEYGSPGILE FFHHQLKDII EYAELKTDVF QSLREVGNAI LFCLLIEQAL SQEEVCDLLH AAPFQNILPR VYIKEGERLE VRMKRLEAKY APLHLVPLIE RLGTPQQIAI AREGDLLTKE RLCCGLSMFE VILTRIRSYL QDPIWRGPPP TNGVMHVDEC VEFHRLWSAM QFVYCIPVGT NEFTAEQCFG DGLNWAGCSI IVLLGQQRRF DLFDFCYHLL KVQRQDGKDE IIKNVPLKKM ADRIRKYQIL NNEVFAILNK YMKSVETDSS TVEHVRCFQP PIHQSLATTC (879-1278 aa encoded by BC011762)
CYFIP2 (Cytoplasmic FMR1-interacting protein 2, p53-inducible protein 121) is a 1, 278 amino acid protein encoded by the human gene CYFIP2. CYFIP2 belongs to the CYFIP family and is involved in T-cell adhesion and p53-dependent induction of apoptosis. It interacts with FMR1, FXR1 and FXR2 and is a component of the WAVE1 complex composed of Abi-2, CYFIP2, C3orf10/HSPC300, NAP125 and WASF1/WAVE1. Upon binding to activated Rac 1, CYFIP2 causes the complex to dissociate, releasing activated WASF1. The CYFIP2 promoter contains a p53-responsive element that confers p53 binding as well as transcriptional activation of a heterologous reporter. Induced expression of CYFIP2 is sufficient for caspase activation and cellular apoptosis, reminiscent of p53 activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYFIP1; CYFIP2; Cytoplasmic FMR1-interacting protein 1; Cytoplasmic FMR1-interacting protein 2; cytoplasmic FMRP interacting protein 1; p140sra-1; p53-inducible protein 121; selective hybridizing clone; Specifically Rac1-associated protein 1; Sra-1
Gene Aliases: 1500004I01Rik; 6430511D02Rik; AA930218; AU022376; CYFIP1; CYFIP2; E030028J09Rik; KIAA0068; KIAA1168; l(7)1Rl; l71Rl; l7Rl1; mKIAA0068; mKIAA1168; P140SRA-1; P140sra1; PIR121; pl-1; SHYC; SRA-1; SRA1
UniProt ID: (Human) Q7L576, (Human) Q96F07, (Mouse) Q7TMB8, (Mouse) Q5SQX6
Entrez Gene ID: (Human) 23191, (Human) 26999, (Mouse) 20430, (Mouse) 76884
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support