Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MLETLRERLL SVQQDFTSGL KTLSDKSREA KVKSKPRTVP FLPKYSAGLE LLSRYEDTWA ALHRRAKDCA SAGELVDSEV VMLSAHWEKK KTSLVELQEQ LQQLPALIAD LESMTANLTH LEASFEEVEN NLLHLEDLCG QCELERCKHM QSQQLENYKK NKRKELETFK AELDAEHAQK VLEMEHTQQM KLKERQKFFE EAFQQDMEQY LSTGYLQIAE RREPIGSMSS MEVNVDMLEQ MDLMDISDQE ALDVFLNSGG EENTVLSPAL GPESSTCQNE ITLQVPNPSE LRAKPPSSSS TCTDSATRDI SEGGESPVVQ SDEEEVQVDT ALATSHTDRE ATPDGGEDSD S; Positive Samples: HeLa, Jurkat, HepG2, MCF7, DU145; Cellular Location: Cell junction, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle, Cytoplasmic vesicle membrane, Endoplasmic reticulum, Endosome membrane, Melanosome membrane, Nucleus, Nucleus, Peripheral membrane protein, postsynaptic cell membrane, postsynaptic density, secretory vesicle, synapse, synaptic vesicle membrane
Dtnbp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 8; Biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; DKFZp564K192; DTBP1; Dysbindin; Dysbindin-1; Dystrobrevin-binding protein 1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; Hermansky-Pudlak syndrome 7 protein homolog; HPS7 protein; HPS7 protein homolog; Hps7-like protein; MGC20210; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1; sandy
Gene Aliases: 5430437B18Rik; AW048963; BLOC1S8; DBND; DTNBP1; dysbindin; HPS7; My031; SDY
UniProt ID: (Human) Q96EV8, (Mouse) Q9CY43
Entrez Gene ID: (Human) 84062, (Mouse) 94245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support