Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognizes all of isoforms of dysbindin.
Immunogen sequence: MLETLRERL LSVQQDFTSG LKTLSDKSRE AKVKSKPRTV PFLPKYSAGL ELLSRYEDTW AALHRRAKDC ASAGELVDSE VVMLSAHWEK KKTSLVELQE QLQQLPALIA DLESMTANLT HLEASFEEVE NNLLHLEDLC GQCELERCKH MQSQQLENYK KNKRKELETF KAELDAEHAQ KVLEMEHTQQ MKLKERQKFF EEAFQQDMEQ YLSTGYLQIA ERREPIGSMS SMEVNVDMLE QMDLMDISDQ EALDVFLNSG GEENTVLSPA LGPESSTCQN EITLQVPNPS ELRAKPPSSS STCTDSATRD ISEGGESPVV QSDEEEVQVD TALATSHTDR EATPDGGEDS DS (1-351 aa encoded by BC011912)
Dtnbp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 8; Biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; distrobrevin binding protein 1; DKFZp564K192; DTBP1; Dysbindin; Dysbindin-1; Dystrobrevin-binding protein 1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; Hermansky-Pudlak syndrome 7 protein homolog; HPS7 protein; HPS7 protein homolog; Hps7-like protein; MGC20210; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1; sandy
Gene Aliases: 5430437B18Rik; AW048963; BLOC1S8; DBND; DTNBP1; dysbindin; HPS7; My031; SDY
UniProt ID: (Human) Q96EV8, (Mouse) Q9CY43, (Rat) Q5M834
Entrez Gene ID: (Human) 84062, (Mouse) 94245, (Rat) 641528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support