Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNGKRPAEPG PARVGKKGKK EVMAEFSDAV TEETLKKQVA EAWSRRTPFS HEVIVMDMDP FLHCVIPNFI QSQDFLEGLQ KELMNLDFHE KYNDLYKFQQ SDDLKKRREP HISTLRKILF EDFRSWLSDI SKIDLESTID MSCAKYEFTD ALLCHDDELE GRRIAFILYL VPPWDRSMGG TLDLYSIDEH FQPKQIVKSL IPSWNKLVFF EVSPVSFHQV SEVLSEEKSR LSISGWFHGP SLTRPPNYFE PPIPRSPHIP QDHEILYDWI NPTYLDMDYQ VQIQEEFEES SEILLKEFLK PEKFTKVCEA LEHGHVEWSS RGPPNKRFYE KAEESKLPEI LKECMKLFRS EALFLLLSNF TGLKLHFLAP SEEDEMNDKK EAETTDITEE GTSHSPPEPE NNQMAISNNS QQSNEQTDPE PEENETKKES SVPMCQGELR HWKTGHYTLI HDHSKAEFAL DLILYCGCEG WEPEYGGFTS YIAKGEDEEL LTVNPESNSL ALVYRDRETL KFVKHINHRS LEQKKTFPNR TGFWDFSFIY YE
OGFOD1, a member of the 2-OG-Fe(II) dioxygenase family, plays an important role in ischemic survival and stress. It is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. An OGFOD1 iron binding residue is required for ATPAF1 gene expression. As a novel stress granule component, OGFOD1 plays important proapoptotic roles in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-oxoglutarate and iron-dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; Prolyl 3-hydroxylase OGFOD1; Termination and polyadenylation 1 homolog; TPA1, termination and polyadenylation 1, homolog; uS12 prolyl 3-hydroxylase
Gene Aliases: KIAA1612; OGFOD1; TPA1
UniProt ID: (Human) Q8N543
Entrez Gene ID: (Human) 55239
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support