Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA500G10.4; FLJ11864; glutaredoxin 4; Glutaredoxin-3; PICOT; PKC-interacting cousin of thioredoxin; PKC-theta-interacting protein; PKCq-interacting protein; testicular tissue protein Li 75; Thioredoxin-like protein 2
Gene Aliases: GLRX3; GLRX4; GRX3; GRX4; HUSSY-22; PICOT; TXNL2; TXNL3
UniProt ID: (Human) O76003
Entrez Gene ID: (Human) 10539
Molecular Function:
oxidoreductase
reductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support