Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); HMG-CoA synthase; hydroxymethylglutaryl-CoA synthase 1; Hydroxymethylglutaryl-CoA synthase, cytoplasmic
Gene Aliases: B130032C06Rik; HMGCS; HMGCS1
UniProt ID: (Human) Q01581, (Rat) P17425, (Mouse) Q8JZK9
Entrez Gene ID: (Human) 3157, (Rat) 29637, (Mouse) 208715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support