Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Control: GFP-human IL28-transfected 293T cells
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IL-28B encodes a cytokine distantly related to type I interferons and the IL-10 family. IL-28B, interleukin 28A (IL28A), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytokine Zcyto22; IFN-lambda 3; IFN-lambda-3; IFN-lambda-4; IL-28B; IL-28C; ILN; Interferon lambda-3; interferon, lambda 3; interferon, lambda 4; Interleukin; interleukin 28C; Interleukin-28B; Interleukin-28C
Gene Aliases: IFNL3; IL-28B; IL28B; IL28C; ZCYTO22
UniProt ID: (Human) Q8IZI9
Entrez Gene ID: (Human) 282617
Molecular Function:
interleukin superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support