Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GAEFWSQYR INVTEVNPLG ASTRLLDVSL QSILRPDPPQ GLRVESVPGY PRRLRASWTY PASWPCQPHF LLKFRLQYRP AQHPAWSTVE PAGLEEVITD AVAGLPHAVR VSARDFLDAG TWSTWSPEAW GTPSTGTIPK EIPAWGQLHT QPEVEPQVDS PAPPRPSLQP HPRLLDHRD (184-361 aa encoded by BC003110)
Interleukin 11 is a stromal cell-derived cytokine that belongs to a family of pleiotropic and redundant cytokines that use the gp130 transducing subunit in their high affinity receptors. This gene encodes the IL-11 receptor, which is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jun 2012].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enhancer trap locus homolog 2; Etl-2; IL-11 receptor subunit alpha; IL-11 receptor subunit alpha-1; IL-11R subunit alpha; IL-11R subunit alpha-1; IL-11R-alpha; IL-11R-alpha-1; IL-11RA; Il-11ra-alpha; IL-11RA1; interleukin 11 receptor, alpha; interleukin 11 receptor, alpha chain 2; interleukin-11 receptor alpha chain; Interleukin-11 receptor subunit alpha; Interleukin-11 receptor subunit alpha-1; locus 2; Novel cytokine receptor 1; NR-1; sIL11RA
Gene Aliases: AI314697; CRSDA; Etl2; GP130; Il-11ra; IL11RA; Il11ra1; Il11ra2; NR1
UniProt ID: (Human) Q14626, (Rat) Q99MF4, (Mouse) Q64385
Entrez Gene ID: (Human) 3590, (Rat) 245983, (Mouse) 16157
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support