Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total INPP5E.
INPP5E (inositol polyphosphate 5-phosphatase), also known as phosphatidylinositol polyphosphate 5-phosphatase type IV, is a 644 amino acid peripheral membrane protein associated with Golgi stacks. Belonging to the inositol-1, 4, 5-trisphosphate 5-phosphatase type IV family, INPP5E converts phosphatidylinositol-3, 4, 5-triphosphate (PtdIns 3, 4, 5-P3) to PtdIns-P2. While inactive towards water soluble inositol phosphates, the activity of INPP5E is specific for lipid substrates. INPP5E becomes phosphorylated upon DNA damage and is expressed in brain, heart, pancreas, testis and spleen. Defects in INPP5E are the cause of Joubert syndrome type 1 (JBTS1), a disorder presenting with cerebellar ataxia, oculomotor apraxia, hypotonia, neonatal breathing abnormalities and psychomotor delay. In addition, mutations in the INPP5E gene may lead to mental retardation-truncal obesity-retinal dystrophy-micropenis (MORMS), an autosomal recessive disorder characterized by moderate mental retardation, truncal obesity, congenital non-progressive retinal dystrophy, and micropenis in males.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-phosphatase that induces arborization; 72 kDa inositol polyphosphate 5-phosphatase; inositol polyphosphate 5-phosphatase; Inositol polyphosphate-5-phosphatase E; inositol polyphosphate-5-phosphatase, 72 kDa; Pharbin; Phosphatidylinositol 4,5-bisphosphate 5-phosphatase; Phosphatidylinositol polyphosphate 5-phosphatase type IV; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; phosphatidylinositol-4,5-bisphosphate 5-phosphatase
Gene Aliases: 1200002L24Rik; 72kDa; CORS1; CPD4; INPP5E; JBTS1; mKIAA0123; MORMS; PPI5PIV
UniProt ID: (Human) Q9NRR6, (Rat) Q9WVR1, (Mouse) Q9JII1
Entrez Gene ID: (Human) 56623, (Rat) 114089, (Mouse) 64436
Molecular Function: hydrolase phosphatase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support