Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPARSGAQFC RRMGQKKQRP ARAGQPHSSS DAAQAPAEQP HSSSDAAQAP CPRERCLGPP TTPGPYRSIY FSSPKGHLTR LGLEFFDQPA VPLARAFLGQ VLVRRLPNGT ELRGRIVETE AYLGPEDEAA HSRGGRQTPR NRGMFMKPGT LYVYIIYGMY FCMNISSQGD GACVLLRALE PLEGLETMRQ LRSTLRKGTA SRVLKDRELC SGPSKLCQAL AINKSFDQRD LAQDEAVWLE RGPLEPSEPA VVAAARVGVG HAGEWARKPL RFYVRGSPWV SVVDRVAEQD TQA
MPG functions in the hydrolysis of the deoxyribose N-glycosidic bond to excise 3-methyladenine, and 7-methylguanine from the damaged DNA polymer formed by alkylation lesions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; 3-alkyladenine DNA glycosylase; 3-methyladenine DNA glycosidase; ADPG; CRA36.1 (3-methyl-adenine DNA glycosylase); DNA-3-methyladenine glycosylase; N-methylpurine-DNA glycocylase; N-methylpurine-DNA glycosylase; N-methylpurine-DNA glycosylase, MPG; proliferation-inducing protein 11; proliferation-inducing protein 16
Gene Aliases: AAG; ADPG; ANPG; APNG; CRA36.1; MDG; MID1; MPG; PIG11; PIG16
UniProt ID: (Human) Q13770
Entrez Gene ID: (Human) 4350, (Rat) 24561
Molecular Function: DNA metabolism protein DNA glycosylase nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support