Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MQPGSSRCEE ETPSLLWGLD PVFLAFAKLY IRDILDMKES RQVPGVFLYN GHPIKQVDVL GTVIGVRERD AFYSYGVDDS TGVINCICWK KLNTESVSAA PSAARELSLT SQLKKLQETI EQKTKIEIGD TIRVRGSIRT YREEREIHAT AYYKVDDPVW NIQIARMLEL PTIYRKVYDQ PFHSSALEKE EALSNPGALD LPSLTSLLSE KAKEFLMENR VQSFYQQELE MVESLLSLAN QPVIHSACSD QVNFKKDTTS KAIHSIFKNA IQLLQEKGLV FQKDDGFDNL YYVTREDKDL HRKIHRIIQQ DCQKPNHMEK GCHFLHILAC ARLSIRPGLS EAVLQQVLEL LEDQSDIVST MEHYYTAF
OBFC1 is a component of the CST complex, a complex that binds to single-stranded DNA and is required to protect telomeres from DNA degradation. The CST complex binds single-stranded DNA with high affinity in a sequence-independent manner, while isolated subunits bind DNA with low affinity by themselves. In addition to telomere protection, the CST complex has probably a more general role in DNA metabolism at non-telomeric sites.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha accessory factor 44; CST complex subunit STN1; oligonucleotide/oligosaccharide-binding fold containing 1; Oligonucleotide/oligosaccharide-binding fold-containing protein 1; replication protein A 32 kDa subunit; Suppressor of cdc thirteen homolog
Gene Aliases: AAF-44; AAF44; bA541N10.2; OBFC1; RPA-32; STN1
UniProt ID: (Human) Q9H668
Entrez Gene ID: (Human) 79991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support