Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Rap1 is a small GTPase that, through effectors, regulates Rho GTPase signaling. These effectors- Rasip1, Radil, and the protein encoded by this gene- translocate to the cell membrane, where they form a multiprotein complex. This complex is necessary for Rap1-induced inhibition of Rho signaling. Defects in this gene may be a cause of nonsyndromic cleft lip with or without cleft palate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PTPL1-associated RhoGAP 1; PTPL1-associated RhoGAP 1 (PARG1); PTPL1-associated RhoGAP protein 1; Rho GTPase-activating protein 29; Rho-type GTPase-activating protein 29; RP11-255E17.1
Gene Aliases: 6720461J18Rik; A830014I19; ARHGAP29; AU040217; B130017I01Rik; C76601; PARG1
UniProt ID: (Human) Q52LW3, (Mouse) Q8CGF1
Entrez Gene ID: (Human) 9411, (Mouse) 214137
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support