Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-dimethyl-L-lysine(36) demethylase PHF8; [histone H3]-dimethyl-L-lysine(9) demethylase PHF8; Histone lysine demethylase PHF8; jumonji C domain-containing histone demethylase 1F; PHD finger protein 8
Gene Aliases: 9830141C09Rik; JHDM1F; KIAA1111; mKIAA1111; MRXSSD; PHF8; RGD1561065; ZNF422
UniProt ID: (Human) Q9UPP1, (Mouse) Q80TJ7
Entrez Gene ID: (Human) 23133, (Mouse) 320595, (Rat) 317425
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support