Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GSCSLGTLR DHQSSPYAAV PYKLFTDHGG LNEKRKQRRI RTTFTSAQLK ELERVFAETH YPDIYTREEL ALKIDLTEAR VQVWFQNRRA KFRKQERAAA AAAAAAKNGS SGKKSDSSRD DESKEAKSTD PDSTGGPGPN PNPTPSCGAN GGGGGGPSPA GAPGAAGPGG PGGEPGKGGA AAAAAAAAAA AAAAAAAAAG GLAAAGGPGQ GWAPGPGPIT SIPDSLGGPF ASVLSSLQRP NGAKAALVKS SMF (63-314 aa encoded by BC017199)
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dilated pupils 1; GENA 269; NBPhox; neuroblastoma paired-type homeobox protein; Neuroblastoma Phox; paired mesoderm homeobox 2b; Paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B homeodomain protein
Gene Aliases: Dilp1; NBLST2; NBPhox; PHOX2B; PMX2B
UniProt ID: (Human) Q99453, (Mouse) O35690
Entrez Gene ID: (Human) 8929, (Mouse) 18935
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support