Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
The PR65 subunit of protein phosphatase 2A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. Upon interaction with GNA12 promotes dephosphorylation of microtubule associated protein TAU/MAPT. Required for proper chromosome segregation and for centromeric localization of SGO1 in mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha isoform of regulatory subunit A, protein phosphatase 2; Medium tumor antigen-associated 61 kDa protein; MGC786; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; PP2Ac; PP2Calpha; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase PP2A; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; testicular secretory protein Li 1
Gene Aliases: 6330556D22Rik; MRD36; PP2A; PP2A-Aalpha; PP2AAALPHA; PPP2R1A; PR65; PR65A
UniProt ID: (Human) P30153, (Mouse) Q76MZ3
Entrez Gene ID: (Human) 5518, (Mouse) 51792, (Rat) 117281
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support