Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEQLLSSVG FCTEVEEDLI DAVTGLSGSG PAYAFTALDA LADGGVKMGL PRRLAVRLGA QALLGAAKML LHSEQHPGQL KDNVSSPGGA TIHALHVLES GGFRSLLINA VEASCIRTRE LQSMADQEQV SPAAIKKTIL DKVKLDSPAG TALSPSGHTK LLPRSLAPAG KD (149-319 aa encoded by BC022244)
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: P5C reductase 1; P5CR 1; proliferation-inducing protein 45; Pyrroline-5-carboxylate reductase 1, mitochondrial
Gene Aliases: ARCL2B; ARCL3B; P5C; P5CR; PIG45; PP222; PRO3; PYCR; PYCR1
UniProt ID: (Human) P32322, (Mouse) Q922W5
Entrez Gene ID: (Human) 5831, (Mouse) 209027, (Rat) 287877
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support