Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17. LLGL2 is a protein similar to lethal giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DLG4; HGL; Hugl-1; human giant larvae homolog; Human homolog to the D-lgl gene protein; lethal giant larvae homolog 1; lethal giant larvae homolog 1, scribble cell polarity complex component; lethal giant larvae homolog 2; lethal giant larvae homolog 2, scribble cell polarity complex component; Lethal giant larvae-like protein 2; Lethal(2) giant larvae protein homolog 1; Lethal(2) giant larvae protein homolog 2; LLGL; LLGL scribble cell polarity complex component 2; Mgl-1; Mlgl
Gene Aliases: 9130006H11Rik; AI325176; DLG4; HGL; HUGL; HUGL-1; Hugl-2; HUGL1; Lgl1; LGL2; LLGL; LLGL1; LLGL2; Llglh; Llglh2; Mgl1; RGD1560307; Rgl-1; Rgl1
UniProt ID: (Human) Q15334, (Human) Q6P1M3, (Mouse) Q80Y17, (Mouse) Q3TJ91
Entrez Gene ID: (Human) 3996, (Human) 3993, (Rat) 54265, (Mouse) 16897, (Rat) 360661, (Mouse) 217325
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support