Bethyl Laboratories
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
H2AX is a variant of the H2A protein family and is a component of the histone octomer in nucleosomes. H2AX is phosphorylated by kinases such as ataxia telangiectasia mutated (ATM) and ATM-Rad3-related (ATR) in the PI3K pathway. Gamma-H2AX is the first step in recruiting and localizing DNA repair proteins. DSBs can be induced by mechanisms such as ionizing radiation or cytotoxic agents and subsequently, gamma-H2AX foci quickly form. These foci represent the DSBs in a 1:1 manner and can be used as a biomarker for damage. The visualization of gamma-H2AX by can allow the assessment of DNA damage, related DNA damage proteins and DNA repair. Gamma-H2AX has applications in the detection of genomic damage caused by cytotoxic chemical agents and environmental and physical damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma H2A X; gamma H2A.X; gamma-H2A.X; gh2ax; H2A histone family, member X; H2a/x; H2AX histone; histone 5 protein 2ax; Histone H2A.X; Histone H2AX
Gene Aliases: AW228881; gammaH2ax; H2A.X; H2A/X; H2AFX; H2AX; Hist5-2ax
UniProt ID: (Human) P16104, (Mouse) P27661
Entrez Gene ID: (Human) 3014, (Mouse) 15270
Molecular Function: histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support