Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive test controls include: MCF7, NCI-H460, HeLa, Mouse spleen, Mouse brain, Mouse thymus, Rat liver. The target is usually found in the following locations: Nucleus.
Immunogen sequence: SIWAQNQVEN WSAFAPDFKE LDENVEYEER ESEFDIEDED KSEPEQTGAD AAEDEEVDVT SVDPIAAFCS SDEELEDSKA LLYLPIAPEV EDPEENPYGP PPDAVQTSLM DEGASSEKKR QSSADGSQPP KKKPKTTNIE LQGVPNDEVH PLLGVKGDGK SKKKQAGRPK GSKGKEKDSP FKPKLYKGDR GLPLEGSAKG KVQAELSQPL TAGGAISELL
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. The encoded protein binds directly to retinoblastoma protein, which regulates cell proliferation. It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RBBP 5; RBBP-5; RBQ 3; retinoblastoma binding protein 5; Retinoblastoma-binding protein 5; Retinoblastoma-binding protein RBQ-3; SWD1, Set1c WD40 repeat protein, homolog
Gene Aliases: 4933411J24Rik; C330016J05; RBBP5; RBQ3; SWD1
UniProt ID: (Human) Q15291, (Mouse) Q8BX09
Entrez Gene ID: (Human) 5929, (Rat) 304794, (Mouse) 213464
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support