FIGURE: 1 / 1
Sequence of this protein is as follows: MTSFQEVPLQ TSNFAHVIFQ NVAKSYLPNA HLECHYTLTP YIHPHPKDWV GIFKVGWSTA RDYYTFLWSP MPEHYVEGST VNCVLAFQGY YLPNDDGEFY QFCYVTHKGE IRGASTPFQF RASSPVEELL TMEDEGNSDM LVVTTKAGLL ELKIEKTMKE KEELLKLIAV LEKETAQLRE QVGRMERELN HEKERCDQLQ AEQKGLTEVT QSLKMENEEF KKRFSDATSK AHQLEEDIVS VTHKAIEKET ELDSLKDKLK KAQHEREQLE CQLKTEKDEK ELYKVHLKNT EIENTKLMSE VQTLKNIDGN KESVITHFKE EIGRLQLCLA EKENLQRTFL LTTSSKEDTC FLKEQLRKAE EQVQATRQEV VFLAKELSDA VNVRDRTMAD LHTARLENEK VKKQLADAVA ELKLNAMKKD QDKTDTLEHE LRREVEDLKL RLQMAADHYK EKFKECQRLQ KQINKLSDQS ANNNNVFTKK TGNQQKVNDA SVNTDPATSA STVDVKPSPS AAEADFDIVT KGQVCEMTKE IADKTEKYNK CKQLLQDEKA KCNKYADELA KMELKWKEQV KIAENVKLEL AEVQDNYKEL KRSLENPAER KMEGQNSQSP QCFKTCSEQN GYVLTLSNAQ PVLQYGNPYA SQETRDGADG AFYPDEIQRP PVRVPSWGLE DNVVCSQPAR NFSRPDGLED SEDSKEDENV PTAPDPPSQH LRGHGTGFCF DSSFDVHKKC PLCELMFPPN YDQSKFEEHV ESHWKVCPMC SEQFPPDYDQ QVFERHVQTH FDQNVLNFD
This gene encodes a HTLV-1 tax1 binding protein. The encoded protein interacts with TNFAIP3, and inhibits TNF-induced apoptosis by mediating the TNFAIP3 anti-apoptotic activity. Degradation of this protein by caspase-3-like family proteins is associated with apoptosis induced by TNF. This protein may also have a role in the inhibition of inflammatory signaling pathways. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1-binding protein 1; TRAF6-binding protein
Gene Aliases: CALCOCO3; PRO0105; T6BP; TAX1BP1; TXBP151
UniProt ID: (Human) Q86VP1
Entrez Gene ID: (Human) 8887
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support