Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: GQSCAGEPDE ELVGAFPIFV RNAILGQKQP KRPRSESSCS KDVVRTGFDG LGGRTKFIQP TDTVMIRPLP VKPKTKVKQR VRVKTVPSLF QAKLDTFLWS
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRAIP; RING finger protein 206; TRAF-interacting protein
Gene Aliases: RNF206; SCKL9; TRAIP; TRIP
UniProt ID: (Human) Q9BWF2
Entrez Gene ID: (Human) 10293
Molecular Function: ubiquitin-protein ligase protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support