Search Thermo Fisher Scientific
All of our high-content screening platforms offer high-speed image acquisition, powerful image processing algorithms and unparalleled real-time analysis. Platforms feature more than 30 validated assays and options including live-cell imaging and in-depth life sciences analysis, including single-cell analysis capabilities. Browse the categories and specifications below, then link to your models of choice for more information.
|
||
CellInsight CX7 LED Pro | CellInsight CX5 | |
Best option for |
||
High-throughput screening and 3D organoid and spheroid research involving thick specimens. |
Multiplex biology, including time-lapse and live-cell imaging. |
Researchers needing optics that deliver optimal assay sensitivity. |
Camera | ||
sCMOS | sCMOS | CCD |
Illumination | ||
7 channels |
7 channels |
5 channels |
Widefield |
||
7-color laser excitation |
7-color LED excitation |
color LED excitation |
Brightfield |
||
4 specific LED + white light for higher contrast imaging (5-color total) |
4 specific LED + white light for higher contrast imaging (5-color total) |
White |
Confocal |
||
Nipkow spinning disk with 40 µm & 70 µm pinholes |
Nipkow spinning disk with 40 µm & 70 µm pinholes |
NA |
Objectives |
||
Olympus X-line objectives, magnifications 2x-60x |
||
Focus |
||
Laser and software |
Laser and software |
Software |
Microplate types |
||
6 to 1,536 wells |
||
Onboard real-time analysis software |
||
HCS Studio 5.0 Cell Analysis Software with Cell-painting Bioapplication |
HCS Studio 4.0 Cell Analysis Software | |
EurekaScan Finder software |
||
yes |
yes |
no |
Z-stack acquisition | ||
yes | yes |
yes |
Amira Software compatibility | ||
yes | yes | yes |
Microscope slides | ||
yes | yes | yes |
Live-cell and time-lapse imaging | ||
Optional on-stage incubator with kinetic scheduling of acquisition and analysis. CO2 and N2 for hypoxia conditions available. | ||
Database | ||
Store Express Image and Database Management Software Include: scalable SQL/Oracle database options available |