Search Thermo Fisher Scientific
With over 25 years of experience, Thermo Fisher Scientific is dedicated to making quality and custom DNA and RNA oligos for your application. We are proud to offer large-scale oligo synthesis services to help ensure our customers obtain desired customized oligos in bulk quantities. Whether for scale-up, manufacturing, or OEM, Thermo Fisher Scientific is here to support your large-scale oligo synthesis needs.
Thermo Fisher Scientific's mission is to enable our customers to make the world healthier, cleaner, and safer in industries such as:
Whether you're looking for milligram, gram, or kilogram amounts, Thermo Fisher Scientific can support you with large-scale oligo synthesis services. We offer a complete range of custom-synthesized oligonucleotides built to your specifications. To customize oligos for stability and function choose from a variety of 5', 3', and internal modifications. Table 1 lists some of the modifications available for large-scale oligo synthesis. Oligos can be ordered desalted or purified via cartridge, HPLC, or PAGE methods to match the oligo design and your downstream application.
Explore the evolving world of large-scale oligo synthesis in our on-demand webinar, where we delve into modern techniques, technologies, and emerging applications from R&D to clinical diagnostics and therapeutics. Watch and gain insights into the challenges associated with scaling up synthesis and learn how Thermo Fisher Scientific can support you with customized support throughout the development journey.
Modification type | Purpose in oligo design | Examples | Applications |
---|---|---|---|
Attachment chemistry or linkers | Linking oligo to another molecule or surface | Amino C6, C12 Biotin C6, TEG Thiol C3, C6 |
|
Phosphorylation | For oligos used as a substrate for DNA ligase | Phosphate 3’ or 5’ |
|
Phosphorothioate bonds | Increase resistance to nuclease degradation | F, O, E, Z: IUPAC codes for phosphorothioated A, C, G, T |
|
Spacers | Physically create space between a moiety and hybridizing region of the oligo | C3 spacer, PC spacer |
|
Modified bases | Aid in cross-linking and nuclease resistance | LNA, L-DNA |
|
Fluorophores and quenchers | Re-emit/absorb light and emit heat | Alexa Fluor, Cy3, FAM, and Rhodamine Red dyes |
|
To manufacture products according to your specifications, leverage our industry-leading expertise and infrastructure as a comprehensive third-party OEM service. We provide kitting, labeling, and commercialization services for oligos. As a company providing thousands of products to customers globally, Thermo Fisher Scientific has developed expertise in supply chain management, product manufacturing, and order fulfillment. As a vertically integrated organization, we control reagent quality throughout the nucleic acid synthesis process which reduces batch-to-batch variability of oligos.
To inquire about OEM oligos: Contact us
Learn more: OEM & commercial supply services
Thermo Fisher Scientific’s oligo manufacturing suite in Pleasanton, CA is powered by more than 100 years of combined experience. We offer dedicated ordering and fulfillment services that are integrated into your organization's systems and processes. Our capabilities extend to the production of oligonucleotide components for in vitro diagnostics (IVDs) and analyte-specific reagents (ASRs).
Our oligo laboratories operate under ISO 9001 and ISO 13485 certifications and are maintained under our quality management systems. We use a scalable quality approach that is customized to your needs and requirements and provides traceability and good documentation practices, including certificates of analysis, to help ensure consistent quality and safety downstream.
Proposal |
---|
|
Development support | Process development |
---|---|
|
|
Custom manufacturing | Support | Distribution |
---|---|---|
|
|
|
Proposal |
---|
|
Development support | Process development |
---|---|
|
|
Custom manufacturing | Support | Distribution |
---|---|---|
|
|
|
* Process may be modified on a project-by-project basis.