Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
So far, vasopressin is rarely measured because of technical difficulties and the limited reliability of available assays. That's why vasopressin measurement is not adequately implemented in diagnostic pathways. With the vasopressin surrogate, Copeptin proAVP , new possibilities open up, and the full range of the hormone's diagnostic potential can be used. Copeptin proAVP shows a significant correlation with vasopressin; it is stable ex vivo and performs precise measurements. In addition, it is easy to manage with results available in less than one hour.
Informational: This product is not available for purchase by the general public
The availability of products may vary in each country depending on local regulatory requirements.
Contact your local specialist for more details