Cleaning and Disinfection Wipes
Equipment and surface infection control
Explore disinfection wipes range
PDI is helping your fight against preventable infections
Explore our PDI Sani-Cloth® ready to use wipes. Here at Thermo Fisher Scientific we are focused on providing our customers with the right wipe for your infection control solutions.  
  Informational: This product is not available for purchase by the general public
Our newest additions to the PDI range
Sani-Cloth® AF (Alcohol – Free)
Sani-Cloth® AF is a ready to use, alcohol-free, quaternary ammonium wipe for non-invasive medical devices and general surfaces. Ideally used for routine cleaning and disinfecting in high traffic areas, high touch point surfaces and non-invasive medical devices.
Sani-Cloth AF pre moistened wipes are effective against Human Corona virus and Norovirus in 30 seconds.
Super Sani-Cloth® 
Super Sani-Cloth is a ready to use, two-in-one dual quaternary ammonium and alcohol-based wipe, ideal to use in fast-paced environments, on non-invasive medical devices, non-porous surfaces. Super Sani-Cloth is effective against Human Corona virus and Norovirus in as little as 60 seconds.
Wipe out infections without wiping out your equipment. View the medical equipment compatibility guide to identify if Super Sani-Cloth is compatible with your medical device.
It’s time to trial a new 2 in 1 wipe, let’s start with a free sample
With the brand-new Sani-Cloth AF and Super Sani Cloth to our Australian and New Zealand markets, now is the time to start with a sample.
This could be the alternative that you are looking for. Start with a sample, and our sales representative will seek your feedback. 
Wipe selection guide
At Thermo Fisher Scientific, we offer a comprehensive range of ready to use wipes. From hospitals to pathology clinics, our range offers comprehensive solutions. We understand that each environment has specific needs, view the range and the quick selection guide to see which PDI wipe solution would work best for you. 
Quick selection guide
Cleaning and disinfecting surfaces and non-invasive medical devices:
  • Sani-Cloth AF, 2 in 1 wipe (Alcohol-free, Quaternary Ammonium)
  • Sani-Cloth Super Sani, 2 in 1 wipe (Alcohol and Quaternary Ammonium)
  • Sani-Cloth 70% alcohol wipe (70% isopropanol alcohol)
  • Sani-Cloth Chlor sporicidal wipe (hypochlorite)
Cleaning and damp dusting of all general surfaces and equipment :
  • Sani-Cloth detergent wipe (detergent only) 
Explore the rest of the range
Sani-Cloth® 70 is the multi-purpose solution, ideal for disinfection.
  • Ready to use: pre-dosed, low linting disposable alcohol disinfection wipes, requiring no mixing or measuring, with no smear formulation.
  • Proven efficacy: effective against bacteria, yeast and enveloped viruses.
  • Improved efficiencies: quick drying, no residue formulation containing 70% Isopropanol alcohol allowing the disinfected area or equipment to be used shortly after application. 
  • Maximum flexibility: can be used on non-invasive medical devices; non-porous hard surfaces. 
  • Recommended usage: as part of a two-step process with Sani-Cloth® Detergent to clean prior to disinfection. Where surfaces are soiled, pre-cleaning is required.
Sani-Cloth® Chlor is a sporicidal cleaning and disinfectant solution ideal for use in high risk, contaminated environments.
  • Ready to use: pre-dosed, low linting disposable cleaning and disinfection wipe, requiring no mixing or measuring.
  • Proven efficacy: effective against all spores, including C.difficle and fully virucidal in 60 seconds.
  • Improved efficiencies: the product solution is impregnated with sodium hypochlorite and surfactant to clean and disinfect providing you with consistently reliable results and significant time savings delivering approximately 6000 ppm available chlorine.
  • Optimal protection: can be used on non-invasive medical devices; non-porous hard surfaces with supporting compatibility data.
  • Cost effective: using the same solution for cleaning and disinfecting saves you time and money.
Sani-Cloth® Detergent wipes are impregnated with a low-level detergent for the cleaning & dampdusting of all general surfaces and equipment, including beds and mattresses. They contain aclean, fresh fragrance with odour-neutralising properties, utilising a low-smear, biodegradable formulation.
Recommended uses:
  • Routine cleaning procedures
  • Prior to disinfecting in a two-step cleaning and disinfecting process