Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Medical procedures and diagnostic laboratory tests generate medical waste that requires special handling and disposal. Reduce costs, and keep staff safe with on-site solutions for autoclave- and microwave-based medical waste treatment and disposal.
The medical autoclave line is designed to service hospitals, day clinics as well as medical centers that need to process small to large volumes of medical waste safely and on site. Tuttnauer provides autoclaves that satisfy the needs of any hospital operating room, central sterile services department or medical clinic.
Healthcare activities protect and save lives, but what about the waste and by-products that they generate? Biomedical waste management can be expensive, and managing the potential risks associated with the prevention of infection, sharps-related hazards, and the environmental impact of treatment and disposal can be a major challenge.
The Sterilwave® instrument is the alternative on-site solution to incineration and landfills. This method reduces the risk of contamination and is recommended by the World Health Organisation (WHO).
Sterilwave 100 |
Sterilwave 250 |
Sterilwave 440 |
|
Particularly suited to medical facilities generating only small amounts of waste. Ultra-compact and comes in a mobile version, for use in a container for example. | Allows medium-sized institutions to treat their biomedical waste, guaranteeing safety and traceability at greatly reduced operating costs. | The ideal treatment solution for institutions producing a large quantity of biomedical waste, with minimal operating costs. | |
Waste capacity | 200 L/h–up to 20 kg/h (with a density of 1 L = 0.1 kg) | 500 L/h–up to 50 kg/h (with a density of 1 L = 0.1 kg) | 880 L/h–up to 70 kg/h (with a density of 1 L = 0.1 kg) |
Vessel capacity | 100 L | 250 L | 440 L |
Average cycle time | 30 minutes | 30 minutes | 30 – 32 minutes |
Dimensions ( L x W x D ) | 1.7 x 1.2 x 1.1 m | 1.6 x 1.1 x 1.5 m | 2 .0 x 1.1 x 2.0 m |
Weight | 700 kg | 1,200 kg | 1,400 kg |
Recommended working area | 10 sq m Ceiling height 2 m |
10 sq m (full workflow area) Ceiling height 2.2 m |
10 sq m (full workflow area) Ceiling height 3 m (3.85 m with automatic loading option) |
Electricity | 400 V/ 3-Phase, 40 A | 400 V/ 3-Phase, 63 A | 400 V/ 3-Phase, 63 A |
Technology | Grinding by rotating blades and heating by microwaves | ||
Volume reduction | Up to 80% | ||
Weight reduction | Up to 25% |