Our techs are here to answer your questions, just email us.
Ask our techs ›
If you like this month's Q&A, check out our Talk Tech archive.
Question
Answer
There are several possible reasons that the Thermo Scientific™ Model 5030i SHARP Monitor or Thermo Scientific™ 5014i Beta Continuous Ambient Particulate Monitor would throw a "Filter tape change alarm."  
First, check to make sure the unit has not run out of tape.  To order replacement tape call 1 (866) 282-0430, option 1 and order part number 106443-00. If the unit has tape, troubleshoot using the below guide.  For troubleshooting purposes, the possible issues are in order of most likely cause.
talk-tech-april-5030i
Filter wheel configuration
Possible issue #1
Filter tape take up spindle (A) is turning in hub of filter tape spool (B) but doesn’t turn filter tape.      
Solution:  Reel hub nut on the end of take up spindle is too loose and needs to be tightened.
Possible issue #2
There is excess slack in filter tape from the filter tape supply hub (C). While performing an automatic filter tape advance, the filter tape counter wheel (D) did not detect movement of the counter wheel.
Solution:  Perform a manual filter tape advance until the counter wheel (right side under the filter tape supply reel) senses movement of the counter wheel.
Possible issue #3
The counter wheel sensor is defective or dirty (E).
Solution:  Use compressed air to clean sensor wheel with detection window is located. Ensure contacts on sensor pcb are clean and void of corrosion.  Ensure the sensor harness is properly connected. Replace sensor.  To order a new sensor, call 1 (866) 282-0430, option 1 and order part number 105938-00.
Possible issue #4
Defective tape advance motor assembly.  Motor doesn’t operate or turn freely during a filter tape advance.
Solution:  Replace the defective motor.   To order a new motor, call 1 (866) 282-0430, option 1 and order part number 106032-00.
tech-talk-showcase-image-230x195
Don't miss out. Subscribe today.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.
Subscribe to Talk Tech ›