Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Blackline Safety is a leader in connected safety monitoring, with innovative solutions to support lone workers, evacuation management, and your environmental gas detection needs. The safety monitoring solutions deliver instant awareness and connectivity for every industry, location, job, and scenario.
The Blackline Safety G7 lone worker device wirelessly connects your team to real people. Blackline puts the tools in your hands to watch over your own team, or you can leave the monitoring to the Blackline in-house 24/7 Safety Operations Center (SOC).
Employees who face an incident are under incredible pressure. Even if they can’t respond to a two-way voice call or text message, G7 brings comfort when its blue LiveResponse™ light turns on—which means the alert was received, and help is on the way.
For situations where a threat needs to be communicated broadly to your team, monitoring personnel can mass-notify your choice of personnel at one or more sites. Confidently account for everyone’s well-being and whereabouts in real-time, and be equipped to act when someone needs a hand.
The G7 is completely customisable with an exclusive modular design to accommodate diverse gas detection and safety monitoring needs.
Blackline G7c |
Blackline G7x and G7 Bridge |
---|---|
Lone worker monitoring, with fall detection, no motion detection, SOS alert, worker check-in | Lone worker monitoring, with fall detection, no motion detection, SOS alert, worker check-in |
Two-way speakerphone or text messaging for worker contact | Text messaging only, to worker |
Connectivity via 3G/4G cellular network | Connectivity via 3G/4G cellular network or satellite via G7 Bridge, with automatic switchover |
Rechargeable battery, with up to 18 hr run-time |
Rechargeable battery, with up to 18 hr run-time, for G7x and 44 hrs for G7 Bridge |
GPS location tracking | GPS location tracking, within cellular range |
Push-to-Talk option turns a G7c into a two-radio, for communication with other G7c users | -- |
Optional integral gas sensors, for up to 5 gases, pumped or diffusion, with lifetime sensor warranty, for a fully connected worker solution | Optional integral gas sensors, for up to 5 gases, pumped or diffusion, with lifetime sensor warranty, for a fully connected worker solution |
Choose gas sensors for toxic gases, oxygen, and combustible (LEL) gases; PID detector for VOCs; IR detector for flammable gases | Choose gas sensors for toxic gases, oxygen, and combustible (LEL) gases; PID detector for VOCs; IR detector for flammable gases |
In certain facilities, GPS signals alone may not provide sufficient positioning precision to locate employees during an emergency response. In such circumstances, every second counts, and time wasted searching for someone could mean the difference between rescue and recovery.
Location Beacons are compact wireless positioning transmitters that are installed where employees operate, from one per floor to one per room, depending on facility size and operations. Beacon locations are then recorded on custom floor or site plans on a live map in Blackline Live, Blackline's safety monitoring software, so you always know exactly where your people are in the context of their surroundings. Beacons are self-powered and affordable, taking only minutes to install without any calibration or triangulation. When personnel are within proximity of beacons, the G7 devices store the nearby beacon identities and report their location to Blackline Live.
Blackline Safety’s G7 Dock is the simple solution to easily calibrating, bump testing, and charging G7 gas monitoring devices. Docks can be used with both G7c and G7x devices and supports G7 Single-gas and Quad-gas cartridges.
Bump tests and calibrations are straightforward, performed by simply placing G7 into the dock and using the inserted G7 device's LCD screen and menu system. If a G7 device is due for a bump test and/or calibration, it will automatically perform the required test actions when placed into the dock and the lid closed. All test results are communicated in real-time to the user's Blackline Live account after test actions are complete.
Blackline's safety monitoring and gas detection solutions deliver complete customisation and 24/7 real-time safety monitoring for any application.
The Blackline Live web portal provides businesses self-monitoring and situational awareness of their workers, in real-time, allowing them to respond quickly to emergency situations.
Alternatively, this service may be carried out by Blackline’s Safety Operations Center (SOC), focusing exclusively on monitoring the well-being of employees' safety. Unlike traditional call centres that often split focus between telemarketing and IT support, the SOC is staffed with professionally-trained monitoring agents 365 days a year.