Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Portable gas detectors are ideal for confined spaces, spot leak testing, and mobile use, marked by flexibility and quality. A variety of single- or multi-gas detectors are offered in compact, lightweight designs—from simple, alarm-only units to advanced, fully configurable, and serviceable instruments.
Applications include underground utilities, vaults, boiler rooms, post-fire sites, sewers, industrial plants, industrial hygiene, first responder crews, and remote fleets.
Single gas detectors are portable gas detectors that only have the ability of detecting a single gas threat in a variety of applications and environments, from CO2 in wineries and brewing, to phosphine fumigation in agriculture, to ammonia in refrigeration and wastewater, and many more.
Honeywell BW™ Solo |
MSA ALTAIR® 2X |
Honeywell
|
Blackline G7
|
Blackline G6 |
---|---|---|---|---|
BW™ Solo is a next generation, easy to service single-gas detector with BLE connectivity and 3-year warranty. | Rugged single or dual gas detector, with 3-year warranty. | Wireless single-gas detector for a large range of toxic gases, VOCs, and combustible gases; with man-down alarm. | Cloud connected single gas detector with optional real-time lone-worker alerts and GPS positioning. | Cloud connected single gas detector with 4-year data access and warranty and GPS positioning. |
15 gases to choose from–common to exotic, H2S to CO2 to HCN and more. | 6 gases to choose from common to exotic: H2S to CO to NH3 and more, as single gas or two-gas | 17 gases to choose from–VOCs (PID) to CO to NO2 and more | 17 gases to choose from VOCs (PID) to CO to NO2 and more. | 4 gases to choose from CO, H2S, O2, or SO2 |
One button operation | One button operation | Two button operation | Three button operation | Three button operation |
Easy battery and sensor replacement | Replaceable battery with 6-12 month life | Re-chargeable battery | Re-chargeable battery | Re-chargeable battery, 1-yr operation |
1-series sensors for fast response and long life | Long-life, fast XCell Technology with end-of-life indicator | Over 20 intelligent field-replaceable electrochemical sensors |
Field replaceable sensor cartridge for fast swap-out | Large graphical, colour display |
Standard, BLE, and Lite versions | Charcoal and glow-in-the-dark versions | Standard, wireless, and Lite versions | Options for cellular and satellite cloud connection, real-time alerts and push-to-talk | Over-the-air updates, GPS location technology |
Dock: IntelliDoX | Dock: Galaxy GX2 | Dock: AutoRAE 2 | Dock: G7 Dock | Dock: G6 Dock |
BW™ Solo is a next generation, easy to service single-gas detector with BLE connectivity and 3-year warranty.
When monitoring the atmosphere in a confined space, there are several important issues that need to be considered, reviewed, and managed. One of the major issues centers on air quality and what you are breathing, both prior to entry and during occupation of a confined space. You need to know what the oxygen content of the atmosphere is and whether there are explosive or toxic gases that could threaten the safety of the environment or your life. The standard four confined space gases are Combustible gas %LEL, Oxygen (O2), Carbon Monoxide (CO), and Hydrogen Sulfide (H2S).
MSA ALTAIR® 4XR |
Honeywell BW™ ICON/ICON+ |
Honeywell BW™ Flex4 |
Blackline G7 |
---|---|---|---|
Rugged 4-gas detector with 4 or 5-year warranty and man-down alerts. | 4-gas detector with simple icon display and 2-year (Icon) or 3-year (Icon+) warranty. | 4-gas detector with full display and 3-year warranty. | Cloud connected 4-gas detector with optional real-time lone-worker alerts and GPS positioning. |
6 gases to choose from: standard Quad-Gas, SO2 and NO2 | 6 gases to choose from: standard Quad-Gas and SO2 | 15 gases to choose from: standard Quad-Gas, to NH3 and SO2 | 17 gases to choose from VOCs (PID) to CO to NO2 and more, pumped or diffusion |
Two button operation | One button operation | One button operation | Three button operation |
Rechargeable Li-polymer battery, 24-hour run-time | Rechargeable Li-polymer battery, 60-day run-time | Rechargeable Li-polymer battery, 60-day run-time | Re-chargeable battery |
Long-life, fast XCell Technology with end-of-life indicator | 1-series sensors for fast response and long life. Intelligent sensors for predictive sensor health (Icon+) | 1-series sensors for fast response and long life. Intelligent sensors for predictive sensor health |
Field replaceable sensor cartridge for fast swap-out, pumped or diffusion |
Charcoal and glow-in-the-dark versions. Bluetooth connectivity | Always-ON option. Bluetooth connectivity. Basic (Icon) or serviceable (Icon+) | Bluetooth connectivity. Serviceable |
Options for cellular and satellite cloud connection, real-time alerts and push-to-talk |
Dock: Galaxy GX2 | Dock: IntelliDoX | Dock: IntelliDoX | Dock: G7 Dock |
Also available, standard quad-gas detectors:
A range of portable gas detectors monitoring the standard gases for confined space gases entry plus a number of toxic gases for everyday use.
Honeywell BW™ Ultra |
MSA ALTAIR® 5XR |
Honeywell MultiRAE |
Honeywell BW™ Flex5 |
---|---|---|---|
BW™ Ultra is a next-generation five-gas detector with smart digital sensors, designed specifically for sampling and monitoring confined spaces. | Rugged 6-gas detector with up to 3-year warranty and man-down alerts. | MultiRAE family offers wireless multi-gas detection for up to 5 gases plus volatile organic compounds, with man-down alarm. | 5-gas detector with full display and 3-year warranty. |
Standard 4-gases plus one sensor for toxic gas or VOCs (PID) | 16 gases to choose from VOCs (PID) to standard Quad-Gas, SO2 and NO2 and more | 17 gases to choose from VOCs (PID) to CO to NO2 and more | 15 gases to choose from: standard Quad-Gas, to NH3 and SO2 |
One button operation | Two button operation | Two button operation | One button operation |
Re-chargeable battery, 18-hour run-time | Rechargeable Li-polymer battery, 20-hour run-time, or alkaline battery | Re-chargeable battery, 12-hour run-time | Rechargeable Li-polymer battery, 60-day run-time |
1-series sensors for fast response and long life. Integral pump | Long-life, fast XCell Technology with end-of-life indicator. Integral pump | Integral pump option | 1-series sensors for fast response and long life. Intelligent sensors for predictive sensor health |
Integral Bluetooth | Options of monochrome or colour display. Bluetooth connectivity | Standard or wireless versions | Bluetooth connectivity. Serviceable |
Dock: IntelliDoX | Dock: Galaxy GX2 | Dock: AutoRAE 2 | Dock: IntelliDoX |
Also available:
Honeywell BW™ Micro 5 and 5 IR for 5 gas detector is a flexible gas monitor 1 to 5 gases, with alkaline or re-chargeable battery pack
Gastec endeavours to achieve highest quality detector tubes for analysing airborne gases and vapours as well as pollutants in soil and water through advanced state-of-the-art research and development.
Colorimetric tubes |
Sampling pump |
---|---|
Gastec Detector Tubes indicate concentrations directly by way of a calibrated scale printed on the tubes. Tubes are available for more than 500 different applications. |
The Gastec Sampling Pump Set for short-term measurements comes in two models, GV-100 and GV-110, with the GV110 having the automatic stroke counter built in. |
Honeywell
|
MSA GALAXY®
|
Honeywell
|
Blackline
|
Blackline
|
---|---|---|---|---|
Wall/bench mount | Wall/bench mount, with gas cylinder holder | Wall/bench mount | Wall/bench mount | Bench mount |
Fast calibration and fast bump test solution avoids gas wastage | Fast calibration and fast bump test solution avoids gas wastage. Auto-detector of calibration gas and printer option | Easy, one-touch bump testing, calibration, charging, and records management | Cloud connected, bump testing, calibration, charging and records management | Cloud connected, bump testing, calibration, charging and records management |
Up to 5 docking modules per base station | Test up to 10 gas detectors, simultaneously | Up to 10 docking cradles per system | Up to 5 docking modules per system | Standalone system for 1 gas detector |
Used with BW™ Clip, Solo, Clip 4, Ultra, Max XT II, MicroClip XL/X3, Icon, Icon+, Flex4, Flex5 | Used with Altair® series of gas detectors | Used with ToxiRAE Pro, MultiRAE | Used with Blackline G7c and G7x | Used with Blackline G6 |
Also available:
Honeywell MicroDock II used with BW Quattro, Micro 5, Max XT II, and MicroClip XL.X3