Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Bearing health is critical to highly reliable production. Early detection of any unusual changes in bearing and gear health is important to production efficiency and reduction of downtime. Our selection of smart sensors can be readily integrated into your plant and schedule for continuous monitoring of your system’s health.
Be informed with early warning of faults in rotating elements of bearings and gears
Bearing faults create metal-to-metal impacts that manifest in vibration monitoring as high frequency, high amplitude peaks. Bearing fault detectors can accurately identify these peaks in ways vibration transmitters cannot, with their RMS process smoothing the shock impacts caused by spalling, cracking, and lubrication problems.
Bearing Fault Detector | Fluke 805 Vibration Meter |
---|---|
The Bearing Fault Detector is a DIN rail mount vibration transmitter (for use with ICP sensor) with dual 4–20 mA outputs for overall and peak vibrations. | The Fluke 805 Vibration Meter screening device is available for frontline mechanical troubleshooting teams that need repeatable, severity-scaled readings of overall vibration, bearing health, and machine health. |
Output range 4–20 mA | - |
Frequency Response: (Overall Vibration) (±3 dB) 10 to 1 kHz (10 to 1 kHz) | - |
Supply Voltage: 20 to 28 VDC | - |
Request information | Fluke 805 Vibration Meter |