Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Whether you’re in an industrial plant, manufacturing facility, or a utility, Fluke power quality and energy management tools are designed to help identify your power quality and energy efficiency issues with easy and effective recording and analysis. Flexible and powerful software is also included with every Fluke Power Quality and Energy Analysis tool, allowing you visibility to valuable insights about the performance of your electrical system.
Power and energy loggers are used for conducting three-phase energy and load studies to discover where savings are possible. With Fluke Energy Analyze Plus software, it is possible to create detailed reports to focus in on the problem areas.
ScopeMeter® Portable Oscilloscopes take you into territory that standard bench oscilloscopes cannot readily withstand, such as harsh, hazardous, and dirty industrial environments. These hand-held instruments combine the performance of a bench oscilloscope with a multimeter and paperless recorder for installing, commissioning, and maintaining industrial and electronic equipment out in the field. Fluke ScopeMeter test tools use a unique Connect-and-View™ mode to automatically and continuously setup the oscilloscope, making them easy to use even on complex signals.
Fluke instruments have the highest accuracy and are designed to handle the most difficult waveforms, whether testing the performance of transformers, lighting, or switching electronics such as inverters or power supplies. For very low power factor or high-frequency waveforms, Fluke has solutions that can measure on single-phase, three-phase, or six-phase systems.
Our precision power quality analysers will perform 'gap-less' measurements and capture all details of every power cycle during measurement and logging; this capability is key to evaluating the health of your power system. These are easy and reliable for use in the field or as a bench unit in laboratories and on test benches. These high-precision analysers will assist you in the development of the best power electronics and will help improve the efficiency of your products.