Empowering you with technology to achieve more today and be ready to master future challenges, triple quadrupole ICP-MS helps ensure that your laboratory can meet evolving legislation requirements, enables you to explore developing markets, and pushes the boundaries of your research.
Whether you are seeking better data quality by reducing the number of false positive results or looking for a new system to take on tomorrow's challenges, there is a dedicated solution available for every laboratory. Harness the superior interference removal capabilities of triple quadrupole technology without compromising detection power.
The Thermo Scientific iCAP TQ ICP-MS is the solution to future-proofing your laboratory: explore developing markets, push the boundaries of your research and meet the requirements of evolving legislation.
  • ‘Right-first-time’ analysis with powerful interference removal, even in the toughest matrices.
  • Reduced re-runs in the daily workload with assured accuracy and repeatability.
  • Maintains the flexibility of single quadrupole (SQ) mode for less challenging analyses.
We understand the challenges you face when analyzing complex sample matrices, which inspired us to redefine triple quadrupole technology with unique ease of use. Find out how the iCAP TQ ICP-MS, which includes triple quadrupole technology, significantly improves interference removal.
Unique ease-of-use 
  • Combining user-inspired hardware with intelligent software, our triple quadrupole ICP-MS systems enable laboratories to easily develop and maintain methods that ensure confidence in data quality. 
  • Reaction Finder, the systems’ unique method development assistant, allows you to tackle challenging matrices without wasting time on complex method development
  • For ultimate flexibility, the iCAP TQ ICP-MS enables you to easily switch from SQ to TQ mode in a single sample analysis, providing the best results for your complete suite of elements.
'Right-first-time' results 
  • Triple quadrupole technology enables you to analyze complex matrices with superior limits of detection due to its advanced interference removal capabilities. Our TQ-ICP-MS systems provide uncomplicated analyses with superior accuracy for both research and routine applications.
Learn how the iSC-65 Autosampler offers exceptional performance in streamlining workflows and sample throughput.
The Thermo Scientific iSC-65 Autosampler has been designed with reliability from the ground up to ensure all samples are analyzed the first time.
  • Efficiency and productivity: The iSC-65 Autosampler allows sample-specific probe depths, avoiding unnecessary reruns due to blocked sample introduction systems. With the advanced Step Ahead feature, the autosampler probe is sent to rinse while data is still being acquired, using sample in the probe line to complete the analysis. 
  • Durable and robust design: Acid-resistant construction ensures confident sampling as well as longevity and robust operation in laboratory environments.
  • Visual feedback: Through LED lighting, provides clear, easy-to-see visual feedback at a distance on autosampler and analysis run status for technicians in crowded laboratories.
  • Software integration: Native Qtegra ISDS Software plugin allows user to control all automation parameters, including alignment, individual depth locations, pump speed, XYZ movement speeds, and direction of sampling sequences.
  • Unattended analyses: With the advanced quality control capabilities in Qtegra ISDS Software, the iSC-65 Autosampler instills confidence in high throughout, unattended ICP-OES and ICP-MS analyses.
  • Low contamination sample introduction: Utilize the iSC-65 Autosampler with the Thermo Scientific iCAP Qnova Series ICP-MS to eliminate manual sample introduction and the potential of contamination when analyzing ultra-pure high sensitivity samples.
Featured videos
Application Notes
Brochures and Specifications
* Required field
Style Sheet for Global Design System
CMD SchemaApp code